Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105209198-105209688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRF1
Entrez Gene ID
2972
Gene Name
BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
Gene Aliases
BRF, BRF-1, CFDS, GTF3B, HEL-S-76p, TAF3B2, TAF3C, TAFIII90, TF3B90, TFIIIB90, hBRF
UniGene
Hs.424484
Gene Chromosome Location
Chr.14: 105209286-105315577 on Build GRCh38
Transcripts
NM_001242786.1 NM_001242787.1 NM_001242788.1 NM_001242789.1 NM_001519.3 NM_145685.2 XM_005267561.3 XM_005267563.3 XM_006720123.2 XM_011536673.1 XM_011536674.1

SNP Detail

SNP ID Chromosome Location
rs1031027245 Chr.14: 105209253-105209259 on Build GRCh38
rs923356405 Chr.14: 105209610-105209610 on Build GRCh38
rs1031058243 Chr.14: 105209437-105209437 on Build GRCh38
rs1028842900 Chr.14: 105209397-105209410 on Build GRCh38
rs1010935008 Chr.14: 105209658-105209658 on Build GRCh38
rs778912742 Chr.14: 105209516-105209516 on Build GRCh38
rs762964707 Chr.14: 105209524-105209524 on Build GRCh38
rs113551076 Chr.14: 105209358-105209358 on Build GRCh38
rs763965388 Chr.14: 105209641-105209641 on Build GRCh38
rs149073034 Chr.14: 105209559-105209559 on Build GRCh38
rs183332136 Chr.14: 105209356-105209356 on Build GRCh38
rs887935960 Chr.14: 105209628-105209628 on Build GRCh38
rs756285929 Chr.14: 105209482-105209482 on Build GRCh38
rs1007630076 Chr.14: 105209233-105209233 on Build GRCh38
rs979341137 Chr.14: 105209580-105209580 on Build GRCh38
rs865864321 Chr.14: 105209555-105209555 on Build GRCh38
rs34745426 Chr.14: 105209465-105209465 on Build GRCh38
rs776248945 Chr.14: 105209655-105209655 on Build GRCh38
rs968989406 Chr.14: 105209509-105209509 on Build GRCh38
rs587715509 Chr.14: 105209240-105209240 on Build GRCh38
rs587690785 Chr.14: 105209570-105209570 on Build GRCh38
rs587748226 Chr.14: 105209579-105209579 on Build GRCh38
rs938486768 Chr.14: 105209506-105209506 on Build GRCh38
rs189190499 Chr.14: 105209535-105209535 on Build GRCh38
rs587672069 Chr.14: 105209264-105209265 on Build GRCh38
rs1026532098 Chr.14: 105209241-105209241 on Build GRCh38
rs1008987568 Chr.14: 105209427-105209427 on Build GRCh38
rs746339146 Chr.14: 105209235-105209235 on Build GRCh38
rs1006413641 Chr.14: 105209443-105209443 on Build GRCh38
rs772132321 Chr.14: 105209584-105209584 on Build GRCh38
rs587760506 Chr.14: 105209551-105209551 on Build GRCh38
rs757319706 Chr.14: 105209396-105209397 on Build GRCh38
rs1040419996 Chr.14: 105209388-105209388 on Build GRCh38
rs753996882 Chr.14: 105209624-105209624 on Build GRCh38
rs118016083 Chr.14: 105209609-105209609 on Build GRCh38
rs758769277 Chr.14: 105209602-105209602 on Build GRCh38
rs587655620 Chr.14: 105209547-105209547 on Build GRCh38
rs757507809 Chr.14: 105209632-105209632 on Build GRCh38
rs763922838 Chr.14: 105209466-105209466 on Build GRCh38
rs587722335 Chr.14: 105209390-105209390 on Build GRCh38
rs1016586998 Chr.14: 105209450-105209450 on Build GRCh38
rs587697474 Chr.14: 105209255-105209255 on Build GRCh38
rs879012851 Chr.14: 105209548-105209548 on Build GRCh38
rs8270 Chr.14: 105209364-105209364 on Build GRCh38
rs951062953 Chr.14: 105209250-105209250 on Build GRCh38
rs1021563599 Chr.14: 105209451-105209451 on Build GRCh38
rs988016798 Chr.14: 105209540-105209540 on Build GRCh38
rs587652391 Chr.14: 105209441-105209441 on Build GRCh38
rs914464523 Chr.14: 105209256-105209256 on Build GRCh38
rs587748065 Chr.14: 105209252-105209252 on Build GRCh38
rs747292268 Chr.14: 105209272-105209272 on Build GRCh38
rs776087865 Chr.14: 105209347-105209347 on Build GRCh38
rs1047911466 Chr.14: 105209625-105209625 on Build GRCh38
rs757273565 Chr.14: 105209222-105209222 on Build GRCh38
rs943487929 Chr.14: 105209382-105209382 on Build GRCh38
rs772889932 Chr.14: 105209487-105209487 on Build GRCh38
rs1048464624 Chr.14: 105209423-105209423 on Build GRCh38
rs587601398 Chr.14: 105209416-105209416 on Build GRCh38
rs770464203 Chr.14: 105209650-105209650 on Build GRCh38
rs927073328 Chr.14: 105209293-105209293 on Build GRCh38
rs750677399 Chr.14: 105209478-105209478 on Build GRCh38
rs779026564 Chr.14: 105209229-105209229 on Build GRCh38
rs983727193 Chr.14: 105209251-105209251 on Build GRCh38
rs769067856 Chr.14: 105209325-105209325 on Build GRCh38
rs113283999 Chr.14: 105209275-105209275 on Build GRCh38
rs775957397 Chr.14: 105209652-105209653 on Build GRCh38
rs994157354 Chr.14: 105209239-105209239 on Build GRCh38
rs759189237 Chr.14: 105209603-105209603 on Build GRCh38
rs913724545 Chr.14: 105209558-105209558 on Build GRCh38
rs1050628245 Chr.14: 105209583-105209583 on Build GRCh38
rs927792034 Chr.14: 105209286-105209286 on Build GRCh38
rs779421036 Chr.14: 105209560-105209560 on Build GRCh38
rs775564399 Chr.14: 105209486-105209486 on Build GRCh38
rs587702609 Chr.14: 105209657-105209657 on Build GRCh38
rs587649434 Chr.14: 105209284-105209284 on Build GRCh38
rs913798093 Chr.14: 105209332-105209332 on Build GRCh38
rs1037998515 Chr.14: 105209365-105209365 on Build GRCh38
rs902007635 Chr.14: 105209238-105209238 on Build GRCh38
rs902016300 Chr.14: 105209392-105209392 on Build GRCh38
rs587625876 Chr.14: 105209254-105209254 on Build GRCh38
rs1044594506 Chr.14: 105209649-105209649 on Build GRCh38
rs927145753 Chr.14: 105209505-105209505 on Build GRCh38
rs993651123 Chr.14: 105209401-105209401 on Build GRCh38
rs939166063 Chr.14: 105209306-105209306 on Build GRCh38
rs587607390 Chr.14: 105209538-105209538 on Build GRCh38

View Full Product Details