Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105209992-105210495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRF1
Entrez Gene ID
2972
Gene Name
BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
Gene Aliases
BRF, BRF-1, CFDS, GTF3B, HEL-S-76p, TAF3B2, TAF3C, TAFIII90, TF3B90, TFIIIB90, hBRF
UniGene
Hs.424484
Gene Chromosome Location
Chr.14: 105209286-105315577 on Build GRCh38
Transcripts
NM_001242786.1 NM_001242787.1 NM_001242788.1 NM_001242789.1 NM_001519.3 NM_145685.2 XM_005267561.3 XM_005267563.3 XM_006720123.2 XM_011536673.1 XM_011536674.1

SNP Detail

SNP ID Chromosome Location
rs1040172899 Chr.14: 105210419-105210419 on Build GRCh38
rs1038803843 Chr.14: 105210392-105210392 on Build GRCh38
rs1027501675 Chr.14: 105210188-105210188 on Build GRCh38
rs770481474 Chr.14: 105210074-105210074 on Build GRCh38
rs963309204 Chr.14: 105210265-105210265 on Build GRCh38
rs930593320 Chr.14: 105210357-105210357 on Build GRCh38
rs924931488 Chr.14: 105210165-105210165 on Build GRCh38
rs913450532 Chr.14: 105210123-105210123 on Build GRCh38
rs587676229 Chr.14: 105210411-105210411 on Build GRCh38
rs1007389576 Chr.14: 105210375-105210375 on Build GRCh38
rs924941304 Chr.14: 105210321-105210321 on Build GRCh38
rs917998369 Chr.14: 105210311-105210311 on Build GRCh38
rs1018637917 Chr.14: 105210071-105210071 on Build GRCh38
rs1049408292 Chr.14: 105210358-105210358 on Build GRCh38
rs765401407 Chr.14: 105210276-105210276 on Build GRCh38
rs587599546 Chr.14: 105210409-105210409 on Build GRCh38
rs111764937 Chr.14: 105210298-105210298 on Build GRCh38
rs950739768 Chr.14: 105210050-105210050 on Build GRCh38
rs587610579 Chr.14: 105210451-105210451 on Build GRCh38
rs985846722 Chr.14: 105210070-105210070 on Build GRCh38
rs587628139 Chr.14: 105210128-105210128 on Build GRCh38
rs1016259510 Chr.14: 105210257-105210257 on Build GRCh38
rs587625087 Chr.14: 105210044-105210044 on Build GRCh38
rs113861810 Chr.14: 105210045-105210045 on Build GRCh38
rs897721955 Chr.14: 105210171-105210171 on Build GRCh38
rs980155276 Chr.14: 105210099-105210099 on Build GRCh38
rs774099871 Chr.14: 105210093-105210093 on Build GRCh38
rs1051588195 Chr.14: 105210228-105210228 on Build GRCh38
rs587776133 Chr.14: 105210284-105210284 on Build GRCh38
rs587746719 Chr.14: 105210100-105210100 on Build GRCh38
rs587763190 Chr.14: 105210191-105210191 on Build GRCh38
rs1004749629 Chr.14: 105210238-105210238 on Build GRCh38
rs191266777 Chr.14: 105210021-105210021 on Build GRCh38
rs35017042 Chr.14: 105210306-105210306 on Build GRCh38
rs146924498 Chr.14: 105210256-105210256 on Build GRCh38
rs965734460 Chr.14: 105210230-105210230 on Build GRCh38
rs139395472 Chr.14: 105210332-105210332 on Build GRCh38
rs587703701 Chr.14: 105210430-105210430 on Build GRCh38
rs965661449 Chr.14: 105210091-105210091 on Build GRCh38
rs1036726615 Chr.14: 105210155-105210155 on Build GRCh38
rs936286020 Chr.14: 105210378-105210378 on Build GRCh38
rs1047002 Chr.14: 105210107-105210107 on Build GRCh38
rs754458444 Chr.14: 105210287-105210288 on Build GRCh38
rs888958892 Chr.14: 105210359-105210359 on Build GRCh38
rs967655217 Chr.14: 105210297-105210297 on Build GRCh38
rs896499553 Chr.14: 105210132-105210132 on Build GRCh38
rs946900127 Chr.14: 105210114-105210114 on Build GRCh38
rs374972527 Chr.14: 105210474-105210474 on Build GRCh38
rs1001635607 Chr.14: 105210267-105210267 on Build GRCh38
rs1039152824 Chr.14: 105210209-105210209 on Build GRCh38
rs886389587 Chr.14: 105210234-105210234 on Build GRCh38
rs933073323 Chr.14: 105210454-105210454 on Build GRCh38
rs376390782 Chr.14: 105210154-105210154 on Build GRCh38
rs960450997 Chr.14: 105210103-105210103 on Build GRCh38
rs587698093 Chr.14: 105210088-105210088 on Build GRCh38
rs983441121 Chr.14: 105210053-105210053 on Build GRCh38
rs995107272 Chr.14: 105210181-105210181 on Build GRCh38
rs940038065 Chr.14: 105210319-105210319 on Build GRCh38
rs750645251 Chr.14: 105210285-105210285 on Build GRCh38
rs77996637 Chr.14: 105210270-105210270 on Build GRCh38
rs775309996 Chr.14: 105210211-105210211 on Build GRCh38
rs987807626 Chr.14: 105210260-105210260 on Build GRCh38
rs116738756 Chr.14: 105210455-105210455 on Build GRCh38
rs767894908 Chr.14: 105210129-105210129 on Build GRCh38
rs77331892 Chr.14: 105210041-105210041 on Build GRCh38
rs80206469 Chr.14: 105210320-105210320 on Build GRCh38
rs993106940 Chr.14: 105210105-105210105 on Build GRCh38
rs587618223 Chr.14: 105210055-105210055 on Build GRCh38
rs926766635 Chr.14: 105210126-105210126 on Build GRCh38
rs116254029 Chr.14: 105210047-105210047 on Build GRCh38
rs997737304 Chr.14: 105210221-105210221 on Build GRCh38

View Full Product Details