Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105212021-105212525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRF1
Entrez Gene ID
2972
Gene Name
BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
Gene Aliases
BRF, BRF-1, CFDS, GTF3B, HEL-S-76p, TAF3B2, TAF3C, TAFIII90, TF3B90, TFIIIB90, hBRF
UniGene
Hs.424484
Gene Chromosome Location
Chr.14: 105209286-105315577 on Build GRCh38
Transcripts
NM_001242786.1 NM_001242787.1 NM_001242788.1 NM_001242789.1 NM_001519.3 NM_145685.2 XM_005267561.3 XM_005267563.3 XM_006720123.2 XM_011536673.1 XM_011536674.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
231853 c.1824+10C>T p.? Chr.14: 105212103-105212103 on Build GRCh38
5456423 c.1817C>T p.T606M Chr.14: 105212120-105212120 on Build GRCh38
5456424 c.1538C>T p.T513M Chr.14: 105212120-105212120 on Build GRCh38
5018887 c.1805delA p.K602fs*60 Chr.14: 105212132-105212132 on Build GRCh38
5018888 c.1526delA p.K509fs*60 Chr.14: 105212132-105212132 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747793853 Chr.14: 105212184-105212184 on Build GRCh38
rs78582404 Chr.14: 105212431-105212431 on Build GRCh38
rs749648343 Chr.14: 105212167-105212167 on Build GRCh38
rs1032338783 Chr.14: 105212409-105212409 on Build GRCh38
rs747005106 Chr.14: 105212260-105212260 on Build GRCh38
rs144088839 Chr.14: 105212144-105212144 on Build GRCh38
rs899570567 Chr.14: 105212386-105212386 on Build GRCh38
rs766255830 Chr.14: 105212198-105212198 on Build GRCh38
rs201049171 Chr.14: 105212182-105212182 on Build GRCh38
rs996569714 Chr.14: 105212386-105212387 on Build GRCh38
rs767687594 Chr.14: 105212208-105212208 on Build GRCh38
rs776580059 Chr.14: 105212199-105212199 on Build GRCh38
rs778919760 Chr.14: 105212209-105212209 on Build GRCh38
rs754778045 Chr.14: 105212119-105212119 on Build GRCh38
rs948754689 Chr.14: 105212340-105212340 on Build GRCh38
rs991425071 Chr.14: 105212486-105212486 on Build GRCh38
rs371012949 Chr.14: 105212098-105212098 on Build GRCh38
rs761336269 Chr.14: 105212076-105212076 on Build GRCh38
rs371720870 Chr.14: 105212370-105212370 on Build GRCh38
rs1044985241 Chr.14: 105212043-105212043 on Build GRCh38
rs955662189 Chr.14: 105212268-105212268 on Build GRCh38
rs587706907 Chr.14: 105212401-105212401 on Build GRCh38
rs749022159 Chr.14: 105212067-105212067 on Build GRCh38
rs3784232 Chr.14: 105212482-105212482 on Build GRCh38
rs587645432 Chr.14: 105212337-105212337 on Build GRCh38
rs934398778 Chr.14: 105212385-105212385 on Build GRCh38
rs952965470 Chr.14: 105212272-105212272 on Build GRCh38
rs1030356080 Chr.14: 105212219-105212219 on Build GRCh38
rs374100422 Chr.14: 105212181-105212181 on Build GRCh38
rs780509058 Chr.14: 105212161-105212161 on Build GRCh38
rs376321122 Chr.14: 105212099-105212099 on Build GRCh38
rs772572837 Chr.14: 105212188-105212188 on Build GRCh38
rs916747445 Chr.14: 105212494-105212494 on Build GRCh38
rs957528295 Chr.14: 105212297-105212297 on Build GRCh38
rs587775517 Chr.14: 105212069-105212069 on Build GRCh38
rs764429837 Chr.14: 105212134-105212134 on Build GRCh38
rs751770016 Chr.14: 105212143-105212143 on Build GRCh38
rs886904625 Chr.14: 105212125-105212125 on Build GRCh38
rs781421657 Chr.14: 105212150-105212150 on Build GRCh38
rs113844484 Chr.14: 105212449-105212449 on Build GRCh38
rs886979398 Chr.14: 105212374-105212374 on Build GRCh38
rs1047387251 Chr.14: 105212373-105212373 on Build GRCh38
rs1005385282 Chr.14: 105212384-105212384 on Build GRCh38
rs770264603 Chr.14: 105212056-105212056 on Build GRCh38
rs776450731 Chr.14: 105212413-105212413 on Build GRCh38
rs929289631 Chr.14: 105212485-105212485 on Build GRCh38
rs773953415 Chr.14: 105212072-105212072 on Build GRCh38
rs1019578397 Chr.14: 105212447-105212447 on Build GRCh38
rs374085127 Chr.14: 105212207-105212207 on Build GRCh38
rs587596351 Chr.14: 105212298-105212298 on Build GRCh38
rs61996214 Chr.14: 105212481-105212481 on Build GRCh38
rs1049559905 Chr.14: 105212418-105212418 on Build GRCh38
rs775707572 Chr.14: 105212057-105212057 on Build GRCh38
rs1008089979 Chr.14: 105212441-105212441 on Build GRCh38
rs763063078 Chr.14: 105212422-105212422 on Build GRCh38
rs766946634 Chr.14: 105212080-105212080 on Build GRCh38
rs753542257 Chr.14: 105212115-105212115 on Build GRCh38
rs906439103 Chr.14: 105212049-105212049 on Build GRCh38
rs757755740 Chr.14: 105212132-105212137 on Build GRCh38
rs769455295 Chr.14: 105212262-105212262 on Build GRCh38
rs756181206 Chr.14: 105212212-105212212 on Build GRCh38
rs990233994 Chr.14: 105212322-105212322 on Build GRCh38
rs759413665 Chr.14: 105212202-105212202 on Build GRCh38
rs746447007 Chr.14: 105212155-105212155 on Build GRCh38
rs1046912896 Chr.14: 105212104-105212104 on Build GRCh38
rs949649165 Chr.14: 105212333-105212333 on Build GRCh38
rs750441546 Chr.14: 105212211-105212211 on Build GRCh38
rs779906095 Chr.14: 105212257-105212257 on Build GRCh38
rs760705833 Chr.14: 105212189-105212189 on Build GRCh38
rs771628822 Chr.14: 105212185-105212185 on Build GRCh38
rs757420676 Chr.14: 105212145-105212145 on Build GRCh38
rs916841083 Chr.14: 105212301-105212301 on Build GRCh38
rs1129274 Chr.14: 105212157-105212157 on Build GRCh38
rs587595492 Chr.14: 105212473-105212473 on Build GRCh38
rs370558099 Chr.14: 105212089-105212089 on Build GRCh38
rs990265128 Chr.14: 105212429-105212429 on Build GRCh38
rs955173811 Chr.14: 105212407-105212407 on Build GRCh38
rs111552628 Chr.14: 105212310-105212310 on Build GRCh38
rs373243258 Chr.14: 105212408-105212408 on Build GRCh38
rs1045745634 Chr.14: 105212351-105212351 on Build GRCh38
rs991105333 Chr.14: 105212291-105212291 on Build GRCh38
rs370326351 Chr.14: 105212206-105212206 on Build GRCh38

View Full Product Details