Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 29120518-29121007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189A1
Entrez Gene ID
23359
Gene Name
family with sequence similarity 189 member A1
Gene Aliases
TMEM228
UniGene
Hs.383564
Gene Chromosome Location
Chr.15: 29120252-29570723 on Build GRCh38
Transcripts
NM_015307.1 XM_011521407.2

SNP Detail

SNP ID Chromosome Location
rs758092682 Chr.15: 29120676-29120676 on Build GRCh38
rs991849671 Chr.15: 29120792-29120792 on Build GRCh38
rs925341500 Chr.15: 29120703-29120703 on Build GRCh38
rs754522535 Chr.15: 29120636-29120636 on Build GRCh38
rs1047375676 Chr.15: 29120855-29120855 on Build GRCh38
rs35482946 Chr.15: 29120548-29120548 on Build GRCh38
rs557646637 Chr.15: 29120948-29120967 on Build GRCh38
rs949744856 Chr.15: 29120814-29120814 on Build GRCh38
rs78814938 Chr.15: 29120553-29120553 on Build GRCh38
rs769216503 Chr.15: 29120650-29120651 on Build GRCh38
rs4779803 Chr.15: 29120860-29120860 on Build GRCh38
rs928306645 Chr.15: 29120787-29120787 on Build GRCh38
rs920494284 Chr.15: 29120612-29120612 on Build GRCh38
rs746392118 Chr.15: 29120771-29120771 on Build GRCh38
rs552450221 Chr.15: 29120769-29120769 on Build GRCh38
rs986862150 Chr.15: 29120735-29120735 on Build GRCh38
rs1020062359 Chr.15: 29120671-29120671 on Build GRCh38
rs931077029 Chr.15: 29120854-29120854 on Build GRCh38
rs997511741 Chr.15: 29120707-29120707 on Build GRCh38
rs181457926 Chr.15: 29120600-29120600 on Build GRCh38
rs372456844 Chr.15: 29120916-29120916 on Build GRCh38
rs899858301 Chr.15: 29120871-29120871 on Build GRCh38
rs898210275 Chr.15: 29120828-29120828 on Build GRCh38
rs533381300 Chr.15: 29120628-29120628 on Build GRCh38
rs963996563 Chr.15: 29120678-29120678 on Build GRCh38
rs536866545 Chr.15: 29120918-29120918 on Build GRCh38
rs74568850 Chr.15: 29120586-29120586 on Build GRCh38
rs148231959 Chr.15: 29120727-29120727 on Build GRCh38
rs1045486335 Chr.15: 29120638-29120638 on Build GRCh38
rs531836534 Chr.15: 29120801-29120801 on Build GRCh38
rs1055562703 Chr.15: 29120827-29120827 on Build GRCh38
rs930563399 Chr.15: 29120615-29120615 on Build GRCh38
rs550752158 Chr.15: 29120927-29120927 on Build GRCh38
rs76010417 Chr.15: 29120564-29120564 on Build GRCh38
rs184646515 Chr.15: 29120635-29120635 on Build GRCh38
rs988232847 Chr.15: 29120962-29120962 on Build GRCh38
rs188469841 Chr.15: 29120697-29120697 on Build GRCh38
rs4779804 Chr.15: 29120892-29120892 on Build GRCh38
rs570761200 Chr.15: 29120981-29120981 on Build GRCh38
rs1045391483 Chr.15: 29120837-29120837 on Build GRCh38
rs955385934 Chr.15: 29120730-29120730 on Build GRCh38
rs893357036 Chr.15: 29120684-29120684 on Build GRCh38
rs955405498 Chr.15: 29120550-29120550 on Build GRCh38
rs1052714197 Chr.15: 29120551-29120551 on Build GRCh38
rs997093495 Chr.15: 29120933-29120933 on Build GRCh38
rs567526990 Chr.15: 29120760-29120760 on Build GRCh38
rs1021567844 Chr.15: 29120975-29120975 on Build GRCh38
rs745559867 Chr.15: 29120573-29120573 on Build GRCh38
rs574961636 Chr.15: 29120670-29120670 on Build GRCh38
rs1015661473 Chr.15: 29120738-29120738 on Build GRCh38
rs181653168 Chr.15: 29120862-29120862 on Build GRCh38
rs915565803 Chr.15: 29120810-29120810 on Build GRCh38
rs936965858 Chr.15: 29120819-29120819 on Build GRCh38
rs936818780 Chr.15: 29120539-29120539 on Build GRCh38
rs1001311958 Chr.15: 29120611-29120611 on Build GRCh38
rs73372321 Chr.15: 29120657-29120657 on Build GRCh38
rs545469293 Chr.15: 29120653-29120653 on Build GRCh38
rs886933780 Chr.15: 29120856-29120856 on Build GRCh38
rs1029014614 Chr.15: 29120951-29120951 on Build GRCh38
rs1055533221 Chr.15: 29120618-29120618 on Build GRCh38
rs4779801 Chr.15: 29120744-29120744 on Build GRCh38
rs4779802 Chr.15: 29120748-29120748 on Build GRCh38

View Full Product Details