Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 29121614-29122106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189A1
Entrez Gene ID
23359
Gene Name
family with sequence similarity 189 member A1
Gene Aliases
TMEM228
UniGene
Hs.383564
Gene Chromosome Location
Chr.15: 29120252-29570723 on Build GRCh38
Transcripts
NM_015307.1 XM_011521407.2

SNP Detail

SNP ID Chromosome Location
rs550994562 Chr.15: 29121658-29121658 on Build GRCh38
rs3829467 Chr.15: 29122034-29122034 on Build GRCh38
rs972862763 Chr.15: 29121864-29121864 on Build GRCh38
rs114451486 Chr.15: 29122086-29122086 on Build GRCh38
rs779183979 Chr.15: 29121807-29121807 on Build GRCh38
rs553270407 Chr.15: 29121854-29121854 on Build GRCh38
rs939307007 Chr.15: 29121995-29121995 on Build GRCh38
rs906514568 Chr.15: 29121711-29121711 on Build GRCh38
rs965398087 Chr.15: 29121842-29121842 on Build GRCh38
rs1003653259 Chr.15: 29121757-29121757 on Build GRCh38
rs57544714 Chr.15: 29121704-29121704 on Build GRCh38
rs962951594 Chr.15: 29121825-29121826 on Build GRCh38
rs573440517 Chr.15: 29121889-29121889 on Build GRCh38
rs1028326002 Chr.15: 29121806-29121806 on Build GRCh38
rs978396213 Chr.15: 29121923-29121923 on Build GRCh38
rs778749758 Chr.15: 29121808-29121808 on Build GRCh38
rs561767487 Chr.15: 29121919-29121919 on Build GRCh38
rs976488806 Chr.15: 29121872-29121872 on Build GRCh38
rs577054155 Chr.15: 29121783-29121783 on Build GRCh38
rs994972476 Chr.15: 29121805-29121805 on Build GRCh38
rs750228922 Chr.15: 29121952-29121952 on Build GRCh38
rs544357280 Chr.15: 29121992-29121992 on Build GRCh38
rs1027999473 Chr.15: 29121874-29121874 on Build GRCh38
rs965138186 Chr.15: 29121848-29121848 on Build GRCh38
rs930711282 Chr.15: 29122011-29122011 on Build GRCh38
rs931556526 Chr.15: 29121951-29121951 on Build GRCh38
rs1028030645 Chr.15: 29122042-29122042 on Build GRCh38
rs768686310 Chr.15: 29121765-29121765 on Build GRCh38
rs776588925 Chr.15: 29122087-29122087 on Build GRCh38
rs972915016 Chr.15: 29121684-29121684 on Build GRCh38
rs796726525 Chr.15: 29122078-29122078 on Build GRCh38
rs989314152 Chr.15: 29121887-29121887 on Build GRCh38
rs748846291 Chr.15: 29121750-29121750 on Build GRCh38
rs541460862 Chr.15: 29121895-29121895 on Build GRCh38
rs181930057 Chr.15: 29122071-29122071 on Build GRCh38
rs750894404 Chr.15: 29121688-29121688 on Build GRCh38
rs948133669 Chr.15: 29121730-29121730 on Build GRCh38
rs115162474 Chr.15: 29121692-29121692 on Build GRCh38
rs1015286183 Chr.15: 29121841-29121841 on Build GRCh38
rs1049261747 Chr.15: 29122018-29122018 on Build GRCh38
rs944211281 Chr.15: 29121674-29121674 on Build GRCh38
rs545961687 Chr.15: 29121846-29121846 on Build GRCh38
rs781537240 Chr.15: 29121661-29121661 on Build GRCh38
rs918766373 Chr.15: 29121869-29121869 on Build GRCh38
rs1005155859 Chr.15: 29122027-29122027 on Build GRCh38
rs557021602 Chr.15: 29121764-29121764 on Build GRCh38
rs891666941 Chr.15: 29121804-29121804 on Build GRCh38
rs886592069 Chr.15: 29122019-29122019 on Build GRCh38
rs530414185 Chr.15: 29121967-29121967 on Build GRCh38
rs932463111 Chr.15: 29121647-29121647 on Build GRCh38
rs895194591 Chr.15: 29122008-29122008 on Build GRCh38
rs546782732 Chr.15: 29122059-29122059 on Build GRCh38
rs1031509670 Chr.15: 29121769-29121769 on Build GRCh38
rs984425161 Chr.15: 29121957-29121957 on Build GRCh38
rs948081722 Chr.15: 29121981-29121981 on Build GRCh38
rs953887379 Chr.15: 29121885-29121885 on Build GRCh38
rs74007097 Chr.15: 29122030-29122030 on Build GRCh38
rs1021700538 Chr.15: 29121816-29121816 on Build GRCh38
rs997772688 Chr.15: 29122035-29122035 on Build GRCh38
rs60978477 Chr.15: 29121694-29121694 on Build GRCh38
rs1055225273 Chr.15: 29121999-29121999 on Build GRCh38
rs867542657 Chr.15: 29121918-29121918 on Build GRCh38
rs1005536273 Chr.15: 29121811-29121811 on Build GRCh38

View Full Product Details