Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 29122406-29122909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189A1
Entrez Gene ID
23359
Gene Name
family with sequence similarity 189 member A1
Gene Aliases
TMEM228
UniGene
Hs.383564
Gene Chromosome Location
Chr.15: 29120252-29570723 on Build GRCh38
Transcripts
NM_015307.1 XM_011521407.2

SNP Detail

SNP ID Chromosome Location
rs556046078 Chr.15: 29122754-29122754 on Build GRCh38
rs896197592 Chr.15: 29122752-29122752 on Build GRCh38
rs1043489337 Chr.15: 29122433-29122433 on Build GRCh38
rs986197364 Chr.15: 29122704-29122704 on Build GRCh38
rs966112209 Chr.15: 29122879-29122879 on Build GRCh38
rs188090039 Chr.15: 29122615-29122615 on Build GRCh38
rs745652192 Chr.15: 29122682-29122682 on Build GRCh38
rs1044394270 Chr.15: 29122838-29122838 on Build GRCh38
rs911046691 Chr.15: 29122571-29122571 on Build GRCh38
rs537849473 Chr.15: 29122841-29122841 on Build GRCh38
rs902163056 Chr.15: 29122632-29122632 on Build GRCh38
rs531801586 Chr.15: 29122575-29122575 on Build GRCh38
rs540327828 Chr.15: 29122488-29122488 on Build GRCh38
rs373186670 Chr.15: 29122448-29122448 on Build GRCh38
rs962047939 Chr.15: 29122704-29122704 on Build GRCh38
rs769532361 Chr.15: 29122727-29122727 on Build GRCh38
rs997633181 Chr.15: 29122880-29122880 on Build GRCh38
rs575974311 Chr.15: 29122758-29122758 on Build GRCh38
rs560336484 Chr.15: 29122495-29122495 on Build GRCh38
rs3812919 Chr.15: 29122681-29122681 on Build GRCh38
rs139006999 Chr.15: 29122555-29122555 on Build GRCh38
rs1034878359 Chr.15: 29122479-29122479 on Build GRCh38
rs756513616 Chr.15: 29122490-29122490 on Build GRCh38
rs988566043 Chr.15: 29122438-29122438 on Build GRCh38
rs1056231909 Chr.15: 29122732-29122732 on Build GRCh38
rs968158886 Chr.15: 29122553-29122553 on Build GRCh38
rs761489479 Chr.15: 29122432-29122432 on Build GRCh38
rs1017627733 Chr.15: 29122862-29122862 on Build GRCh38
rs1030289658 Chr.15: 29122856-29122856 on Build GRCh38
rs529063517 Chr.15: 29122507-29122507 on Build GRCh38
rs755028924 Chr.15: 29122585-29122585 on Build GRCh38
rs1015363447 Chr.15: 29122703-29122703 on Build GRCh38
rs1005750861 Chr.15: 29122517-29122517 on Build GRCh38
rs953498606 Chr.15: 29122869-29122869 on Build GRCh38
rs1010957559 Chr.15: 29122429-29122429 on Build GRCh38
rs570612980 Chr.15: 29122643-29122643 on Build GRCh38
rs190173926 Chr.15: 29122705-29122705 on Build GRCh38
rs983132202 Chr.15: 29122711-29122711 on Build GRCh38
rs772280052 Chr.15: 29122554-29122554 on Build GRCh38
rs764182310 Chr.15: 29122773-29122773 on Build GRCh38
rs1022720275 Chr.15: 29122549-29122549 on Build GRCh38
rs567086337 Chr.15: 29122706-29122706 on Build GRCh38
rs905397497 Chr.15: 29122518-29122518 on Build GRCh38
rs947657235 Chr.15: 29122715-29122715 on Build GRCh38
rs977681166 Chr.15: 29122718-29122718 on Build GRCh38
rs566469305 Chr.15: 29122731-29122731 on Build GRCh38
rs533012587 Chr.15: 29122487-29122487 on Build GRCh38
rs947783346 Chr.15: 29122753-29122753 on Build GRCh38
rs149358212 Chr.15: 29122620-29122620 on Build GRCh38
rs896331319 Chr.15: 29122644-29122644 on Build GRCh38
rs887730253 Chr.15: 29122691-29122691 on Build GRCh38
rs986303974 Chr.15: 29122712-29122712 on Build GRCh38
rs542714964 Chr.15: 29122481-29122481 on Build GRCh38
rs113250886 Chr.15: 29122704-29122705 on Build GRCh38
rs532936026 Chr.15: 29122703-29122704 on Build GRCh38
rs564185520 Chr.15: 29122467-29122467 on Build GRCh38
rs557964194 Chr.15: 29122545-29122547 on Build GRCh38
rs1003637815 Chr.15: 29122666-29122666 on Build GRCh38
rs1009810664 Chr.15: 29122807-29122807 on Build GRCh38
rs892663265 Chr.15: 29122791-29122791 on Build GRCh38
rs976120767 Chr.15: 29122539-29122539 on Build GRCh38
rs897828685 Chr.15: 29122853-29122853 on Build GRCh38
rs376275735 Chr.15: 29122522-29122522 on Build GRCh38
rs923460253 Chr.15: 29122547-29122547 on Build GRCh38
rs557834508 Chr.15: 29122855-29122855 on Build GRCh38
rs914739988 Chr.15: 29122713-29122713 on Build GRCh38
rs952087237 Chr.15: 29122492-29122492 on Build GRCh38
rs561116750 Chr.15: 29122542-29122542 on Build GRCh38
rs1023526007 Chr.15: 29122485-29122485 on Build GRCh38
rs535650029 Chr.15: 29122707-29122707 on Build GRCh38
rs1006128160 Chr.15: 29122692-29122692 on Build GRCh38
rs371451106 Chr.15: 29122641-29122641 on Build GRCh38
rs758671272 Chr.15: 29122616-29122616 on Build GRCh38
rs1006570806 Chr.15: 29122768-29122768 on Build GRCh38
rs56411615 Chr.15: 29122496-29122496 on Build GRCh38
rs935097972 Chr.15: 29122633-29122633 on Build GRCh38
rs942588603 Chr.15: 29122468-29122468 on Build GRCh38
rs962468272 Chr.15: 29122531-29122531 on Build GRCh38
rs550524054 Chr.15: 29122637-29122637 on Build GRCh38
rs1012186439 Chr.15: 29122482-29122482 on Build GRCh38
rs946798831 Chr.15: 29122458-29122459 on Build GRCh38
rs771380123 Chr.15: 29122541-29122541 on Build GRCh38

View Full Product Details