Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32111698-32112205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA7
Entrez Gene ID
1139
Gene Name
cholinergic receptor nicotinic alpha 7 subunit
Gene Aliases
CHRNA7-2, NACHRA7
UniGene
Hs.511772
Gene Chromosome Location
Chr.15: 32030462-32172521 on Build GRCh38
Transcripts
NM_000746.5 NM_001190455.2 NR_046324.1 XM_011521176.2 XM_011521178.2 XM_017021882.1 XM_017021883.1 XM_017021884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5812810 c.339T>A p.L113L Chr.15: 32111888-32111888 on Build GRCh38
174801 c.332A>G p.D111G Chr.15: 32111881-32111881 on Build GRCh38
1678333 c.246G>A p.W82* Chr.15: 32111795-32111795 on Build GRCh38
4672273 c.267G>A p.W89* Chr.15: 32111816-32111816 on Build GRCh38
4672272 c.354G>A p.W118* Chr.15: 32111816-32111816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769357181 Chr.15: 32111911-32111911 on Build GRCh38
rs200908085 Chr.15: 32111893-32111893 on Build GRCh38
rs187555054 Chr.15: 32112134-32112134 on Build GRCh38
rs1050477531 Chr.15: 32111729-32111729 on Build GRCh38
rs372841547 Chr.15: 32111861-32111861 on Build GRCh38
rs539097141 Chr.15: 32112096-32112096 on Build GRCh38
rs1015084885 Chr.15: 32112136-32112136 on Build GRCh38
rs752984260 Chr.15: 32111900-32111900 on Build GRCh38
rs781363844 Chr.15: 32111754-32111754 on Build GRCh38
rs199822557 Chr.15: 32111757-32111757 on Build GRCh38
rs558958460 Chr.15: 32111727-32111727 on Build GRCh38
rs200260877 Chr.15: 32111918-32111918 on Build GRCh38
rs977261838 Chr.15: 32112039-32112039 on Build GRCh38
rs762881799 Chr.15: 32111860-32111860 on Build GRCh38
rs775075161 Chr.15: 32111809-32111809 on Build GRCh38
rs528035597 Chr.15: 32111976-32111976 on Build GRCh38
rs1003244214 Chr.15: 32111840-32111840 on Build GRCh38
rs765144098 Chr.15: 32111851-32111851 on Build GRCh38
rs75599783 Chr.15: 32112120-32112120 on Build GRCh38
rs746590780 Chr.15: 32111812-32111812 on Build GRCh38
rs751683272 Chr.15: 32111867-32111867 on Build GRCh38
rs558874562 Chr.15: 32111958-32111958 on Build GRCh38
rs771303377 Chr.15: 32111757-32111758 on Build GRCh38
rs745360664 Chr.15: 32111783-32111783 on Build GRCh38
rs140842241 Chr.15: 32111883-32111883 on Build GRCh38
rs79187520 Chr.15: 32111732-32111732 on Build GRCh38
rs182243630 Chr.15: 32112026-32112026 on Build GRCh38
rs953560059 Chr.15: 32111821-32111821 on Build GRCh38
rs759731321 Chr.15: 32111919-32111920 on Build GRCh38
rs61341441 Chr.15: 32112034-32112034 on Build GRCh38
rs200196560 Chr.15: 32111886-32111886 on Build GRCh38
rs1018013093 Chr.15: 32112180-32112180 on Build GRCh38
rs776256188 Chr.15: 32111842-32111842 on Build GRCh38
rs947485249 Chr.15: 32112038-32112038 on Build GRCh38
rs947892840 Chr.15: 32112025-32112025 on Build GRCh38
rs761715059 Chr.15: 32111850-32111850 on Build GRCh38
rs61639061 Chr.15: 32112027-32112027 on Build GRCh38
rs1048565965 Chr.15: 32112111-32112111 on Build GRCh38
rs1006376823 Chr.15: 32111928-32111928 on Build GRCh38
rs894125176 Chr.15: 32112103-32112103 on Build GRCh38
rs764493580 Chr.15: 32112069-32112069 on Build GRCh38
rs189690056 Chr.15: 32111999-32111999 on Build GRCh38
rs545403606 Chr.15: 32111909-32111909 on Build GRCh38
rs1022530098 Chr.15: 32112056-32112056 on Build GRCh38
rs761003833 Chr.15: 32112149-32112149 on Build GRCh38
rs755117831 Chr.15: 32111882-32111882 on Build GRCh38
rs924475926 Chr.15: 32112042-32112042 on Build GRCh38
rs554543323 Chr.15: 32112157-32112157 on Build GRCh38
rs774677667 Chr.15: 32111773-32111774 on Build GRCh38
rs148971791 Chr.15: 32111781-32111781 on Build GRCh38
rs1012635598 Chr.15: 32111868-32111868 on Build GRCh38
rs558996708 Chr.15: 32112102-32112102 on Build GRCh38
rs757856659 Chr.15: 32111937-32111937 on Build GRCh38
rs543528898 Chr.15: 32111834-32111834 on Build GRCh38
rs948382566 Chr.15: 32112110-32112110 on Build GRCh38
rs754334488 Chr.15: 32111916-32111916 on Build GRCh38
rs755241662 Chr.15: 32111753-32111753 on Build GRCh38
rs145804770 Chr.15: 32111728-32111728 on Build GRCh38
rs773200568 Chr.15: 32111859-32111859 on Build GRCh38
rs746565649 Chr.15: 32111946-32111946 on Build GRCh38
rs188164539 Chr.15: 32111736-32111736 on Build GRCh38
rs1041995242 Chr.15: 32111778-32111778 on Build GRCh38
rs771779618 Chr.15: 32111788-32111788 on Build GRCh38
rs570180270 Chr.15: 32112035-32112035 on Build GRCh38
rs202080401 Chr.15: 32111914-32111914 on Build GRCh38
rs779387500 Chr.15: 32111942-32111942 on Build GRCh38
rs779554435 Chr.15: 32112022-32112022 on Build GRCh38
rs938548579 Chr.15: 32112085-32112085 on Build GRCh38
rs1004409495 Chr.15: 32112135-32112135 on Build GRCh38

View Full Product Details