Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32730598-32731100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GREM1
Entrez Gene ID
26585
Gene Name
gremlin 1, DAN family BMP antagonist
Gene Aliases
C15DUPq, CKTSF1B1, CRAC1, CRCS4, DAND2, DRM, DUP15q, GREMLIN, HMPS, HMPS1, IHG-2, MPSH, PIG2
UniGene
Hs.40098
Gene Chromosome Location
Chr.15: 32717940-32734669 on Build GRCh38
Transcripts
NM_001191322.1 NM_001191323.1 NM_013372.6 XM_005254301.2 XM_017022077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
386659 c.322T>A p.C108S Chr.15: 32731012-32731012 on Build GRCh38
700176 c.87C>T p.S29S Chr.15: 32730777-32730777 on Build GRCh38
6895426 c.192G>T p.R64R Chr.15: 32730882-32730882 on Build GRCh38
700173 c.186A>T p.Q62H Chr.15: 32730876-32730876 on Build GRCh38
3706511 c.111C>T p.D37D Chr.15: 32730801-32730801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749057876 Chr.15: 32730930-32730930 on Build GRCh38
rs997413825 Chr.15: 32730720-32730720 on Build GRCh38
rs868208266 Chr.15: 32730698-32730698 on Build GRCh38
rs572405915 Chr.15: 32730825-32730825 on Build GRCh38
rs370674293 Chr.15: 32730997-32730997 on Build GRCh38
rs775520243 Chr.15: 32731053-32731053 on Build GRCh38
rs745622778 Chr.15: 32731044-32731044 on Build GRCh38
rs372172128 Chr.15: 32730822-32730822 on Build GRCh38
rs12911221 Chr.15: 32730955-32730955 on Build GRCh38
rs755799933 Chr.15: 32730900-32730900 on Build GRCh38
rs202162184 Chr.15: 32730677-32730677 on Build GRCh38
rs1002880323 Chr.15: 32730984-32730984 on Build GRCh38
rs748340321 Chr.15: 32731068-32731068 on Build GRCh38
rs779454972 Chr.15: 32730794-32730794 on Build GRCh38
rs753975618 Chr.15: 32730860-32730861 on Build GRCh38
rs750874632 Chr.15: 32730880-32730880 on Build GRCh38
rs776296145 Chr.15: 32730700-32730700 on Build GRCh38
rs1043315001 Chr.15: 32730816-32730816 on Build GRCh38
rs1027553263 Chr.15: 32731030-32731030 on Build GRCh38
rs2280738 Chr.15: 32730897-32730897 on Build GRCh38
rs193125649 Chr.15: 32730655-32730655 on Build GRCh38
rs1035305843 Chr.15: 32730991-32730991 on Build GRCh38
rs34096580 Chr.15: 32731047-32731047 on Build GRCh38
rs759636569 Chr.15: 32730750-32730750 on Build GRCh38
rs777005561 Chr.15: 32731073-32731073 on Build GRCh38
rs769511007 Chr.15: 32730721-32730721 on Build GRCh38
rs762932572 Chr.15: 32730738-32730738 on Build GRCh38
rs900035234 Chr.15: 32730669-32730669 on Build GRCh38
rs764435884 Chr.15: 32730944-32730944 on Build GRCh38
rs545952537 Chr.15: 32730830-32730830 on Build GRCh38
rs78005246 Chr.15: 32730999-32730999 on Build GRCh38
rs748915906 Chr.15: 32731035-32731035 on Build GRCh38
rs761624816 Chr.15: 32730701-32730701 on Build GRCh38
rs944445481 Chr.15: 32730665-32730665 on Build GRCh38
rs759010741 Chr.15: 32731000-32731000 on Build GRCh38
rs752903637 Chr.15: 32730772-32730772 on Build GRCh38
rs778837795 Chr.15: 32731038-32731038 on Build GRCh38
rs200285291 Chr.15: 32730846-32730846 on Build GRCh38
rs777378898 Chr.15: 32730917-32730917 on Build GRCh38
rs267604152 Chr.15: 32730762-32730762 on Build GRCh38
rs553840621 Chr.15: 32730812-32730812 on Build GRCh38
rs779317921 Chr.15: 32730875-32730875 on Build GRCh38
rs776954783 Chr.15: 32730949-32730949 on Build GRCh38
rs775753186 Chr.15: 32730942-32730942 on Build GRCh38
rs762152055 Chr.15: 32731076-32731076 on Build GRCh38
rs769422903 Chr.15: 32730817-32730817 on Build GRCh38
rs764380818 Chr.15: 32730855-32730855 on Build GRCh38
rs927970468 Chr.15: 32730777-32730777 on Build GRCh38
rs538020861 Chr.15: 32730667-32730667 on Build GRCh38
rs764243456 Chr.15: 32730655-32730656 on Build GRCh38
rs762082199 Chr.15: 32730827-32730827 on Build GRCh38
rs778343488 Chr.15: 32730792-32730792 on Build GRCh38
rs745880464 Chr.15: 32730935-32730935 on Build GRCh38
rs759541309 Chr.15: 32730837-32730837 on Build GRCh38
rs765373204 Chr.15: 32731079-32731079 on Build GRCh38
rs1054066828 Chr.15: 32730723-32730723 on Build GRCh38
rs770819924 Chr.15: 32730833-32730833 on Build GRCh38
rs869312795 Chr.15: 32730963-32730963 on Build GRCh38
rs371194430 Chr.15: 32731062-32731062 on Build GRCh38
rs768170664 Chr.15: 32730806-32730806 on Build GRCh38
rs752249342 Chr.15: 32731010-32731010 on Build GRCh38
rs757246207 Chr.15: 32730865-32730876 on Build GRCh38
rs367997246 Chr.15: 32730810-32730810 on Build GRCh38
rs770695786 Chr.15: 32730931-32730931 on Build GRCh38
rs766332214 Chr.15: 32730746-32730746 on Build GRCh38
rs111262341 Chr.15: 32730793-32730793 on Build GRCh38
rs1017180089 Chr.15: 32730711-32730711 on Build GRCh38
rs766836254 Chr.15: 32731002-32731002 on Build GRCh38
rs968111910 Chr.15: 32731008-32731008 on Build GRCh38
rs774292923 Chr.15: 32730834-32730834 on Build GRCh38
rs148668967 Chr.15: 32730749-32730749 on Build GRCh38
rs945879966 Chr.15: 32730815-32730815 on Build GRCh38
rs747649135 Chr.15: 32730898-32730898 on Build GRCh38
rs199894051 Chr.15: 32730767-32730767 on Build GRCh38
rs997876408 Chr.15: 32730856-32730856 on Build GRCh38
rs756849803 Chr.15: 32731037-32731037 on Build GRCh38
rs771903480 Chr.15: 32730941-32730941 on Build GRCh38
rs367676745 Chr.15: 32731034-32731034 on Build GRCh38
rs754459001 Chr.15: 32730885-32730885 on Build GRCh38
rs755568850 Chr.15: 32731027-32731027 on Build GRCh38
rs757593929 Chr.15: 32730861-32730861 on Build GRCh38
rs368472880 Chr.15: 32730933-32730933 on Build GRCh38
rs768242805 Chr.15: 32730697-32730697 on Build GRCh38
rs767579208 Chr.15: 32730840-32730840 on Build GRCh38
rs773145682 Chr.15: 32730732-32730732 on Build GRCh38
rs754206331 Chr.15: 32730859-32730859 on Build GRCh38
rs756565137 Chr.15: 32730788-32730788 on Build GRCh38
rs762087325 Chr.15: 32730959-32730959 on Build GRCh38
rs746525315 Chr.15: 32730795-32730795 on Build GRCh38

View Full Product Details