Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32730939-32731452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GREM1
Entrez Gene ID
26585
Gene Name
gremlin 1, DAN family BMP antagonist
Gene Aliases
C15DUPq, CKTSF1B1, CRAC1, CRCS4, DAND2, DRM, DUP15q, GREMLIN, HMPS, HMPS1, IHG-2, MPSH, PIG2
UniGene
Hs.40098
Gene Chromosome Location
Chr.15: 32717940-32734669 on Build GRCh38
Transcripts
NM_001191322.1 NM_001191323.1 NM_013372.6 XM_005254301.2 XM_017022077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6871620 c.280T>A p.C94S Chr.15: 32730970-32730970 on Build GRCh38
386659 c.322T>A p.C108S Chr.15: 32731012-32731012 on Build GRCh38
7088608 c.355_356insA p.Y119fs*1 Chr.15: 32731045-32731046 on Build GRCh38
6554850 c.386G>T p.R129M Chr.15: 32731076-32731076 on Build GRCh38
6857851 c.418_419insTCC p.S140>FP Chr.15: 32731108-32731109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775560885 Chr.15: 32731368-32731368 on Build GRCh38
rs770099270 Chr.15: 32731206-32731206 on Build GRCh38
rs771216139 Chr.15: 32731230-32731230 on Build GRCh38
rs974551417 Chr.15: 32731187-32731187 on Build GRCh38
rs758228532 Chr.15: 32731170-32731170 on Build GRCh38
rs781399623 Chr.15: 32731203-32731203 on Build GRCh38
rs933638318 Chr.15: 32731319-32731319 on Build GRCh38
rs1042416265 Chr.15: 32731393-32731393 on Build GRCh38
rs370674293 Chr.15: 32730997-32730997 on Build GRCh38
rs750254980 Chr.15: 32731155-32731155 on Build GRCh38
rs775520243 Chr.15: 32731053-32731053 on Build GRCh38
rs749572971 Chr.15: 32731288-32731288 on Build GRCh38
rs745622778 Chr.15: 32731044-32731044 on Build GRCh38
rs753442229 Chr.15: 32731247-32731247 on Build GRCh38
rs774688129 Chr.15: 32731233-32731233 on Build GRCh38
rs1035231292 Chr.15: 32731426-32731426 on Build GRCh38
rs752018538 Chr.15: 32731098-32731098 on Build GRCh38
rs1002880323 Chr.15: 32730984-32730984 on Build GRCh38
rs748340321 Chr.15: 32731068-32731068 on Build GRCh38
rs1027553263 Chr.15: 32731030-32731030 on Build GRCh38
rs199760237 Chr.15: 32731153-32731153 on Build GRCh38
rs756977802 Chr.15: 32731140-32731140 on Build GRCh38
rs202104240 Chr.15: 32731139-32731139 on Build GRCh38
rs34096580 Chr.15: 32731047-32731047 on Build GRCh38
rs1035305843 Chr.15: 32730991-32730991 on Build GRCh38
rs777005561 Chr.15: 32731073-32731073 on Build GRCh38
rs748146251 Chr.15: 32731204-32731204 on Build GRCh38
rs78005246 Chr.15: 32730999-32730999 on Build GRCh38
rs12915554 Chr.15: 32731285-32731285 on Build GRCh38
rs794727286 Chr.15: 32731107-32731107 on Build GRCh38
rs748915906 Chr.15: 32731035-32731035 on Build GRCh38
rs760105878 Chr.15: 32731240-32731240 on Build GRCh38
rs759010741 Chr.15: 32731000-32731000 on Build GRCh38
rs748278261 Chr.15: 32731276-32731276 on Build GRCh38
rs778837795 Chr.15: 32731038-32731038 on Build GRCh38
rs780051578 Chr.15: 32731268-32731268 on Build GRCh38
rs755005942 Chr.15: 32731273-32731273 on Build GRCh38
rs768700770 Chr.15: 32731182-32731182 on Build GRCh38
rs867785991 Chr.15: 32731287-32731287 on Build GRCh38
rs755636439 Chr.15: 32731116-32731116 on Build GRCh38
rs762152055 Chr.15: 32731076-32731076 on Build GRCh38
rs1002398152 Chr.15: 32731385-32731385 on Build GRCh38
rs921662109 Chr.15: 32731209-32731209 on Build GRCh38
rs763654384 Chr.15: 32731125-32731125 on Build GRCh38
rs199522589 Chr.15: 32731092-32731092 on Build GRCh38
rs561986028 Chr.15: 32731275-32731275 on Build GRCh38
rs765373204 Chr.15: 32731079-32731079 on Build GRCh38
rs749149648 Chr.15: 32731376-32731376 on Build GRCh38
rs368726521 Chr.15: 32731226-32731226 on Build GRCh38
rs763336404 Chr.15: 32731215-32731215 on Build GRCh38
rs1038505414 Chr.15: 32731357-32731357 on Build GRCh38
rs746883700 Chr.15: 32731179-32731179 on Build GRCh38
rs767985360 Chr.15: 32731241-32731241 on Build GRCh38
rs752249342 Chr.15: 32731010-32731010 on Build GRCh38
rs371194430 Chr.15: 32731062-32731062 on Build GRCh38
rs869312795 Chr.15: 32730963-32730963 on Build GRCh38
rs777740546 Chr.15: 32731084-32731084 on Build GRCh38
rs371474885 Chr.15: 32731386-32731386 on Build GRCh38
rs764973719 Chr.15: 32731258-32731258 on Build GRCh38
rs886820859 Chr.15: 32731347-32731347 on Build GRCh38
rs1046774219 Chr.15: 32731342-32731342 on Build GRCh38
rs780146359 Chr.15: 32731173-32731173 on Build GRCh38
rs145439767 Chr.15: 32731081-32731081 on Build GRCh38
rs766836254 Chr.15: 32731002-32731002 on Build GRCh38
rs372399812 Chr.15: 32731259-32731259 on Build GRCh38
rs548075623 Chr.15: 32731353-32731353 on Build GRCh38
rs968111910 Chr.15: 32731008-32731008 on Build GRCh38
rs945576519 Chr.15: 32731378-32731378 on Build GRCh38
rs766891324 Chr.15: 32731088-32731088 on Build GRCh38
rs201006159 Chr.15: 32731365-32731365 on Build GRCh38
rs756849803 Chr.15: 32731037-32731037 on Build GRCh38
rs773255112 Chr.15: 32731212-32731212 on Build GRCh38
rs367676745 Chr.15: 32731034-32731034 on Build GRCh38
rs755568850 Chr.15: 32731027-32731027 on Build GRCh38
rs751395218 Chr.15: 32731271-32731271 on Build GRCh38
rs761180040 Chr.15: 32731254-32731254 on Build GRCh38
rs780077846 Chr.15: 32731296-32731296 on Build GRCh38
rs769719167 Chr.15: 32731284-32731284 on Build GRCh38
rs906573252 Chr.15: 32731404-32731404 on Build GRCh38
rs942578260 Chr.15: 32731225-32731225 on Build GRCh38
rs1003947342 Chr.15: 32731421-32731421 on Build GRCh38

View Full Product Details