Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32733320-32733835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GREM1
Entrez Gene ID
26585
Gene Name
gremlin 1, DAN family BMP antagonist
Gene Aliases
C15DUPq, CKTSF1B1, CRAC1, CRCS4, DAND2, DRM, DUP15q, GREMLIN, HMPS, HMPS1, IHG-2, MPSH, PIG2
UniGene
Hs.40098
Gene Chromosome Location
Chr.15: 32717940-32734669 on Build GRCh38
Transcripts
NM_001191322.1 NM_001191323.1 NM_013372.6 XM_005254301.2 XM_017022077.1

SNP Detail

SNP ID Chromosome Location
rs535069918 Chr.15: 32733423-32733423 on Build GRCh38
rs114363828 Chr.15: 32733714-32733714 on Build GRCh38
rs182520997 Chr.15: 32733345-32733345 on Build GRCh38
rs11853876 Chr.15: 32733579-32733579 on Build GRCh38
rs995052243 Chr.15: 32733603-32733607 on Build GRCh38
rs143200567 Chr.15: 32733708-32733708 on Build GRCh38
rs894737673 Chr.15: 32733621-32733621 on Build GRCh38
rs756258403 Chr.15: 32733522-32733522 on Build GRCh38
rs543809612 Chr.15: 32733529-32733529 on Build GRCh38
rs992799596 Chr.15: 32733617-32733617 on Build GRCh38
rs1030990646 Chr.15: 32733447-32733447 on Build GRCh38
rs956743878 Chr.15: 32733743-32733743 on Build GRCh38
rs539219397 Chr.15: 32733488-32733488 on Build GRCh38
rs1054094654 Chr.15: 32733485-32733485 on Build GRCh38
rs1050091072 Chr.15: 32733530-32733530 on Build GRCh38
rs3743103 Chr.15: 32733426-32733426 on Build GRCh38
rs555297512 Chr.15: 32733634-32733634 on Build GRCh38
rs923786135 Chr.15: 32733420-32733420 on Build GRCh38
rs960138472 Chr.15: 32733604-32733604 on Build GRCh38
rs140081414 Chr.15: 32733541-32733541 on Build GRCh38
rs997742976 Chr.15: 32733440-32733440 on Build GRCh38
rs1048154751 Chr.15: 32733388-32733388 on Build GRCh38
rs909653580 Chr.15: 32733678-32733678 on Build GRCh38
rs1006358058 Chr.15: 32733421-32733421 on Build GRCh38
rs746725988 Chr.15: 32733762-32733762 on Build GRCh38
rs765617880 Chr.15: 32733386-32733388 on Build GRCh38
rs373252623 Chr.15: 32733693-32733693 on Build GRCh38
rs915201424 Chr.15: 32733486-32733486 on Build GRCh38
rs539300179 Chr.15: 32733602-32733604 on Build GRCh38
rs981888119 Chr.15: 32733417-32733417 on Build GRCh38
rs141078860 Chr.15: 32733661-32733661 on Build GRCh38
rs1052044805 Chr.15: 32733751-32733751 on Build GRCh38
rs1036944069 Chr.15: 32733694-32733694 on Build GRCh38
rs983723821 Chr.15: 32733642-32733642 on Build GRCh38
rs1007585888 Chr.15: 32733789-32733789 on Build GRCh38
rs933480324 Chr.15: 32733715-32733715 on Build GRCh38
rs10318 Chr.15: 32733778-32733778 on Build GRCh38
rs1031780931 Chr.15: 32733719-32733719 on Build GRCh38
rs571835053 Chr.15: 32733429-32733429 on Build GRCh38
rs752649459 Chr.15: 32733632-32733632 on Build GRCh38
rs765146186 Chr.15: 32733580-32733580 on Build GRCh38
rs146793673 Chr.15: 32733590-32733590 on Build GRCh38
rs888182988 Chr.15: 32733416-32733416 on Build GRCh38
rs567560278 Chr.15: 32733393-32733393 on Build GRCh38
rs1007544882 Chr.15: 32733535-32733535 on Build GRCh38
rs944451644 Chr.15: 32733567-32733567 on Build GRCh38

View Full Product Details