Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32733629-32734173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GREM1
Entrez Gene ID
26585
Gene Name
gremlin 1, DAN family BMP antagonist
Gene Aliases
C15DUPq, CKTSF1B1, CRAC1, CRCS4, DAND2, DRM, DUP15q, GREMLIN, HMPS, HMPS1, IHG-2, MPSH, PIG2
UniGene
Hs.40098
Gene Chromosome Location
Chr.15: 32717940-32734669 on Build GRCh38
Transcripts
NM_001191322.1 NM_001191323.1 NM_013372.6 XM_005254301.2 XM_017022077.1

SNP Detail

SNP ID Chromosome Location
rs922324563 Chr.15: 32734041-32734041 on Build GRCh38
rs894709644 Chr.15: 32734038-32734038 on Build GRCh38
rs1032170952 Chr.15: 32733837-32733837 on Build GRCh38
rs114363828 Chr.15: 32733714-32733714 on Build GRCh38
rs983753385 Chr.15: 32733986-32733986 on Build GRCh38
rs998906133 Chr.15: 32734078-32734078 on Build GRCh38
rs998975236 Chr.15: 32733827-32733827 on Build GRCh38
rs1021597921 Chr.15: 32733814-32733814 on Build GRCh38
rs993057058 Chr.15: 32733859-32733859 on Build GRCh38
rs757902225 Chr.15: 32733907-32733913 on Build GRCh38
rs527518402 Chr.15: 32734096-32734096 on Build GRCh38
rs571747225 Chr.15: 32734134-32734134 on Build GRCh38
rs182180121 Chr.15: 32734126-32734126 on Build GRCh38
rs530791359 Chr.15: 32733863-32733863 on Build GRCh38
rs1052343766 Chr.15: 32734068-32734068 on Build GRCh38
rs909653580 Chr.15: 32733678-32733678 on Build GRCh38
rs746725988 Chr.15: 32733762-32733762 on Build GRCh38
rs373252623 Chr.15: 32733693-32733693 on Build GRCh38
rs1043322630 Chr.15: 32734133-32734133 on Build GRCh38
rs867598266 Chr.15: 32733855-32733855 on Build GRCh38
rs141078860 Chr.15: 32733661-32733661 on Build GRCh38
rs185751333 Chr.15: 32733816-32733816 on Build GRCh38
rs933530010 Chr.15: 32734056-32734056 on Build GRCh38
rs933480324 Chr.15: 32733715-32733715 on Build GRCh38
rs746624633 Chr.15: 32733989-32733993 on Build GRCh38
rs10318 Chr.15: 32733778-32733778 on Build GRCh38
rs989695199 Chr.15: 32734115-32734115 on Build GRCh38
rs1031780931 Chr.15: 32733719-32733719 on Build GRCh38
rs1036517722 Chr.15: 32733904-32733904 on Build GRCh38
rs369985830 Chr.15: 32733860-32733860 on Build GRCh38
rs939679784 Chr.15: 32734023-32734023 on Build GRCh38
rs373869559 Chr.15: 32733982-32733982 on Build GRCh38
rs528491836 Chr.15: 32734097-32734097 on Build GRCh38
rs189731184 Chr.15: 32733958-32733958 on Build GRCh38
rs969923679 Chr.15: 32733948-32733948 on Build GRCh38
rs909517758 Chr.15: 32733988-32733988 on Build GRCh38
rs143200567 Chr.15: 32733708-32733708 on Build GRCh38
rs1032166032 Chr.15: 32734079-32734079 on Build GRCh38
rs956743878 Chr.15: 32733743-32733743 on Build GRCh38
rs780822127 Chr.15: 32733821-32733821 on Build GRCh38
rs959923532 Chr.15: 32733852-32733852 on Build GRCh38
rs1024542786 Chr.15: 32734059-32734059 on Build GRCh38
rs972546496 Chr.15: 32734032-32734032 on Build GRCh38
rs915103558 Chr.15: 32733815-32733815 on Build GRCh38
rs796165203 Chr.15: 32734150-32734150 on Build GRCh38
rs550918345 Chr.15: 32734063-32734064 on Build GRCh38
rs1052044805 Chr.15: 32733751-32733751 on Build GRCh38
rs897605626 Chr.15: 32733913-32733913 on Build GRCh38
rs1036944069 Chr.15: 32733694-32733694 on Build GRCh38
rs1007585888 Chr.15: 32733789-32733789 on Build GRCh38
rs769267630 Chr.15: 32734140-32734140 on Build GRCh38
rs911546193 Chr.15: 32733878-32733878 on Build GRCh38
rs62002611 Chr.15: 32734087-32734087 on Build GRCh38
rs944377163 Chr.15: 32733880-32733880 on Build GRCh38

View Full Product Details