Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 35450534-35451030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH6
Entrez Gene ID
89978
Gene Name
diphthamine biosynthesis 6
Gene Aliases
ATPBD4
UniGene
Hs.107196
Gene Chromosome Location
Chr.15: 35217356-35546222 on Build GRCh38
Transcripts
NM_080650.3 XM_011522160.1 XM_017022707.1 XM_017022708.1 XM_017022709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4548065 c.438G>A p.Q146Q Chr.15: 35450752-35450752 on Build GRCh38
4492906 c.405C>T p.L135L Chr.15: 35450785-35450785 on Build GRCh38
6840914 c.485T>C p.I162T Chr.15: 35450705-35450705 on Build GRCh38
1372537 c.431G>A p.R144K Chr.15: 35450759-35450759 on Build GRCh38
3886871 c.464C>T p.S155L Chr.15: 35450726-35450726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913831763 Chr.15: 35450935-35450935 on Build GRCh38
rs758182103 Chr.15: 35450838-35450838 on Build GRCh38
rs770014356 Chr.15: 35450743-35450743 on Build GRCh38
rs966757159 Chr.15: 35450978-35450978 on Build GRCh38
rs780294614 Chr.15: 35450840-35450840 on Build GRCh38
rs867195336 Chr.15: 35450728-35450728 on Build GRCh38
rs749865625 Chr.15: 35450794-35450794 on Build GRCh38
rs773827693 Chr.15: 35450667-35450667 on Build GRCh38
rs757023821 Chr.15: 35450708-35450708 on Build GRCh38
rs771993124 Chr.15: 35450634-35450634 on Build GRCh38
rs529483952 Chr.15: 35450609-35450609 on Build GRCh38
rs369514948 Chr.15: 35450659-35450659 on Build GRCh38
rs570003815 Chr.15: 35450938-35450938 on Build GRCh38
rs767186937 Chr.15: 35450799-35450799 on Build GRCh38
rs77687650 Chr.15: 35450832-35450832 on Build GRCh38
rs766872767 Chr.15: 35450674-35450674 on Build GRCh38
rs747440013 Chr.15: 35450827-35450830 on Build GRCh38
rs567923256 Chr.15: 35450624-35450624 on Build GRCh38
rs944741984 Chr.15: 35450812-35450812 on Build GRCh38
rs751881870 Chr.15: 35450841-35450841 on Build GRCh38
rs773630188 Chr.15: 35450779-35450779 on Build GRCh38
rs755677220 Chr.15: 35450684-35450684 on Build GRCh38
rs981805252 Chr.15: 35450999-35450999 on Build GRCh38
rs1027998141 Chr.15: 35450888-35450888 on Build GRCh38
rs761767468 Chr.15: 35450827-35450827 on Build GRCh38
rs370800281 Chr.15: 35450705-35450705 on Build GRCh38
rs760144861 Chr.15: 35450805-35450805 on Build GRCh38
rs755065158 Chr.15: 35450847-35450847 on Build GRCh38
rs1033216310 Chr.15: 35450934-35450935 on Build GRCh38
rs777347127 Chr.15: 35450654-35450654 on Build GRCh38
rs756808860 Chr.15: 35450729-35450731 on Build GRCh38
rs547579810 Chr.15: 35450649-35450649 on Build GRCh38
rs930768927 Chr.15: 35450761-35450761 on Build GRCh38
rs777260966 Chr.15: 35450722-35450722 on Build GRCh38
rs764831557 Chr.15: 35450788-35450789 on Build GRCh38
rs1004407569 Chr.15: 35450770-35450770 on Build GRCh38
rs920672304 Chr.15: 35450734-35450734 on Build GRCh38
rs756171501 Chr.15: 35450854-35450854 on Build GRCh38
rs759054480 Chr.15: 35450784-35450784 on Build GRCh38
rs780524111 Chr.15: 35450808-35450813 on Build GRCh38
rs533235178 Chr.15: 35450718-35450718 on Build GRCh38
rs749441429 Chr.15: 35450574-35450574 on Build GRCh38
rs775071387 Chr.15: 35450803-35450803 on Build GRCh38
rs900762823 Chr.15: 35450895-35450895 on Build GRCh38
rs747451842 Chr.15: 35450647-35450647 on Build GRCh38
rs374248836 Chr.15: 35450681-35450681 on Build GRCh38
rs561951808 Chr.15: 35450592-35450592 on Build GRCh38
rs779936508 Chr.15: 35450715-35450715 on Build GRCh38
rs748690837 Chr.15: 35450731-35450731 on Build GRCh38
rs955250403 Chr.15: 35450896-35450896 on Build GRCh38
rs904647795 Chr.15: 35450597-35450597 on Build GRCh38
rs1028040017 Chr.15: 35450577-35450577 on Build GRCh38
rs759264738 Chr.15: 35450669-35450669 on Build GRCh38
rs758988542 Chr.15: 35450807-35450808 on Build GRCh38
rs986733708 Chr.15: 35450918-35450918 on Build GRCh38
rs763642261 Chr.15: 35450811-35450811 on Build GRCh38
rs990622214 Chr.15: 35450707-35450707 on Build GRCh38
rs750653114 Chr.15: 35450709-35450709 on Build GRCh38
rs753725196 Chr.15: 35450814-35450814 on Build GRCh38
rs750244410 Chr.15: 35450835-35450835 on Build GRCh38
rs537760872 Chr.15: 35450953-35450953 on Build GRCh38
rs1021061251 Chr.15: 35450604-35450604 on Build GRCh38
rs377536835 Chr.15: 35450638-35450638 on Build GRCh38
rs201959314 Chr.15: 35450782-35450782 on Build GRCh38
rs935976477 Chr.15: 35450620-35450620 on Build GRCh38
rs962465352 Chr.15: 35450842-35450842 on Build GRCh38
rs565863034 Chr.15: 35450686-35450686 on Build GRCh38
rs758649328 Chr.15: 35450714-35450714 on Build GRCh38
rs979250641 Chr.15: 35450957-35450957 on Build GRCh38
rs752480716 Chr.15: 35450561-35450561 on Build GRCh38
rs762826567 Chr.15: 35450628-35450628 on Build GRCh38

View Full Product Details