Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40252167-40252671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1B-PAK6
Entrez Gene ID
106821730
Gene Name
BUB1B-PAK6 readthrough
Gene Aliases
-
UniGene
Hs.732375
Gene Chromosome Location
Chr.15: 40217428-40277487 on Build GRCh38
Transcripts
NM_001128628.2 NM_001128629.2 NM_001039905.2 NM_001276718.1 NM_020168.5

Gene Symbol
C15orf56
Entrez Gene ID
644809
Gene Name
chromosome 15 open reading frame 56
Gene Aliases
-
UniGene
Hs.631718
Gene Chromosome Location
Chr.15: 40250664-40252909 on Build GRCh38
Transcripts
NM_001128628.2 NM_001128629.2 NM_001039905.2 NM_001276718.1 NM_020168.5

Gene Symbol
PAK6
Entrez Gene ID
56924
Gene Name
p21 (RAC1) activated kinase 6
Gene Aliases
PAK5
UniGene
Hs.732375
Gene Chromosome Location
Chr.15: 40239091-40277487 on Build GRCh38
Transcripts
NM_001128628.2 NM_001128629.2 NM_001039905.2 NM_001276718.1 NM_020168.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5564398 c.394C>T p.R132W Chr.15: 40252377-40252377 on Build GRCh38
4884006 c.440C>A p.T147K Chr.15: 40252331-40252331 on Build GRCh38
4054298 c.254G>T p.R85L Chr.15: 40252517-40252517 on Build GRCh38
4853835 c.457C>A p.R153S Chr.15: 40252314-40252314 on Build GRCh38
6425611 c.430T>C p.C144R Chr.15: 40252341-40252341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562091767 Chr.15: 40252336-40252336 on Build GRCh38
rs1013896700 Chr.15: 40252478-40252478 on Build GRCh38
rs764361427 Chr.15: 40252332-40252333 on Build GRCh38
rs55863440 Chr.15: 40252416-40252416 on Build GRCh38
rs766770622 Chr.15: 40252458-40252458 on Build GRCh38
rs764313551 Chr.15: 40252323-40252323 on Build GRCh38
rs747589956 Chr.15: 40252282-40252282 on Build GRCh38
rs113214405 Chr.15: 40252577-40252577 on Build GRCh38
rs1024199965 Chr.15: 40252512-40252512 on Build GRCh38
rs756696424 Chr.15: 40252581-40252581 on Build GRCh38
rs893750258 Chr.15: 40252309-40252309 on Build GRCh38
rs777674535 Chr.15: 40252284-40252284 on Build GRCh38
rs540327479 Chr.15: 40252225-40252225 on Build GRCh38
rs769758263 Chr.15: 40252619-40252619 on Build GRCh38
rs763281874 Chr.15: 40252444-40252444 on Build GRCh38
rs758795556 Chr.15: 40252348-40252348 on Build GRCh38
rs769966205 Chr.15: 40252545-40252545 on Build GRCh38
rs373664252 Chr.15: 40252498-40252498 on Build GRCh38
rs375354593 Chr.15: 40252642-40252642 on Build GRCh38
rs758357182 Chr.15: 40252591-40252591 on Build GRCh38
rs550607348 Chr.15: 40252396-40252396 on Build GRCh38
rs770713785 Chr.15: 40252389-40252389 on Build GRCh38
rs191930557 Chr.15: 40252570-40252570 on Build GRCh38
rs763610558 Chr.15: 40252576-40252576 on Build GRCh38
rs1033505269 Chr.15: 40252605-40252605 on Build GRCh38
rs868669376 Chr.15: 40252608-40252608 on Build GRCh38
rs761960069 Chr.15: 40252426-40252426 on Build GRCh38
rs746816528 Chr.15: 40252522-40252522 on Build GRCh38
rs754286655 Chr.15: 40252564-40252567 on Build GRCh38
rs756820191 Chr.15: 40252503-40252503 on Build GRCh38
rs1047283469 Chr.15: 40252195-40252195 on Build GRCh38
rs55974545 Chr.15: 40252319-40252319 on Build GRCh38
rs767970171 Chr.15: 40252643-40252643 on Build GRCh38
rs548218786 Chr.15: 40252563-40252563 on Build GRCh38
rs527893521 Chr.15: 40252338-40252338 on Build GRCh38
rs772386466 Chr.15: 40252310-40252310 on Build GRCh38
rs776112408 Chr.15: 40252645-40252645 on Build GRCh38
rs780852906 Chr.15: 40252268-40252268 on Build GRCh38
rs775828779 Chr.15: 40252412-40252412 on Build GRCh38
rs774867987 Chr.15: 40252636-40252636 on Build GRCh38
rs1027020778 Chr.15: 40252617-40252617 on Build GRCh38
rs758267511 Chr.15: 40252520-40252520 on Build GRCh38
rs535240512 Chr.15: 40252254-40252254 on Build GRCh38
rs750962796 Chr.15: 40252438-40252438 on Build GRCh38
rs574579676 Chr.15: 40252200-40252200 on Build GRCh38
rs368409250 Chr.15: 40252484-40252484 on Build GRCh38
rs776944954 Chr.15: 40252538-40252538 on Build GRCh38
rs982781177 Chr.15: 40252640-40252640 on Build GRCh38
rs908755805 Chr.15: 40252480-40252480 on Build GRCh38
rs759897213 Chr.15: 40252571-40252571 on Build GRCh38
rs970391269 Chr.15: 40252524-40252524 on Build GRCh38
rs200940821 Chr.15: 40252367-40252367 on Build GRCh38
rs1015338196 Chr.15: 40252448-40252448 on Build GRCh38
rs546368665 Chr.15: 40252376-40252376 on Build GRCh38
rs894586717 Chr.15: 40252612-40252612 on Build GRCh38
rs761588408 Chr.15: 40252646-40252646 on Build GRCh38
rs777493626 Chr.15: 40252495-40252495 on Build GRCh38
rs369650878 Chr.15: 40252582-40252582 on Build GRCh38
rs370638959 Chr.15: 40252306-40252306 on Build GRCh38
rs748128283 Chr.15: 40252541-40252541 on Build GRCh38
rs537704201 Chr.15: 40252606-40252606 on Build GRCh38
rs199772587 Chr.15: 40252649-40252649 on Build GRCh38
rs752341297 Chr.15: 40252462-40252462 on Build GRCh38
rs772010836 Chr.15: 40252404-40252404 on Build GRCh38
rs748442071 Chr.15: 40252613-40252613 on Build GRCh38
rs55799438 Chr.15: 40252292-40252292 on Build GRCh38
rs372004712 Chr.15: 40252252-40252252 on Build GRCh38
rs117614422 Chr.15: 40252330-40252330 on Build GRCh38
rs906365628 Chr.15: 40252357-40252357 on Build GRCh38
rs775863490 Chr.15: 40252311-40252311 on Build GRCh38
rs745846311 Chr.15: 40252517-40252517 on Build GRCh38
rs749252121 Chr.15: 40252287-40252287 on Build GRCh38
rs780775279 Chr.15: 40252352-40252352 on Build GRCh38
rs750689116 Chr.15: 40252341-40252341 on Build GRCh38
rs372777407 Chr.15: 40252569-40252569 on Build GRCh38
rs773338047 Chr.15: 40252621-40252621 on Build GRCh38
rs1053373850 Chr.15: 40252301-40252301 on Build GRCh38
rs868858964 Chr.15: 40252313-40252313 on Build GRCh38
rs566276564 Chr.15: 40252386-40252386 on Build GRCh38
rs944723251 Chr.15: 40252346-40252346 on Build GRCh38
rs780148206 Chr.15: 40252595-40252595 on Build GRCh38
rs896811391 Chr.15: 40252459-40252459 on Build GRCh38
rs376088458 Chr.15: 40252391-40252391 on Build GRCh38
rs372019677 Chr.15: 40252583-40252583 on Build GRCh38
rs749382893 Chr.15: 40252630-40252630 on Build GRCh38
rs754536400 Chr.15: 40252261-40252261 on Build GRCh38
rs768935793 Chr.15: 40252526-40252526 on Build GRCh38
rs759399859 Chr.15: 40252308-40252308 on Build GRCh38
rs753548560 Chr.15: 40252580-40252580 on Build GRCh38
rs765624068 Chr.15: 40252437-40252437 on Build GRCh38
rs769306575 Chr.15: 40252283-40252283 on Build GRCh38
rs763517941 Chr.15: 40252559-40252559 on Build GRCh38
rs1036200308 Chr.15: 40252372-40252372 on Build GRCh38
rs966016707 Chr.15: 40252626-40252626 on Build GRCh38
rs761049600 Chr.15: 40252414-40252414 on Build GRCh38
rs773561939 Chr.15: 40252551-40252551 on Build GRCh38
rs1015223378 Chr.15: 40252196-40252196 on Build GRCh38
rs778663383 Chr.15: 40252508-40252508 on Build GRCh38
rs776518212 Chr.15: 40252425-40252425 on Build GRCh38
rs577310277 Chr.15: 40252260-40252260 on Build GRCh38
rs752396383 Chr.15: 40252354-40252354 on Build GRCh38
rs983430667 Chr.15: 40252470-40252470 on Build GRCh38
rs749847018 Chr.15: 40252363-40252363 on Build GRCh38
rs184309892 Chr.15: 40252602-40252602 on Build GRCh38
rs746763061 Chr.15: 40252598-40252598 on Build GRCh38
rs969896778 Chr.15: 40252291-40252291 on Build GRCh38

View Full Product Details