Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40363539-40364040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DISP2
Entrez Gene ID
85455
Gene Name
dispatched RND transporter family member 2
Gene Aliases
DISPB, HsT16908, LINC00594
UniGene
Hs.355645
Gene Chromosome Location
Chr.15: 40354359-40378755 on Build GRCh38
Transcripts
NM_033510.2 XM_011522125.2 XM_017022687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7313755 c.212G>T p.G71V Chr.15: 40363717-40363717 on Build GRCh38
3969085 c.397C>A p.R133S Chr.15: 40363902-40363902 on Build GRCh38
5529750 c.150A>C p.A50A Chr.15: 40363655-40363655 on Build GRCh38
2269328 c.309delG p.G105fs*60 Chr.15: 40363814-40363814 on Build GRCh38
1203668 c.290G>A p.R97Q Chr.15: 40363795-40363795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775421210 Chr.15: 40363861-40363861 on Build GRCh38
rs766387287 Chr.15: 40363959-40363959 on Build GRCh38
rs779372854 Chr.15: 40364001-40364001 on Build GRCh38
rs747298491 Chr.15: 40363754-40363754 on Build GRCh38
rs777269574 Chr.15: 40363615-40363615 on Build GRCh38
rs111238199 Chr.15: 40363591-40363591 on Build GRCh38
rs200355085 Chr.15: 40363860-40363860 on Build GRCh38
rs772544294 Chr.15: 40363953-40363953 on Build GRCh38
rs1052981910 Chr.15: 40363700-40363700 on Build GRCh38
rs202103960 Chr.15: 40363612-40363612 on Build GRCh38
rs775609424 Chr.15: 40363605-40363605 on Build GRCh38
rs774643565 Chr.15: 40363849-40363849 on Build GRCh38
rs143063914 Chr.15: 40363904-40363904 on Build GRCh38
rs773758681 Chr.15: 40363845-40363845 on Build GRCh38
rs934791433 Chr.15: 40363995-40363995 on Build GRCh38
rs758173883 Chr.15: 40363751-40363751 on Build GRCh38
rs762582272 Chr.15: 40363957-40363957 on Build GRCh38
rs777648349 Chr.15: 40363916-40363916 on Build GRCh38
rs745637626 Chr.15: 40363741-40363741 on Build GRCh38
rs753355404 Chr.15: 40363727-40363727 on Build GRCh38
rs772292183 Chr.15: 40363601-40363601 on Build GRCh38
rs746810167 Chr.15: 40363815-40363815 on Build GRCh38
rs781595288 Chr.15: 40363836-40363836 on Build GRCh38
rs948380000 Chr.15: 40363939-40363939 on Build GRCh38
rs764718671 Chr.15: 40363873-40363873 on Build GRCh38
rs546489298 Chr.15: 40363999-40363999 on Build GRCh38
rs766714035 Chr.15: 40363786-40363786 on Build GRCh38
rs763699203 Chr.15: 40363861-40363861 on Build GRCh38
rs749400005 Chr.15: 40363925-40363925 on Build GRCh38
rs750749839 Chr.15: 40363656-40363656 on Build GRCh38
rs759920069 Chr.15: 40363859-40363859 on Build GRCh38
rs760255017 Chr.15: 40363791-40363791 on Build GRCh38
rs150939222 Chr.15: 40363902-40363902 on Build GRCh38
rs762737284 Chr.15: 40363895-40363895 on Build GRCh38
rs112633102 Chr.15: 40363643-40363644 on Build GRCh38
rs1898882 Chr.15: 40363672-40363672 on Build GRCh38
rs769001268 Chr.15: 40363757-40363757 on Build GRCh38
rs372047339 Chr.15: 40363968-40363968 on Build GRCh38
rs12443160 Chr.15: 40363940-40363940 on Build GRCh38
rs1898883 Chr.15: 40363644-40363644 on Build GRCh38
rs529851949 Chr.15: 40363984-40363984 on Build GRCh38
rs910592719 Chr.15: 40363603-40363603 on Build GRCh38
rs530696119 Chr.15: 40363926-40363926 on Build GRCh38
rs750724751 Chr.15: 40363804-40363804 on Build GRCh38
rs149658212 Chr.15: 40363905-40363905 on Build GRCh38
rs139247330 Chr.15: 40363794-40363794 on Build GRCh38
rs757425979 Chr.15: 40363765-40363765 on Build GRCh38
rs755202120 Chr.15: 40363818-40363818 on Build GRCh38
rs763256585 Chr.15: 40363660-40363660 on Build GRCh38
rs755662950 Chr.15: 40363579-40363579 on Build GRCh38
rs866611014 Chr.15: 40363716-40363716 on Build GRCh38
rs776308810 Chr.15: 40363865-40363865 on Build GRCh38
rs775119602 Chr.15: 40363787-40363787 on Build GRCh38
rs750382132 Chr.15: 40363809-40363809 on Build GRCh38
rs201394554 Chr.15: 40363813-40363813 on Build GRCh38
rs948416859 Chr.15: 40363631-40363631 on Build GRCh38
rs1040828649 Chr.15: 40364018-40364018 on Build GRCh38
rs748227152 Chr.15: 40363837-40363837 on Build GRCh38
rs368685493 Chr.15: 40363840-40363840 on Build GRCh38
rs755609441 Chr.15: 40363689-40363689 on Build GRCh38
rs777666589 Chr.15: 40363581-40363581 on Build GRCh38
rs544127960 Chr.15: 40363999-40363999 on Build GRCh38
rs146107339 Chr.15: 40363779-40363779 on Build GRCh38
rs765193725 Chr.15: 40363801-40363801 on Build GRCh38
rs752161326 Chr.15: 40363574-40363574 on Build GRCh38
rs757164133 Chr.15: 40363736-40363736 on Build GRCh38
rs776339006 Chr.15: 40363941-40363941 on Build GRCh38
rs756751597 Chr.15: 40363795-40363795 on Build GRCh38
rs763355413 Chr.15: 40363784-40363784 on Build GRCh38
rs778081583 Chr.15: 40363997-40363997 on Build GRCh38
rs905527822 Chr.15: 40363802-40363802 on Build GRCh38
rs749950163 Chr.15: 40363884-40363884 on Build GRCh38
rs540472041 Chr.15: 40363811-40363811 on Build GRCh38
rs756216107 Chr.15: 40363908-40363908 on Build GRCh38
rs867264440 Chr.15: 40363747-40363747 on Build GRCh38
rs761369737 Chr.15: 40363866-40363866 on Build GRCh38
rs745667583 Chr.15: 40363596-40363596 on Build GRCh38
rs778706980 Chr.15: 40363585-40363585 on Build GRCh38
rs35070171 Chr.15: 40363846-40363846 on Build GRCh38
rs756233199 Chr.15: 40363992-40363992 on Build GRCh38
rs751360468 Chr.15: 40363903-40363903 on Build GRCh38
rs144407194 Chr.15: 40363701-40363701 on Build GRCh38
rs374305547 Chr.15: 40363582-40363582 on Build GRCh38
rs770293316 Chr.15: 40363773-40363773 on Build GRCh38
rs371731373 Chr.15: 40363753-40363753 on Build GRCh38
rs553848134 Chr.15: 40363792-40363792 on Build GRCh38
rs200288583 Chr.15: 40363662-40363662 on Build GRCh38
rs778741100 Chr.15: 40363737-40363737 on Build GRCh38
rs560742995 Chr.15: 40363981-40363981 on Build GRCh38
rs769496877 Chr.15: 40363949-40363949 on Build GRCh38
rs767168573 Chr.15: 40363669-40363669 on Build GRCh38
rs375408757 Chr.15: 40363610-40363610 on Build GRCh38
rs201340864 Chr.15: 40363626-40363626 on Build GRCh38
rs759393095 Chr.15: 40363977-40363977 on Build GRCh38
rs572866435 Chr.15: 40363848-40363848 on Build GRCh38
rs763660511 Chr.15: 40363721-40363721 on Build GRCh38
rs753826127 Chr.15: 40363998-40363998 on Build GRCh38
rs746202282 Chr.15: 40363935-40363935 on Build GRCh38
rs748199427 Chr.15: 40363766-40363766 on Build GRCh38

View Full Product Details