Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40364330-40364834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DISP2
Entrez Gene ID
85455
Gene Name
dispatched RND transporter family member 2
Gene Aliases
DISPB, HsT16908, LINC00594
UniGene
Hs.355645
Gene Chromosome Location
Chr.15: 40354359-40378755 on Build GRCh38
Transcripts
NM_033510.2 XM_011522125.2 XM_017022687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6344412 c.514G>T p.V172L Chr.15: 40364455-40364455 on Build GRCh38
4054332 c.575G>T p.R192L Chr.15: 40364516-40364516 on Build GRCh38
4496918 c.485C>T p.S162F Chr.15: 40364426-40364426 on Build GRCh38
3969086 c.514G>A p.V172M Chr.15: 40364455-40364455 on Build GRCh38
6644436 c.494T>C p.I165T Chr.15: 40364435-40364435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766547080 Chr.15: 40364572-40364572 on Build GRCh38
rs775280557 Chr.15: 40364528-40364528 on Build GRCh38
rs115655873 Chr.15: 40364566-40364566 on Build GRCh38
rs949884158 Chr.15: 40364396-40364397 on Build GRCh38
rs931809905 Chr.15: 40364626-40364626 on Build GRCh38
rs531696813 Chr.15: 40364524-40364524 on Build GRCh38
rs1004846136 Chr.15: 40364400-40364400 on Build GRCh38
rs750849408 Chr.15: 40364762-40364762 on Build GRCh38
rs1050740637 Chr.15: 40364719-40364719 on Build GRCh38
rs770714527 Chr.15: 40364410-40364410 on Build GRCh38
rs921759481 Chr.15: 40364623-40364623 on Build GRCh38
rs141389410 Chr.15: 40364511-40364511 on Build GRCh38
rs966012604 Chr.15: 40364571-40364571 on Build GRCh38
rs200160916 Chr.15: 40364810-40364810 on Build GRCh38
rs940692650 Chr.15: 40364671-40364671 on Build GRCh38
rs752166813 Chr.15: 40364487-40364487 on Build GRCh38
rs758849095 Chr.15: 40364381-40364381 on Build GRCh38
rs369219150 Chr.15: 40364536-40364536 on Build GRCh38
rs377429654 Chr.15: 40364568-40364568 on Build GRCh38
rs189481944 Chr.15: 40364451-40364451 on Build GRCh38
rs969971500 Chr.15: 40364468-40364468 on Build GRCh38
rs755293586 Chr.15: 40364504-40364504 on Build GRCh38
rs755419593 Chr.15: 40364396-40364396 on Build GRCh38
rs1050815714 Chr.15: 40364464-40364464 on Build GRCh38
rs745393435 Chr.15: 40364414-40364414 on Build GRCh38
rs917051852 Chr.15: 40364390-40364390 on Build GRCh38
rs72733418 Chr.15: 40364523-40364523 on Build GRCh38
rs1006627431 Chr.15: 40364738-40364738 on Build GRCh38
rs941238297 Chr.15: 40364640-40364640 on Build GRCh38
rs371189111 Chr.15: 40364577-40364577 on Build GRCh38
rs1045743518 Chr.15: 40364756-40364756 on Build GRCh38
rs146996300 Chr.15: 40364516-40364516 on Build GRCh38
rs762927408 Chr.15: 40364569-40364569 on Build GRCh38
rs747304340 Chr.15: 40364389-40364389 on Build GRCh38
rs775541754 Chr.15: 40364421-40364421 on Build GRCh38
rs534252155 Chr.15: 40364622-40364622 on Build GRCh38
rs371358127 Chr.15: 40364558-40364558 on Build GRCh38
rs750460103 Chr.15: 40364455-40364455 on Build GRCh38
rs765464880 Chr.15: 40364454-40364454 on Build GRCh38
rs572020648 Chr.15: 40364357-40364357 on Build GRCh38
rs749017494 Chr.15: 40364405-40364405 on Build GRCh38
rs1036564456 Chr.15: 40364401-40364401 on Build GRCh38
rs757319729 Chr.15: 40364373-40364373 on Build GRCh38
rs971501115 Chr.15: 40364629-40364629 on Build GRCh38
rs771868462 Chr.15: 40364416-40364416 on Build GRCh38
rs900671551 Chr.15: 40364752-40364752 on Build GRCh38
rs762027272 Chr.15: 40364670-40364670 on Build GRCh38
rs918220588 Chr.15: 40364676-40364676 on Build GRCh38
rs1008999907 Chr.15: 40364492-40364492 on Build GRCh38
rs890598434 Chr.15: 40364484-40364484 on Build GRCh38
rs773941546 Chr.15: 40364412-40364412 on Build GRCh38
rs932115422 Chr.15: 40364354-40364354 on Build GRCh38
rs890861646 Chr.15: 40364730-40364730 on Build GRCh38
rs956514157 Chr.15: 40364812-40364812 on Build GRCh38
rs1038442355 Chr.15: 40364443-40364443 on Build GRCh38
rs200567942 Chr.15: 40364378-40364379 on Build GRCh38
rs748537315 Chr.15: 40364513-40364513 on Build GRCh38
rs979925047 Chr.15: 40364630-40364630 on Build GRCh38
rs753073326 Chr.15: 40364591-40364591 on Build GRCh38
rs776636023 Chr.15: 40364587-40364587 on Build GRCh38
rs750513008 Chr.15: 40364379-40364379 on Build GRCh38
rs778688950 Chr.15: 40364595-40364595 on Build GRCh38
rs758670679 Chr.15: 40364778-40364778 on Build GRCh38
rs375725325 Chr.15: 40364458-40364458 on Build GRCh38
rs532623947 Chr.15: 40364515-40364515 on Build GRCh38
rs1036342251 Chr.15: 40364679-40364679 on Build GRCh38
rs909334771 Chr.15: 40364667-40364667 on Build GRCh38
rs560118609 Chr.15: 40364503-40364503 on Build GRCh38
rs181747268 Chr.15: 40364639-40364639 on Build GRCh38
rs371442025 Chr.15: 40364398-40364398 on Build GRCh38
rs896715411 Chr.15: 40364717-40364717 on Build GRCh38
rs372573100 Chr.15: 40364376-40364376 on Build GRCh38
rs377321174 Chr.15: 40364800-40364800 on Build GRCh38
rs376884591 Chr.15: 40364382-40364382 on Build GRCh38
rs764010466 Chr.15: 40364426-40364426 on Build GRCh38
rs767654662 Chr.15: 40364653-40364654 on Build GRCh38
rs1001270381 Chr.15: 40364512-40364512 on Build GRCh38
rs776356427 Chr.15: 40364564-40364564 on Build GRCh38
rs769968876 Chr.15: 40364567-40364567 on Build GRCh38
rs757846540 Chr.15: 40364801-40364801 on Build GRCh38
rs745645157 Chr.15: 40364509-40364509 on Build GRCh38
rs766771584 Chr.15: 40364479-40364479 on Build GRCh38
rs760367167 Chr.15: 40364424-40364424 on Build GRCh38
rs200805831 Chr.15: 40364430-40364430 on Build GRCh38
rs760623477 Chr.15: 40364575-40364575 on Build GRCh38
rs756589810 Chr.15: 40364594-40364594 on Build GRCh38
rs142651190 Chr.15: 40364585-40364585 on Build GRCh38
rs779555159 Chr.15: 40364802-40364802 on Build GRCh38
rs747038182 Chr.15: 40364534-40364534 on Build GRCh38
rs765444543 Chr.15: 40364749-40364749 on Build GRCh38

View Full Product Details