Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48202931-48203479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTXN2
Entrez Gene ID
399697
Gene Name
cortexin 2
Gene Aliases
-
UniGene
Hs.721408
Gene Chromosome Location
Chr.15: 48178564-48203754 on Build GRCh38
Transcripts
NM_001145668.1

SNP Detail

SNP ID Chromosome Location
rs903434120 Chr.15: 48203062-48203062 on Build GRCh38
rs192951038 Chr.15: 48203283-48203283 on Build GRCh38
rs755368406 Chr.15: 48203033-48203033 on Build GRCh38
rs879131365 Chr.15: 48203036-48203036 on Build GRCh38
rs921400784 Chr.15: 48203349-48203349 on Build GRCh38
rs755243011 Chr.15: 48203076-48203076 on Build GRCh38
rs1052168708 Chr.15: 48203025-48203025 on Build GRCh38
rs912255389 Chr.15: 48203306-48203306 on Build GRCh38
rs1054539535 Chr.15: 48203070-48203070 on Build GRCh38
rs1055479365 Chr.15: 48202999-48202999 on Build GRCh38
rs781511485 Chr.15: 48203137-48203137 on Build GRCh38
rs1035722071 Chr.15: 48203191-48203191 on Build GRCh38
rs960092575 Chr.15: 48203206-48203206 on Build GRCh38
rs1039510288 Chr.15: 48203326-48203326 on Build GRCh38
rs950340042 Chr.15: 48203039-48203039 on Build GRCh38
rs943797533 Chr.15: 48203061-48203061 on Build GRCh38
rs575770717 Chr.15: 48203279-48203279 on Build GRCh38
rs185557876 Chr.15: 48203358-48203358 on Build GRCh38
rs147434428 Chr.15: 48203295-48203295 on Build GRCh38
rs750279055 Chr.15: 48203012-48203012 on Build GRCh38
rs1039877329 Chr.15: 48203075-48203075 on Build GRCh38
rs1017562864 Chr.15: 48203243-48203243 on Build GRCh38
rs1046440337 Chr.15: 48203045-48203045 on Build GRCh38
rs887343520 Chr.15: 48203384-48203385 on Build GRCh38
rs915658568 Chr.15: 48203436-48203436 on Build GRCh38
rs139812569 Chr.15: 48203413-48203413 on Build GRCh38
rs1320052 Chr.15: 48203311-48203311 on Build GRCh38
rs940305955 Chr.15: 48203422-48203422 on Build GRCh38
rs867043474 Chr.15: 48203222-48203222 on Build GRCh38
rs893236425 Chr.15: 48203088-48203088 on Build GRCh38
rs756181537 Chr.15: 48203260-48203260 on Build GRCh38
rs559366462 Chr.15: 48203403-48203403 on Build GRCh38
rs982639041 Chr.15: 48203298-48203298 on Build GRCh38
rs962468734 Chr.15: 48203344-48203344 on Build GRCh38
rs991195607 Chr.15: 48203345-48203345 on Build GRCh38
rs1007575291 Chr.15: 48203141-48203141 on Build GRCh38
rs999454925 Chr.15: 48203069-48203069 on Build GRCh38
rs995768433 Chr.15: 48203133-48203133 on Build GRCh38
rs900042996 Chr.15: 48203091-48203091 on Build GRCh38
rs956372320 Chr.15: 48203278-48203278 on Build GRCh38
rs979791898 Chr.15: 48202959-48202959 on Build GRCh38
rs1049130868 Chr.15: 48203139-48203139 on Build GRCh38
rs748390034 Chr.15: 48203223-48203223 on Build GRCh38
rs1004632223 Chr.15: 48203183-48203183 on Build GRCh38
rs941074023 Chr.15: 48202967-48202967 on Build GRCh38
rs75445284 Chr.15: 48203261-48203261 on Build GRCh38
rs924937221 Chr.15: 48202963-48202963 on Build GRCh38
rs887077468 Chr.15: 48203177-48203177 on Build GRCh38
rs1023416596 Chr.15: 48203233-48203233 on Build GRCh38
rs925208791 Chr.15: 48203264-48203264 on Build GRCh38

View Full Product Details