Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48408961-48409485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBN1
Entrez Gene ID
2200
Gene Name
fibrillin 1
Gene Aliases
ACMICD, ECTOL1, FBN, GPHYSD2, MASS, MFLS, MFS1, OCTD, SGS, SSKS, WMS, WMS2
UniGene
Hs.591133
Gene Chromosome Location
Chr.15: 48408306-48645788 on Build GRCh38
Transcripts
NM_000138.4

SNP Detail

SNP ID Chromosome Location
rs1054860347 Chr.15: 48409009-48409009 on Build GRCh38
rs56194244 Chr.15: 48409001-48409001 on Build GRCh38
rs992846436 Chr.15: 48409148-48409148 on Build GRCh38
rs557820296 Chr.15: 48409408-48409408 on Build GRCh38
rs35929392 Chr.15: 48409263-48409264 on Build GRCh38
rs763023832 Chr.15: 48409114-48409114 on Build GRCh38
rs539021839 Chr.15: 48409269-48409269 on Build GRCh38
rs962748908 Chr.15: 48409192-48409192 on Build GRCh38
rs936258673 Chr.15: 48409211-48409211 on Build GRCh38
rs963155212 Chr.15: 48409446-48409446 on Build GRCh38
rs909998204 Chr.15: 48409165-48409165 on Build GRCh38
rs761904999 Chr.15: 48409457-48409457 on Build GRCh38
rs886051231 Chr.15: 48409318-48409318 on Build GRCh38
rs1029839086 Chr.15: 48409413-48409413 on Build GRCh38
rs887643640 Chr.15: 48409343-48409343 on Build GRCh38
rs183652996 Chr.15: 48409058-48409058 on Build GRCh38
rs886051229 Chr.15: 48409047-48409047 on Build GRCh38
rs557225449 Chr.15: 48409429-48409429 on Build GRCh38
rs4775760 Chr.15: 48409415-48409415 on Build GRCh38
rs927633594 Chr.15: 48409275-48409275 on Build GRCh38
rs545437535 Chr.15: 48409458-48409458 on Build GRCh38
rs1024355910 Chr.15: 48409151-48409151 on Build GRCh38
rs551933552 Chr.15: 48408987-48408987 on Build GRCh38
rs374297351 Chr.15: 48409371-48409371 on Build GRCh38
rs1034349142 Chr.15: 48409105-48409105 on Build GRCh38
rs897587338 Chr.15: 48409318-48409318 on Build GRCh38
rs78442438 Chr.15: 48409428-48409428 on Build GRCh38
rs1029618446 Chr.15: 48409146-48409146 on Build GRCh38
rs187799549 Chr.15: 48408993-48408993 on Build GRCh38
rs1002106856 Chr.15: 48409347-48409347 on Build GRCh38
rs573501850 Chr.15: 48408999-48408999 on Build GRCh38
rs886051230 Chr.15: 48409257-48409257 on Build GRCh38
rs1034030045 Chr.15: 48409392-48409392 on Build GRCh38
rs1046521878 Chr.15: 48408990-48408990 on Build GRCh38
rs1042956547 Chr.15: 48409371-48409372 on Build GRCh38
rs977560524 Chr.15: 48409178-48409178 on Build GRCh38
rs12050562 Chr.15: 48409031-48409031 on Build GRCh38
rs535798341 Chr.15: 48409039-48409040 on Build GRCh38
rs894451140 Chr.15: 48409394-48409394 on Build GRCh38
rs552538828 Chr.15: 48409345-48409346 on Build GRCh38
rs534577080 Chr.15: 48409041-48409041 on Build GRCh38
rs568791870 Chr.15: 48409219-48409219 on Build GRCh38
rs770825180 Chr.15: 48409284-48409284 on Build GRCh38
rs572619690 Chr.15: 48409274-48409274 on Build GRCh38
rs893584590 Chr.15: 48409346-48409346 on Build GRCh38
rs929224138 Chr.15: 48409230-48409230 on Build GRCh38
rs774344373 Chr.15: 48409380-48409380 on Build GRCh38
rs189534565 Chr.15: 48409213-48409213 on Build GRCh38
rs886051232 Chr.15: 48409355-48409355 on Build GRCh38
rs929086444 Chr.15: 48409323-48409323 on Build GRCh38
rs897639741 Chr.15: 48409030-48409030 on Build GRCh38
rs370279934 Chr.15: 48409065-48409065 on Build GRCh38
rs967483418 Chr.15: 48409164-48409164 on Build GRCh38
rs919089483 Chr.15: 48409218-48409218 on Build GRCh38
rs772970380 Chr.15: 48409179-48409179 on Build GRCh38
rs557301792 Chr.15: 48409270-48409270 on Build GRCh38
rs1048886533 Chr.15: 48409340-48409340 on Build GRCh38
rs574169581 Chr.15: 48409411-48409411 on Build GRCh38
rs1027952961 Chr.15: 48409039-48409039 on Build GRCh38
rs17352989 Chr.15: 48409410-48409410 on Build GRCh38

View Full Product Details