Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49155404-49155942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS2
Entrez Gene ID
9318
Gene Name
COP9 signalosome subunit 2
Gene Aliases
ALIEN, CSN2, SGN2, TRIP15
UniGene
Hs.369614
Gene Chromosome Location
Chr.15: 49125274-49155657 on Build GRCh38
Transcripts
NM_001143887.1 NM_004236.3 NM_001001556.2 XM_005254280.3 XM_005254284.3 XM_017022063.1 XM_017022065.1

Gene Symbol
GALK2
Entrez Gene ID
2585
Gene Name
galactokinase 2
Gene Aliases
GK2
UniGene
Hs.122006
Gene Chromosome Location
Chr.15: 49155757-49367883 on Build GRCh38
Transcripts
NM_001143887.1 NM_004236.3 NM_001001556.2 XM_005254280.3 XM_005254284.3 XM_017022063.1 XM_017022065.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1317778 c.28T>G p.C10G Chr.15: 49155551-49155551 on Build GRCh38
4055398 c.44A>G p.D15G Chr.15: 49155535-49155535 on Build GRCh38
1629583 c.34G>A p.D12N Chr.15: 49155545-49155545 on Build GRCh38
6574227 c.54+1G>C p.? Chr.15: 49155524-49155524 on Build GRCh38
3969141 c.5C>G p.S2C Chr.15: 49155574-49155574 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1012993924 Chr.15: 49155667-49155667 on Build GRCh38
rs886230769 Chr.15: 49155735-49155735 on Build GRCh38
rs761285117 Chr.15: 49155540-49155540 on Build GRCh38
rs943014205 Chr.15: 49155713-49155713 on Build GRCh38
rs576328029 Chr.15: 49155588-49155588 on Build GRCh38
rs369648099 Chr.15: 49155596-49155596 on Build GRCh38
rs930420986 Chr.15: 49155725-49155725 on Build GRCh38
rs983040225 Chr.15: 49155794-49155794 on Build GRCh38
rs768760936 Chr.15: 49155715-49155715 on Build GRCh38
rs767136618 Chr.15: 49155505-49155505 on Build GRCh38
rs141286814 Chr.15: 49155636-49155636 on Build GRCh38
rs535454664 Chr.15: 49155789-49155789 on Build GRCh38
rs561223379 Chr.15: 49155674-49155674 on Build GRCh38
rs116224830 Chr.15: 49155762-49155762 on Build GRCh38
rs759205042 Chr.15: 49155590-49155590 on Build GRCh38
rs750448754 Chr.15: 49155617-49155617 on Build GRCh38
rs890081569 Chr.15: 49155451-49155451 on Build GRCh38
rs773694962 Chr.15: 49155501-49155501 on Build GRCh38
rs755145333 Chr.15: 49155569-49155569 on Build GRCh38
rs531010743 Chr.15: 49155751-49155751 on Build GRCh38
rs201706108 Chr.15: 49155484-49155484 on Build GRCh38
rs547021746 Chr.15: 49155473-49155473 on Build GRCh38
rs778108589 Chr.15: 49155765-49155765 on Build GRCh38
rs930326681 Chr.15: 49155877-49155877 on Build GRCh38
rs766913168 Chr.15: 49155494-49155494 on Build GRCh38
rs892074673 Chr.15: 49155909-49155909 on Build GRCh38
rs956485687 Chr.15: 49155731-49155731 on Build GRCh38
rs907826039 Chr.15: 49155796-49155796 on Build GRCh38
rs934848922 Chr.15: 49155649-49155649 on Build GRCh38
rs146730067 Chr.15: 49155811-49155811 on Build GRCh38
rs943045224 Chr.15: 49155464-49155464 on Build GRCh38
rs920292302 Chr.15: 49155876-49155876 on Build GRCh38
rs974370112 Chr.15: 49155837-49155837 on Build GRCh38
rs545963292 Chr.15: 49155640-49155640 on Build GRCh38
rs1044514143 Chr.15: 49155849-49155849 on Build GRCh38
rs773381969 Chr.15: 49155587-49155587 on Build GRCh38
rs373052719 Chr.15: 49155512-49155512 on Build GRCh38
rs559327432 Chr.15: 49155647-49155647 on Build GRCh38
rs971407652 Chr.15: 49155688-49155688 on Build GRCh38
rs956247760 Chr.15: 49155455-49155455 on Build GRCh38
rs763856167 Chr.15: 49155534-49155534 on Build GRCh38
rs1051376631 Chr.15: 49155449-49155449 on Build GRCh38
rs577087676 Chr.15: 49155826-49155826 on Build GRCh38
rs371972527 Chr.15: 49155586-49155586 on Build GRCh38
rs964959638 Chr.15: 49155831-49155831 on Build GRCh38
rs755900606 Chr.15: 49155530-49155530 on Build GRCh38
rs145039314 Chr.15: 49155737-49155737 on Build GRCh38
rs151074018 Chr.15: 49155584-49155584 on Build GRCh38
rs1023040132 Chr.15: 49155607-49155607 on Build GRCh38
rs369810251 Chr.15: 49155625-49155625 on Build GRCh38
rs947848816 Chr.15: 49155827-49155827 on Build GRCh38
rs914365677 Chr.15: 49155476-49155476 on Build GRCh38
rs77933182 Chr.15: 49155590-49155590 on Build GRCh38
rs1047899412 Chr.15: 49155880-49155880 on Build GRCh38
rs1026592890 Chr.15: 49155570-49155570 on Build GRCh38
rs748437187 Chr.15: 49155585-49155585 on Build GRCh38
rs536586238 Chr.15: 49155568-49155568 on Build GRCh38
rs191066853 Chr.15: 49155591-49155591 on Build GRCh38
rs767230489 Chr.15: 49155816-49155816 on Build GRCh38
rs762233437 Chr.15: 49155475-49155475 on Build GRCh38
rs924875221 Chr.15: 49155436-49155436 on Build GRCh38
rs990365612 Chr.15: 49155750-49155750 on Build GRCh38
rs757120988 Chr.15: 49155614-49155614 on Build GRCh38
rs751316748 Chr.15: 49155726-49155726 on Build GRCh38
rs990682323 Chr.15: 49155470-49155470 on Build GRCh38
rs774221875 Chr.15: 49155720-49155720 on Build GRCh38
rs569927295 Chr.15: 49155806-49155806 on Build GRCh38
rs753683808 Chr.15: 49155537-49155537 on Build GRCh38
rs60899981 Chr.15: 49155886-49155886 on Build GRCh38
rs762600614 Chr.15: 49155677-49155677 on Build GRCh38
rs17435932 Chr.15: 49155815-49155815 on Build GRCh38
rs552396498 Chr.15: 49155754-49155754 on Build GRCh38
rs753540257 Chr.15: 49155603-49155603 on Build GRCh38
rs182143807 Chr.15: 49155696-49155696 on Build GRCh38
rs568457747 Chr.15: 49155772-49155772 on Build GRCh38
rs934942855 Chr.15: 49155445-49155445 on Build GRCh38
rs757398343 Chr.15: 49155841-49155841 on Build GRCh38
rs907254706 Chr.15: 49155869-49155869 on Build GRCh38
rs115915876 Chr.15: 49155624-49155624 on Build GRCh38
rs765050392 Chr.15: 49155616-49155616 on Build GRCh38
rs752390101 Chr.15: 49155522-49155522 on Build GRCh38
rs759069771 Chr.15: 49155503-49155503 on Build GRCh38
rs981675533 Chr.15: 49155443-49155443 on Build GRCh38
rs774002549 Chr.15: 49155589-49155590 on Build GRCh38
rs79788893 Chr.15: 49155882-49155882 on Build GRCh38
rs745526465 Chr.15: 49155447-49155447 on Build GRCh38
rs763310748 Chr.15: 49155488-49155488 on Build GRCh38
rs760265463 Chr.15: 49155594-49155594 on Build GRCh38
rs898614974 Chr.15: 49155648-49155648 on Build GRCh38
rs376158843 Chr.15: 49155793-49155793 on Build GRCh38
rs778927689 Chr.15: 49155564-49155564 on Build GRCh38
rs546765497 Chr.15: 49155763-49155763 on Build GRCh38
rs757080604 Chr.15: 49155554-49155554 on Build GRCh38
rs971671099 Chr.15: 49155427-49155427 on Build GRCh38
rs924836054 Chr.15: 49155444-49155444 on Build GRCh38
rs564338389 Chr.15: 49155759-49155759 on Build GRCh38
rs898928324 Chr.15: 49155483-49155483 on Build GRCh38
rs894956211 Chr.15: 49155821-49155821 on Build GRCh38
rs534747533 Chr.15: 49155861-49155861 on Build GRCh38
rs922994078 Chr.15: 49155775-49155775 on Build GRCh38

View Full Product Details