Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49620597-49621110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTWD1
Entrez Gene ID
56986
Gene Name
DTW domain containing 1
Gene Aliases
MDS009
UniGene
Hs.127432
Gene Chromosome Location
Chr.15: 49620980-49654448 on Build GRCh38
Transcripts
NM_001144955.1 NM_020234.5 XM_011521815.1 XM_017022419.1 XM_017022421.1 XM_017022423.1 XM_017022424.1 XM_017022425.1 XM_017022427.1 XM_017022428.1 XM_017022430.1 NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_005254220.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521321.2 XM_011521324.1 XM_011521325.2 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
UniGene
Hs.712959
Gene Chromosome Location
Chr.15: 49326997-49620931 on Build GRCh38
Transcripts
NM_001144955.1 NM_020234.5 XM_011521815.1 XM_017022419.1 XM_017022421.1 XM_017022423.1 XM_017022424.1 XM_017022425.1 XM_017022427.1 XM_017022428.1 XM_017022430.1 NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_005254220.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521321.2 XM_011521324.1 XM_011521325.2 XM_011521326.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1

SNP Detail

SNP ID Chromosome Location
rs1015896836 Chr.15: 49620825-49620825 on Build GRCh38
rs530659491 Chr.15: 49621035-49621035 on Build GRCh38
rs186303270 Chr.15: 49620983-49620983 on Build GRCh38
rs1040211856 Chr.15: 49620949-49620949 on Build GRCh38
rs1033344500 Chr.15: 49621063-49621063 on Build GRCh38
rs535962252 Chr.15: 49621088-49621088 on Build GRCh38
rs565745381 Chr.15: 49620661-49620661 on Build GRCh38
rs746808022 Chr.15: 49620907-49620908 on Build GRCh38
rs867021231 Chr.15: 49620948-49620948 on Build GRCh38
rs368516561 Chr.15: 49620821-49620821 on Build GRCh38
rs972177866 Chr.15: 49620639-49620639 on Build GRCh38
rs1037484803 Chr.15: 49620909-49620910 on Build GRCh38
rs1037854180 Chr.15: 49620904-49620904 on Build GRCh38
rs142111670 Chr.15: 49621070-49621070 on Build GRCh38
rs1008685773 Chr.15: 49620857-49620857 on Build GRCh38
rs1041949007 Chr.15: 49620809-49620809 on Build GRCh38
rs191983039 Chr.15: 49621016-49621016 on Build GRCh38
rs977956258 Chr.15: 49621087-49621087 on Build GRCh38
rs887656836 Chr.15: 49621030-49621030 on Build GRCh38
rs1028681704 Chr.15: 49620622-49620622 on Build GRCh38
rs914620537 Chr.15: 49620901-49620902 on Build GRCh38
rs914818775 Chr.15: 49620807-49620807 on Build GRCh38
rs954705390 Chr.15: 49620976-49620976 on Build GRCh38
rs1024877512 Chr.15: 49620858-49620858 on Build GRCh38
rs962006620 Chr.15: 49620829-49620829 on Build GRCh38
rs576701868 Chr.15: 49620819-49620819 on Build GRCh38
rs774227283 Chr.15: 49620892-49620907 on Build GRCh38
rs926336448 Chr.15: 49620757-49620757 on Build GRCh38
rs1050481482 Chr.15: 49620816-49620816 on Build GRCh38
rs963590741 Chr.15: 49620993-49620993 on Build GRCh38
rs923525873 Chr.15: 49620908-49620909 on Build GRCh38
rs972315650 Chr.15: 49620727-49620727 on Build GRCh38
rs1011327931 Chr.15: 49620830-49620830 on Build GRCh38
rs113956008 Chr.15: 49620929-49620929 on Build GRCh38
rs1016612079 Chr.15: 49620708-49620708 on Build GRCh38
rs1043785075 Chr.15: 49620974-49620974 on Build GRCh38
rs1005858218 Chr.15: 49620650-49620650 on Build GRCh38
rs559458700 Chr.15: 49620951-49620951 on Build GRCh38
rs951915516 Chr.15: 49620681-49620681 on Build GRCh38
rs910781483 Chr.15: 49620985-49620986 on Build GRCh38
rs1057287107 Chr.15: 49620785-49620785 on Build GRCh38
rs926368925 Chr.15: 49620905-49620905 on Build GRCh38
rs77933109 Chr.15: 49620964-49620964 on Build GRCh38
rs1000714148 Chr.15: 49620813-49620813 on Build GRCh38
rs117894747 Chr.15: 49620909-49620909 on Build GRCh38
rs368959908 Chr.15: 49620905-49620907 on Build GRCh38
rs1042146086 Chr.15: 49620808-49620808 on Build GRCh38
rs367663198 Chr.15: 49620801-49620801 on Build GRCh38
rs1018729396 Chr.15: 49620860-49620860 on Build GRCh38
rs541065479 Chr.15: 49620938-49620938 on Build GRCh38
rs377694323 Chr.15: 49620905-49620906 on Build GRCh38
rs549868613 Chr.15: 49620738-49620738 on Build GRCh38
rs958109694 Chr.15: 49620906-49620907 on Build GRCh38
rs16962664 Chr.15: 49620749-49620749 on Build GRCh38
rs764127788 Chr.15: 49620669-49620669 on Build GRCh38
rs960479732 Chr.15: 49621065-49621065 on Build GRCh38
rs895125720 Chr.15: 49620805-49620805 on Build GRCh38
rs113483771 Chr.15: 49620897-49620897 on Build GRCh38
rs962303390 Chr.15: 49620874-49620874 on Build GRCh38
rs927346793 Chr.15: 49620797-49620797 on Build GRCh38
rs1006302072 Chr.15: 49620824-49620824 on Build GRCh38
rs749429826 Chr.15: 49620831-49620831 on Build GRCh38
rs917789666 Chr.15: 49620730-49620730 on Build GRCh38
rs961948298 Chr.15: 49620715-49620715 on Build GRCh38
rs929075189 Chr.15: 49621059-49621059 on Build GRCh38
rs897511344 Chr.15: 49620904-49620905 on Build GRCh38
rs944783483 Chr.15: 49620930-49620930 on Build GRCh38
rs372818436 Chr.15: 49620856-49620856 on Build GRCh38
rs574951279 Chr.15: 49620766-49620766 on Build GRCh38
rs970575897 Chr.15: 49620872-49620873 on Build GRCh38
rs376212407 Chr.15: 49620959-49620959 on Build GRCh38
rs111673878 Chr.15: 49620901-49620901 on Build GRCh38
rs1027585235 Chr.15: 49621008-49621008 on Build GRCh38
rs559330901 Chr.15: 49620926-49620926 on Build GRCh38
rs998818912 Chr.15: 49620952-49620952 on Build GRCh38
rs867641913 Chr.15: 49620910-49620910 on Build GRCh38
rs374915719 Chr.15: 49620908-49620908 on Build GRCh38
rs565871006 Chr.15: 49620812-49620812 on Build GRCh38
rs114332954 Chr.15: 49620906-49620906 on Build GRCh38
rs1055066472 Chr.15: 49621080-49621080 on Build GRCh38
rs377490821 Chr.15: 49620900-49620901 on Build GRCh38
rs1050345054 Chr.15: 49620928-49620928 on Build GRCh38
rs888847038 Chr.15: 49620818-49620818 on Build GRCh38
rs541462775 Chr.15: 49620921-49620921 on Build GRCh38
rs564056345 Chr.15: 49620902-49620903 on Build GRCh38
rs967592350 Chr.15: 49621076-49621076 on Build GRCh38
rs113302861 Chr.15: 49620907-49620907 on Build GRCh38
rs1034438004 Chr.15: 49620877-49620877 on Build GRCh38
rs981544963 Chr.15: 49620751-49620751 on Build GRCh38
rs991896242 Chr.15: 49621069-49621069 on Build GRCh38
rs1041957682 Chr.15: 49620911-49620911 on Build GRCh38
rs906191838 Chr.15: 49620910-49620911 on Build GRCh38
rs926177482 Chr.15: 49620916-49620916 on Build GRCh38
rs374731292 Chr.15: 49620902-49620902 on Build GRCh38
rs973956073 Chr.15: 49621021-49621021 on Build GRCh38
rs576609446 Chr.15: 49620903-49620903 on Build GRCh38
rs375731889 Chr.15: 49620804-49620804 on Build GRCh38
rs897638496 Chr.15: 49620953-49620953 on Build GRCh38
rs16962663 Chr.15: 49620633-49620633 on Build GRCh38
rs993413782 Chr.15: 49620876-49620876 on Build GRCh38
rs1021731261 Chr.15: 49620936-49620936 on Build GRCh38

View Full Product Details