Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51209429-51209944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP19A1
Entrez Gene ID
1588
Gene Name
cytochrome P450 family 19 subfamily A member 1
Gene Aliases
ARO, ARO1, CPV1, CYAR, CYP19, CYPXIX, P-450AROM
UniGene
Hs.260074
Gene Chromosome Location
Chr.15: 51208057-51338598 on Build GRCh38
Transcripts
NM_000103.3 NM_031226.2 XM_005254191.1

SNP Detail

SNP ID Chromosome Location
rs916577936 Chr.15: 51209834-51209834 on Build GRCh38
rs988879442 Chr.15: 51209524-51209524 on Build GRCh38
rs964492469 Chr.15: 51209660-51209660 on Build GRCh38
rs551590351 Chr.15: 51209546-51209546 on Build GRCh38
rs770994265 Chr.15: 51209801-51209801 on Build GRCh38
rs999207741 Chr.15: 51209594-51209594 on Build GRCh38
rs1055082080 Chr.15: 51209814-51209814 on Build GRCh38
rs772138757 Chr.15: 51209605-51209605 on Build GRCh38
rs1055137305 Chr.15: 51209487-51209487 on Build GRCh38
rs577509190 Chr.15: 51209800-51209800 on Build GRCh38
rs1019452336 Chr.15: 51209915-51209915 on Build GRCh38
rs950882104 Chr.15: 51209772-51209772 on Build GRCh38
rs1013615290 Chr.15: 51209540-51209540 on Build GRCh38
rs975828739 Chr.15: 51209738-51209738 on Build GRCh38
rs527275486 Chr.15: 51209739-51209739 on Build GRCh38
rs754516277 Chr.15: 51209766-51209766 on Build GRCh38
rs28757211 Chr.15: 51209569-51209569 on Build GRCh38
rs966452661 Chr.15: 51209541-51209541 on Build GRCh38
rs907017185 Chr.15: 51209577-51209577 on Build GRCh38
rs561231361 Chr.15: 51209776-51209776 on Build GRCh38
rs1803153 Chr.15: 51209704-51209704 on Build GRCh38
rs574201730 Chr.15: 51209477-51209477 on Build GRCh38
rs940823899 Chr.15: 51209637-51209637 on Build GRCh38
rs984125253 Chr.15: 51209789-51209789 on Build GRCh38
rs542540772 Chr.15: 51209775-51209775 on Build GRCh38
rs1050787 Chr.15: 51209885-51209885 on Build GRCh38
rs375328204 Chr.15: 51209617-51209617 on Build GRCh38
rs1054282186 Chr.15: 51209857-51209857 on Build GRCh38
rs966114821 Chr.15: 51209919-51209919 on Build GRCh38
rs533861105 Chr.15: 51209606-51209606 on Build GRCh38
rs566312936 Chr.15: 51209602-51209602 on Build GRCh38
rs28757210 Chr.15: 51209618-51209618 on Build GRCh38
rs1032840751 Chr.15: 51209502-51209502 on Build GRCh38
rs559731751 Chr.15: 51209668-51209668 on Build GRCh38
rs891646590 Chr.15: 51209881-51209881 on Build GRCh38
rs28757212 Chr.15: 51209512-51209512 on Build GRCh38
rs949761088 Chr.15: 51209798-51209798 on Build GRCh38
rs544673636 Chr.15: 51209636-51209636 on Build GRCh38
rs905747200 Chr.15: 51209469-51209469 on Build GRCh38
rs1009994682 Chr.15: 51209907-51209907 on Build GRCh38
rs886051274 Chr.15: 51209789-51209789 on Build GRCh38
rs909425068 Chr.15: 51209796-51209796 on Build GRCh38
rs949495005 Chr.15: 51209902-51209902 on Build GRCh38
rs1019002824 Chr.15: 51209532-51209532 on Build GRCh38
rs773502422 Chr.15: 51209570-51209570 on Build GRCh38
rs942009829 Chr.15: 51209463-51209463 on Build GRCh38
rs1024597772 Chr.15: 51209559-51209559 on Build GRCh38

View Full Product Details