Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51227787-51228290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP19A1
Entrez Gene ID
1588
Gene Name
cytochrome P450 family 19 subfamily A member 1
Gene Aliases
ARO, ARO1, CPV1, CYAR, CYP19, CYPXIX, P-450AROM
UniGene
Hs.260074
Gene Chromosome Location
Chr.15: 51208057-51338598 on Build GRCh38
Transcripts
NM_000103.3 NM_031226.2 XM_005254191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4493292 c.412C>T p.P138S Chr.15: 51227818-51227818 on Build GRCh38
3956724 c.379A>G p.M127V Chr.15: 51227851-51227851 on Build GRCh38
6573463 c.387G>C p.E129D Chr.15: 51227843-51227843 on Build GRCh38
6030695 c.368A>G p.Q123R Chr.15: 51227862-51227862 on Build GRCh38
2216885 c.331C>A p.H111N Chr.15: 51227899-51227899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143453811 Chr.15: 51228151-51228151 on Build GRCh38
rs375975652 Chr.15: 51227847-51227847 on Build GRCh38
rs888511774 Chr.15: 51228111-51228111 on Build GRCh38
rs1013882880 Chr.15: 51228148-51228148 on Build GRCh38
rs755633611 Chr.15: 51227815-51227815 on Build GRCh38
rs770701615 Chr.15: 51227844-51227844 on Build GRCh38
rs111555509 Chr.15: 51228110-51228110 on Build GRCh38
rs572169084 Chr.15: 51228167-51228167 on Build GRCh38
rs774271716 Chr.15: 51227853-51227854 on Build GRCh38
rs763223426 Chr.15: 51227893-51227893 on Build GRCh38
rs1033921193 Chr.15: 51228127-51228127 on Build GRCh38
rs368138645 Chr.15: 51227886-51227886 on Build GRCh38
rs745865546 Chr.15: 51227896-51227918 on Build GRCh38
rs1065778 Chr.15: 51228009-51228009 on Build GRCh38
rs925468485 Chr.15: 51227939-51227939 on Build GRCh38
rs552093024 Chr.15: 51227946-51227946 on Build GRCh38
rs765243938 Chr.15: 51227881-51227881 on Build GRCh38
rs537307360 Chr.15: 51228048-51228048 on Build GRCh38
rs550859249 Chr.15: 51228164-51228165 on Build GRCh38
rs1024617781 Chr.15: 51228149-51228149 on Build GRCh38
rs985681773 Chr.15: 51228218-51228218 on Build GRCh38
rs369622599 Chr.15: 51228226-51228226 on Build GRCh38
rs780888050 Chr.15: 51227812-51227812 on Build GRCh38
rs1803154 Chr.15: 51227908-51227908 on Build GRCh38
rs944252652 Chr.15: 51228051-51228051 on Build GRCh38
rs753195937 Chr.15: 51227941-51227941 on Build GRCh38
rs1041053163 Chr.15: 51228054-51228054 on Build GRCh38
rs79887439 Chr.15: 51228021-51228021 on Build GRCh38
rs184470765 Chr.15: 51228268-51228268 on Build GRCh38
rs916593491 Chr.15: 51227912-51227912 on Build GRCh38
rs950692139 Chr.15: 51228174-51228174 on Build GRCh38
rs1006451038 Chr.15: 51227810-51227810 on Build GRCh38
rs1033623088 Chr.15: 51227839-51227839 on Build GRCh38
rs146277709 Chr.15: 51228176-51228176 on Build GRCh38
rs555191361 Chr.15: 51228066-51228067 on Build GRCh38
rs755257022 Chr.15: 51227925-51227925 on Build GRCh38
rs751920045 Chr.15: 51227902-51227902 on Build GRCh38
rs761906497 Chr.15: 51227866-51227866 on Build GRCh38
rs547448505 Chr.15: 51227959-51227959 on Build GRCh38
rs140027501 Chr.15: 51228096-51228096 on Build GRCh38
rs770719138 Chr.15: 51227816-51227825 on Build GRCh38
rs368588028 Chr.15: 51227931-51227931 on Build GRCh38
rs542617167 Chr.15: 51228168-51228168 on Build GRCh38
rs745845217 Chr.15: 51227854-51227854 on Build GRCh38
rs1057518574 Chr.15: 51227850-51227850 on Build GRCh38
rs977715644 Chr.15: 51228152-51228152 on Build GRCh38
rs144210769 Chr.15: 51228061-51228061 on Build GRCh38
rs777577929 Chr.15: 51227821-51227821 on Build GRCh38
rs748955665 Chr.15: 51227971-51227971 on Build GRCh38
rs773312519 Chr.15: 51227882-51227882 on Build GRCh38
rs1008605680 Chr.15: 51228087-51228087 on Build GRCh38
rs936870869 Chr.15: 51227998-51227998 on Build GRCh38
rs958026076 Chr.15: 51228187-51228187 on Build GRCh38
rs28757183 Chr.15: 51227906-51227906 on Build GRCh38
rs891273529 Chr.15: 51228074-51228074 on Build GRCh38
rs371077463 Chr.15: 51227835-51227835 on Build GRCh38
rs147288710 Chr.15: 51228165-51228166 on Build GRCh38
rs6413418 Chr.15: 51228112-51228112 on Build GRCh38
rs755030218 Chr.15: 51228129-51228129 on Build GRCh38
rs775367580 Chr.15: 51227857-51227857 on Build GRCh38
rs576343700 Chr.15: 51228203-51228203 on Build GRCh38
rs768157788 Chr.15: 51227934-51227934 on Build GRCh38
rs990878332 Chr.15: 51227909-51227909 on Build GRCh38
rs749555631 Chr.15: 51228166-51228167 on Build GRCh38
rs558502162 Chr.15: 51228060-51228060 on Build GRCh38
rs1048537266 Chr.15: 51228106-51228106 on Build GRCh38
rs1034879076 Chr.15: 51228206-51228206 on Build GRCh38
rs1006092077 Chr.15: 51228113-51228113 on Build GRCh38
rs139905959 Chr.15: 51227894-51227894 on Build GRCh38
rs537019588 Chr.15: 51227950-51227950 on Build GRCh38
rs946733381 Chr.15: 51227988-51227988 on Build GRCh38
rs372059801 Chr.15: 51228008-51228008 on Build GRCh38
rs548160872 Chr.15: 51228018-51228018 on Build GRCh38

View Full Product Details