Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51397236-51397752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLDN
Entrez Gene ID
342035
Gene Name
gliomedin
Gene Aliases
CLOM, COLM, CRG-L2, CRGL2, UNC-112
UniGene
Hs.526441
Gene Chromosome Location
Chr.15: 51341516-51413365 on Build GRCh38
Transcripts
NM_181789.2 XM_011521501.2 XM_017022121.1 XM_017022122.1 XM_017022123.1 XM_017022124.1 XM_017022125.1 XM_017022126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5383479 c.754C>T p.P252S Chr.15: 51397535-51397535 on Build GRCh38
701069 c.806G>T p.G269V Chr.15: 51397587-51397587 on Build GRCh38
1678456 c.752G>T p.G251V Chr.15: 51397533-51397533 on Build GRCh38
4055501 c.780C>T p.G260G Chr.15: 51397561-51397561 on Build GRCh38
3728077 c.689-2A>G p.? Chr.15: 51397468-51397468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370328960 Chr.15: 51397618-51397618 on Build GRCh38
rs201795225 Chr.15: 51397586-51397586 on Build GRCh38
rs749671919 Chr.15: 51397488-51397488 on Build GRCh38
rs373750156 Chr.15: 51397552-51397552 on Build GRCh38
rs545825070 Chr.15: 51397262-51397262 on Build GRCh38
rs373661236 Chr.15: 51397595-51397595 on Build GRCh38
rs200306486 Chr.15: 51397620-51397620 on Build GRCh38
rs533453553 Chr.15: 51397450-51397450 on Build GRCh38
rs575390245 Chr.15: 51397461-51397461 on Build GRCh38
rs898339184 Chr.15: 51397415-51397415 on Build GRCh38
rs746201596 Chr.15: 51397561-51397561 on Build GRCh38
rs767004221 Chr.15: 51397420-51397420 on Build GRCh38
rs868024551 Chr.15: 51397354-51397354 on Build GRCh38
rs760002795 Chr.15: 51397603-51397603 on Build GRCh38
rs898455405 Chr.15: 51397696-51397696 on Build GRCh38
rs759149598 Chr.15: 51397422-51397422 on Build GRCh38
rs201901285 Chr.15: 51397519-51397519 on Build GRCh38
rs758677429 Chr.15: 51397472-51397472 on Build GRCh38
rs778826448 Chr.15: 51397462-51397462 on Build GRCh38
rs570468297 Chr.15: 51397463-51397463 on Build GRCh38
rs564372081 Chr.15: 51397617-51397617 on Build GRCh38
rs763323925 Chr.15: 51397483-51397507 on Build GRCh38
rs772414925 Chr.15: 51397633-51397633 on Build GRCh38
rs771413809 Chr.15: 51397491-51397491 on Build GRCh38
rs774891841 Chr.15: 51397496-51397496 on Build GRCh38
rs773563537 Chr.15: 51397451-51397454 on Build GRCh38
rs774927202 Chr.15: 51397421-51397421 on Build GRCh38
rs530197987 Chr.15: 51397394-51397394 on Build GRCh38
rs893466056 Chr.15: 51397304-51397306 on Build GRCh38
rs553016727 Chr.15: 51397495-51397495 on Build GRCh38
rs752745071 Chr.15: 51397523-51397523 on Build GRCh38
rs886041056 Chr.15: 51397539-51397539 on Build GRCh38
rs750413223 Chr.15: 51397512-51397512 on Build GRCh38
rs780003472 Chr.15: 51397518-51397518 on Build GRCh38
rs548746004 Chr.15: 51397407-51397407 on Build GRCh38
rs914762274 Chr.15: 51397298-51397298 on Build GRCh38
rs183416884 Chr.15: 51397448-51397448 on Build GRCh38
rs373267973 Chr.15: 51397571-51397571 on Build GRCh38
rs111907476 Chr.15: 51397346-51397347 on Build GRCh38
rs926458632 Chr.15: 51397532-51397532 on Build GRCh38
rs200060926 Chr.15: 51397349-51397350 on Build GRCh38
rs1036899030 Chr.15: 51397674-51397674 on Build GRCh38
rs765004774 Chr.15: 51397511-51397511 on Build GRCh38
rs1039145716 Chr.15: 51397331-51397331 on Build GRCh38
rs1009270442 Chr.15: 51397333-51397333 on Build GRCh38
rs375789151 Chr.15: 51397565-51397565 on Build GRCh38
rs149543159 Chr.15: 51397468-51397468 on Build GRCh38
rs528101800 Chr.15: 51397290-51397290 on Build GRCh38
rs369454519 Chr.15: 51397508-51397508 on Build GRCh38
rs568180379 Chr.15: 51397392-51397392 on Build GRCh38
rs752443022 Chr.15: 51397438-51397438 on Build GRCh38
rs891015676 Chr.15: 51397361-51397361 on Build GRCh38
rs931128347 Chr.15: 51397719-51397719 on Build GRCh38
rs74013203 Chr.15: 51397346-51397346 on Build GRCh38
rs1052226575 Chr.15: 51397355-51397355 on Build GRCh38
rs143383924 Chr.15: 51397566-51397566 on Build GRCh38
rs768012999 Chr.15: 51397617-51397617 on Build GRCh38
rs17648128 Chr.15: 51397575-51397575 on Build GRCh38
rs753424904 Chr.15: 51397510-51397510 on Build GRCh38
rs750218696 Chr.15: 51397592-51397592 on Build GRCh38
rs774896034 Chr.15: 51397508-51397508 on Build GRCh38
rs989065249 Chr.15: 51397538-51397538 on Build GRCh38
rs375769696 Chr.15: 51397614-51397614 on Build GRCh38
rs1147142 Chr.15: 51397427-51397427 on Build GRCh38
rs749505021 Chr.15: 51397543-51397543 on Build GRCh38
rs776091046 Chr.15: 51397641-51397641 on Build GRCh38
rs766229043 Chr.15: 51397606-51397606 on Build GRCh38
rs763750572 Chr.15: 51397507-51397507 on Build GRCh38
rs187857819 Chr.15: 51397585-51397585 on Build GRCh38
rs775395450 Chr.15: 51397450-51397451 on Build GRCh38
rs1002544475 Chr.15: 51397258-51397258 on Build GRCh38
rs907869578 Chr.15: 51397664-51397664 on Build GRCh38
rs748196701 Chr.15: 51397451-51397452 on Build GRCh38
rs956520639 Chr.15: 51397282-51397282 on Build GRCh38
rs751508597 Chr.15: 51397613-51397613 on Build GRCh38
rs763760292 Chr.15: 51397454-51397454 on Build GRCh38
rs753657467 Chr.15: 51397456-51397456 on Build GRCh38
rs756148812 Chr.15: 51397524-51397524 on Build GRCh38
rs1004103923 Chr.15: 51397391-51397391 on Build GRCh38
rs375203691 Chr.15: 51397493-51397493 on Build GRCh38
rs779226495 Chr.15: 51397627-51397627 on Build GRCh38
rs762763177 Chr.15: 51397603-51397603 on Build GRCh38
rs564062001 Chr.15: 51397283-51397283 on Build GRCh38
rs765060260 Chr.15: 51397587-51397587 on Build GRCh38
rs540246264 Chr.15: 51397663-51397663 on Build GRCh38
rs558835975 Chr.15: 51397460-51397460 on Build GRCh38
rs965148006 Chr.15: 51397381-51397383 on Build GRCh38
rs139856594 Chr.15: 51397303-51397303 on Build GRCh38
rs755147576 Chr.15: 51397466-51397466 on Build GRCh38
rs939904744 Chr.15: 51397665-51397665 on Build GRCh38
rs768206046 Chr.15: 51397638-51397638 on Build GRCh38
rs574578328 Chr.15: 51397535-51397535 on Build GRCh38
rs373065689 Chr.15: 51397636-51397636 on Build GRCh38
rs779389882 Chr.15: 51397560-51397560 on Build GRCh38
rs758241376 Chr.15: 51397517-51397517 on Build GRCh38
rs760182082 Chr.15: 51397497-51397497 on Build GRCh38
rs766826604 Chr.15: 51397498-51397499 on Build GRCh38

View Full Product Details