Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57960460-57961006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A2
Entrez Gene ID
8854
Gene Name
aldehyde dehydrogenase 1 family member A2
Gene Aliases
RALDH(II), RALDH2, RALDH2-T
UniGene
Hs.643455
Gene Chromosome Location
Chr.15: 57953424-58065923 on Build GRCh38
Transcripts
NM_001206897.1 NM_003888.3 NM_170696.2 NM_170697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5577247 c.1469G>A p.G490E Chr.15: 57960785-57960785 on Build GRCh38
401559 c.1480G>A p.E494K Chr.15: 57960774-57960774 on Build GRCh38
963314 c.1427C>T p.A476V Chr.15: 57960827-57960827 on Build GRCh38
3401846 c.1435G>A p.A479T Chr.15: 57960819-57960819 on Build GRCh38
3502731 c.1474G>A p.G492R Chr.15: 57960780-57960780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749322232 Chr.15: 57960756-57960756 on Build GRCh38
rs766067276 Chr.15: 57960757-57960758 on Build GRCh38
rs146966659 Chr.15: 57960892-57960892 on Build GRCh38
rs916962753 Chr.15: 57960692-57960692 on Build GRCh38
rs1040181939 Chr.15: 57960811-57960811 on Build GRCh38
rs372054788 Chr.15: 57960885-57960885 on Build GRCh38
rs770711096 Chr.15: 57960902-57960902 on Build GRCh38
rs34682734 Chr.15: 57960536-57960536 on Build GRCh38
rs759066132 Chr.15: 57960802-57960802 on Build GRCh38
rs547810916 Chr.15: 57960626-57960626 on Build GRCh38
rs747112431 Chr.15: 57960879-57960879 on Build GRCh38
rs781550198 Chr.15: 57960882-57960882 on Build GRCh38
rs111252521 Chr.15: 57960645-57960645 on Build GRCh38
rs747078949 Chr.15: 57960732-57960732 on Build GRCh38
rs371309158 Chr.15: 57960851-57960851 on Build GRCh38
rs979405579 Chr.15: 57960610-57960610 on Build GRCh38
rs3784262 Chr.15: 57960908-57960908 on Build GRCh38
rs779747774 Chr.15: 57960697-57960697 on Build GRCh38
rs574492465 Chr.15: 57960603-57960603 on Build GRCh38
rs901709908 Chr.15: 57960828-57960828 on Build GRCh38
rs1048937758 Chr.15: 57960748-57960748 on Build GRCh38
rs4646634 Chr.15: 57960653-57960653 on Build GRCh38
rs767414203 Chr.15: 57960832-57960832 on Build GRCh38
rs750641775 Chr.15: 57960484-57960484 on Build GRCh38
rs778485212 Chr.15: 57960873-57960875 on Build GRCh38
rs868060815 Chr.15: 57960780-57960780 on Build GRCh38
rs773674202 Chr.15: 57960890-57960890 on Build GRCh38
rs551528782 Chr.15: 57960712-57960712 on Build GRCh38
rs865968982 Chr.15: 57960779-57960779 on Build GRCh38
rs971614692 Chr.15: 57960922-57960922 on Build GRCh38
rs988985015 Chr.15: 57960665-57960665 on Build GRCh38
rs189376070 Chr.15: 57960760-57960760 on Build GRCh38
rs138003657 Chr.15: 57960905-57960905 on Build GRCh38
rs562970741 Chr.15: 57960654-57960654 on Build GRCh38
rs369082445 Chr.15: 57960875-57960875 on Build GRCh38
rs774041681 Chr.15: 57960894-57960894 on Build GRCh38
rs756878707 Chr.15: 57960860-57960861 on Build GRCh38
rs879357298 Chr.15: 57960876-57960876 on Build GRCh38
rs199654940 Chr.15: 57960775-57960775 on Build GRCh38
rs752676181 Chr.15: 57960839-57960839 on Build GRCh38
rs141758881 Chr.15: 57960909-57960909 on Build GRCh38
rs901269150 Chr.15: 57960514-57960514 on Build GRCh38
rs774047547 Chr.15: 57960778-57960778 on Build GRCh38
rs763827962 Chr.15: 57960861-57960861 on Build GRCh38
rs200758020 Chr.15: 57960754-57960754 on Build GRCh38
rs1028733097 Chr.15: 57960569-57960569 on Build GRCh38
rs773017152 Chr.15: 57960757-57960757 on Build GRCh38
rs202059375 Chr.15: 57960880-57960880 on Build GRCh38
rs930347681 Chr.15: 57960735-57960735 on Build GRCh38
rs571598114 Chr.15: 57960535-57960535 on Build GRCh38
rs762713198 Chr.15: 57960643-57960643 on Build GRCh38
rs3784263 Chr.15: 57960864-57960864 on Build GRCh38
rs35789027 Chr.15: 57960706-57960706 on Build GRCh38
rs560892063 Chr.15: 57960574-57960574 on Build GRCh38
rs769090661 Chr.15: 57960745-57960745 on Build GRCh38
rs1033278085 Chr.15: 57960637-57960637 on Build GRCh38

View Full Product Details