Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57963807-57964321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A2
Entrez Gene ID
8854
Gene Name
aldehyde dehydrogenase 1 family member A2
Gene Aliases
RALDH(II), RALDH2, RALDH2-T
UniGene
Hs.643455
Gene Chromosome Location
Chr.15: 57953424-58065923 on Build GRCh38
Transcripts
NM_001206897.1 NM_003888.3 NM_170696.2 NM_170697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598189 c.1007T>G p.F336C Chr.15: 57963964-57963964 on Build GRCh38
3794266 c.1020C>T p.S340S Chr.15: 57963951-57963951 on Build GRCh38
5936111 c.1054C>T p.P352S Chr.15: 57963917-57963917 on Build GRCh38
3936796 c.957C>G p.C319W Chr.15: 57964014-57964014 on Build GRCh38
6598193 c.1049G>A p.G350E Chr.15: 57963922-57963922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568894798 Chr.15: 57964131-57964131 on Build GRCh38
rs995196417 Chr.15: 57964148-57964148 on Build GRCh38
rs963837318 Chr.15: 57963998-57963998 on Build GRCh38
rs923941363 Chr.15: 57964071-57964071 on Build GRCh38
rs774419607 Chr.15: 57964011-57964011 on Build GRCh38
rs763069969 Chr.15: 57963891-57963891 on Build GRCh38
rs1023303759 Chr.15: 57963930-57963930 on Build GRCh38
rs763772319 Chr.15: 57963924-57963924 on Build GRCh38
rs377317390 Chr.15: 57963864-57963864 on Build GRCh38
rs770934217 Chr.15: 57963963-57963963 on Build GRCh38
rs777101147 Chr.15: 57963845-57963845 on Build GRCh38
rs778997757 Chr.15: 57963932-57963932 on Build GRCh38
rs778924533 Chr.15: 57964117-57964117 on Build GRCh38
rs753732065 Chr.15: 57964216-57964216 on Build GRCh38
rs751372190 Chr.15: 57964091-57964091 on Build GRCh38
rs141245344 Chr.15: 57963931-57963931 on Build GRCh38
rs752416528 Chr.15: 57963909-57963909 on Build GRCh38
rs946771251 Chr.15: 57964058-57964058 on Build GRCh38
rs767813198 Chr.15: 57963908-57963908 on Build GRCh38
rs898695294 Chr.15: 57964135-57964135 on Build GRCh38
rs946659347 Chr.15: 57963860-57963860 on Build GRCh38
rs781574976 Chr.15: 57963873-57963873 on Build GRCh38
rs750327790 Chr.15: 57963935-57963935 on Build GRCh38
rs775570769 Chr.15: 57963893-57963893 on Build GRCh38
rs137957671 Chr.15: 57963944-57963944 on Build GRCh38
rs1025294962 Chr.15: 57963945-57963945 on Build GRCh38
rs367804744 Chr.15: 57963960-57963960 on Build GRCh38
rs548803596 Chr.15: 57964213-57964222 on Build GRCh38
rs956695453 Chr.15: 57964275-57964275 on Build GRCh38
rs761085706 Chr.15: 57964010-57964010 on Build GRCh38
rs770042436 Chr.15: 57963875-57963875 on Build GRCh38
rs896626113 Chr.15: 57963926-57963926 on Build GRCh38
rs34666807 Chr.15: 57963872-57963872 on Build GRCh38
rs935344662 Chr.15: 57964083-57964083 on Build GRCh38
rs769945737 Chr.15: 57963874-57963874 on Build GRCh38
rs774853794 Chr.15: 57963990-57963990 on Build GRCh38
rs1047268885 Chr.15: 57964156-57964156 on Build GRCh38
rs537977490 Chr.15: 57964132-57964132 on Build GRCh38
rs750413727 Chr.15: 57964087-57964087 on Build GRCh38
rs779670359 Chr.15: 57963943-57963943 on Build GRCh38
rs758287557 Chr.15: 57963942-57963942 on Build GRCh38
rs769962890 Chr.15: 57963950-57963950 on Build GRCh38
rs4646626 Chr.15: 57963929-57963929 on Build GRCh38
rs752281062 Chr.15: 57964274-57964274 on Build GRCh38
rs765053493 Chr.15: 57964271-57964271 on Build GRCh38
rs35214579 Chr.15: 57964197-57964197 on Build GRCh38
rs763149416 Chr.15: 57963904-57963904 on Build GRCh38
rs745723972 Chr.15: 57963870-57963870 on Build GRCh38
rs996795655 Chr.15: 57964205-57964205 on Build GRCh38
rs775758977 Chr.15: 57964005-57964005 on Build GRCh38
rs772458487 Chr.15: 57964003-57964003 on Build GRCh38
rs979513243 Chr.15: 57964250-57964250 on Build GRCh38
rs755663422 Chr.15: 57963840-57963840 on Build GRCh38
rs917960934 Chr.15: 57964284-57964284 on Build GRCh38
rs114347273 Chr.15: 57963951-57963951 on Build GRCh38
rs773201837 Chr.15: 57963890-57963890 on Build GRCh38
rs573894154 Chr.15: 57964283-57964283 on Build GRCh38
rs757308536 Chr.15: 57963858-57963858 on Build GRCh38
rs761420400 Chr.15: 57964049-57964049 on Build GRCh38
rs372024991 Chr.15: 57964088-57964088 on Build GRCh38
rs759901664 Chr.15: 57963903-57963903 on Build GRCh38
rs145953941 Chr.15: 57964002-57964002 on Build GRCh38
rs138118724 Chr.15: 57964291-57964291 on Build GRCh38
rs375883599 Chr.15: 57963835-57963835 on Build GRCh38
rs751058082 Chr.15: 57964203-57964203 on Build GRCh38
rs139464985 Chr.15: 57964063-57964063 on Build GRCh38
rs4646625 Chr.15: 57964098-57964098 on Build GRCh38
rs577679957 Chr.15: 57964196-57964196 on Build GRCh38
rs371746910 Chr.15: 57964106-57964106 on Build GRCh38
rs757351162 Chr.15: 57964110-57964110 on Build GRCh38
rs1019488419 Chr.15: 57964189-57964189 on Build GRCh38
rs986710758 Chr.15: 57963952-57963952 on Build GRCh38
rs902543056 Chr.15: 57963881-57963881 on Build GRCh38
rs553929285 Chr.15: 57964227-57964227 on Build GRCh38
rs375413897 Chr.15: 57963921-57963921 on Build GRCh38
rs1019962559 Chr.15: 57963978-57963978 on Build GRCh38
rs753463120 Chr.15: 57964027-57964027 on Build GRCh38
rs754792712 Chr.15: 57964240-57964240 on Build GRCh38
rs754801022 Chr.15: 57964097-57964097 on Build GRCh38
rs1008315420 Chr.15: 57964188-57964188 on Build GRCh38
rs774839436 Chr.15: 57963900-57963900 on Build GRCh38
rs767984071 Chr.15: 57963872-57963874 on Build GRCh38
rs34474303 Chr.15: 57964167-57964167 on Build GRCh38
rs184664871 Chr.15: 57963857-57963857 on Build GRCh38
rs766315896 Chr.15: 57964089-57964089 on Build GRCh38

View Full Product Details