Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57994940-57995436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A2
Entrez Gene ID
8854
Gene Name
aldehyde dehydrogenase 1 family member A2
Gene Aliases
RALDH(II), RALDH2, RALDH2-T
UniGene
Hs.643455
Gene Chromosome Location
Chr.15: 57953424-58065923 on Build GRCh38
Transcripts
NM_001206897.1 NM_003888.3 NM_170696.2 NM_170697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4499748 c.549C>T p.I183I Chr.15: 57995084-57995084 on Build GRCh38
4055885 c.547A>T p.I183F Chr.15: 57995086-57995086 on Build GRCh38
132656 c.535G>C p.V179L Chr.15: 57995098-57995098 on Build GRCh38
3420476 c.518G>T p.R173I Chr.15: 57995115-57995115 on Build GRCh38
4842536 c.549C>A p.I183I Chr.15: 57995084-57995084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575040621 Chr.15: 57995134-57995134 on Build GRCh38
rs770784333 Chr.15: 57995099-57995099 on Build GRCh38
rs773006216 Chr.15: 57995367-57995367 on Build GRCh38
rs964988102 Chr.15: 57995274-57995274 on Build GRCh38
rs145691810 Chr.15: 57995194-57995194 on Build GRCh38
rs78794515 Chr.15: 57995275-57995275 on Build GRCh38
rs548247204 Chr.15: 57995232-57995232 on Build GRCh38
rs575127213 Chr.15: 57995394-57995394 on Build GRCh38
rs1022450046 Chr.15: 57995316-57995316 on Build GRCh38
rs796534531 Chr.15: 57995236-57995237 on Build GRCh38
rs528578248 Chr.15: 57995230-57995230 on Build GRCh38
rs756352718 Chr.15: 57995054-57995054 on Build GRCh38
rs372661426 Chr.15: 57995030-57995030 on Build GRCh38
rs34068380 Chr.15: 57995217-57995218 on Build GRCh38
rs778397345 Chr.15: 57995183-57995184 on Build GRCh38
rs765752318 Chr.15: 57995037-57995037 on Build GRCh38
rs746906780 Chr.15: 57995353-57995353 on Build GRCh38
rs201538569 Chr.15: 57995162-57995162 on Build GRCh38
rs140216186 Chr.15: 57995062-57995062 on Build GRCh38
rs188321672 Chr.15: 57995413-57995413 on Build GRCh38
rs752207299 Chr.15: 57995172-57995172 on Build GRCh38
rs567915706 Chr.15: 57995233-57995233 on Build GRCh38
rs756058906 Chr.15: 57995081-57995081 on Build GRCh38
rs762663712 Chr.15: 57995390-57995390 on Build GRCh38
rs755649051 Chr.15: 57995176-57995176 on Build GRCh38
rs761939819 Chr.15: 57995133-57995133 on Build GRCh38
rs377293453 Chr.15: 57995247-57995247 on Build GRCh38
rs777860277 Chr.15: 57995179-57995179 on Build GRCh38
rs539418255 Chr.15: 57994990-57994990 on Build GRCh38
rs777839828 Chr.15: 57995086-57995086 on Build GRCh38
rs762265558 Chr.15: 57995035-57995035 on Build GRCh38
rs3985733 Chr.15: 57995237-57995237 on Build GRCh38
rs570798483 Chr.15: 57995359-57995359 on Build GRCh38
rs550950579 Chr.15: 57995355-57995355 on Build GRCh38
rs753773193 Chr.15: 57995187-57995187 on Build GRCh38
rs539098274 Chr.15: 57995243-57995243 on Build GRCh38
rs927703908 Chr.15: 57995356-57995356 on Build GRCh38
rs780438722 Chr.15: 57995106-57995106 on Build GRCh38
rs754541558 Chr.15: 57995061-57995061 on Build GRCh38
rs202076416 Chr.15: 57995160-57995160 on Build GRCh38
rs766166060 Chr.15: 57995225-57995237 on Build GRCh38
rs776236915 Chr.15: 57995141-57995141 on Build GRCh38
rs745463934 Chr.15: 57995191-57995192 on Build GRCh38
rs186254060 Chr.15: 57994966-57994966 on Build GRCh38
rs376037780 Chr.15: 57995055-57995055 on Build GRCh38
rs148475880 Chr.15: 57995101-57995101 on Build GRCh38
rs1055011612 Chr.15: 57995010-57995010 on Build GRCh38
rs61999890 Chr.15: 57995292-57995292 on Build GRCh38
rs543992207 Chr.15: 57995396-57995396 on Build GRCh38
rs756713979 Chr.15: 57995075-57995076 on Build GRCh38
rs375746194 Chr.15: 57995237-57995237 on Build GRCh38
rs776622530 Chr.15: 57995129-57995129 on Build GRCh38
rs572962104 Chr.15: 57995001-57995001 on Build GRCh38
rs3922093 Chr.15: 57995029-57995029 on Build GRCh38
rs559453634 Chr.15: 57995195-57995195 on Build GRCh38
rs183256839 Chr.15: 57995323-57995323 on Build GRCh38
rs114333284 Chr.15: 57995168-57995168 on Build GRCh38
rs890415711 Chr.15: 57995236-57995236 on Build GRCh38
rs201972956 Chr.15: 57995226-57995237 on Build GRCh38
rs766678904 Chr.15: 57995156-57995156 on Build GRCh38
rs763296066 Chr.15: 57995042-57995042 on Build GRCh38
rs747192456 Chr.15: 57995111-57995111 on Build GRCh38
rs4646606 Chr.15: 57995328-57995328 on Build GRCh38
rs61999889 Chr.15: 57995241-57995241 on Build GRCh38
rs752224198 Chr.15: 57995064-57995064 on Build GRCh38
rs879071220 Chr.15: 57995217-57995217 on Build GRCh38
rs71441491 Chr.15: 57995218-57995219 on Build GRCh38
rs768890166 Chr.15: 57995122-57995122 on Build GRCh38
rs750841249 Chr.15: 57995038-57995038 on Build GRCh38
rs763318081 Chr.15: 57995154-57995154 on Build GRCh38
rs946028466 Chr.15: 57995211-57995211 on Build GRCh38
rs773638758 Chr.15: 57995146-57995146 on Build GRCh38
rs749163730 Chr.15: 57995096-57995096 on Build GRCh38
rs745544251 Chr.15: 57995411-57995411 on Build GRCh38
rs757007375 Chr.15: 57995365-57995365 on Build GRCh38
rs550639196 Chr.15: 57995240-57995240 on Build GRCh38
rs74017097 Chr.15: 57994996-57994996 on Build GRCh38
rs1018055984 Chr.15: 57995270-57995270 on Build GRCh38

View Full Product Details