Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59438009-59438506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM81A
Entrez Gene ID
145773
Gene Name
family with sequence similarity 81 member A
Gene Aliases
-
UniGene
Hs.531168
Gene Chromosome Location
Chr.15: 59399662-59523555 on Build GRCh38
Transcripts
NM_152450.2 XM_005254166.2 XM_006720398.3 XM_006720399.2 XM_011521247.2 XM_011521248.2 XM_011521249.2 XM_011521250.2 XM_017021932.1

SNP Detail

SNP ID Chromosome Location
rs998166936 Chr.15: 59438483-59438483 on Build GRCh38
rs12907533 Chr.15: 59438246-59438246 on Build GRCh38
rs760816978 Chr.15: 59438469-59438469 on Build GRCh38
rs749833932 Chr.15: 59438320-59438320 on Build GRCh38
rs929883086 Chr.15: 59438431-59438431 on Build GRCh38
rs1047441784 Chr.15: 59438440-59438440 on Build GRCh38
rs112849510 Chr.15: 59438086-59438086 on Build GRCh38
rs906448289 Chr.15: 59438417-59438417 on Build GRCh38
rs767535520 Chr.15: 59438355-59438355 on Build GRCh38
rs570983650 Chr.15: 59438064-59438064 on Build GRCh38
rs564784619 Chr.15: 59438458-59438458 on Build GRCh38
rs111732908 Chr.15: 59438233-59438233 on Build GRCh38
rs538324272 Chr.15: 59438214-59438214 on Build GRCh38
rs899954818 Chr.15: 59438482-59438482 on Build GRCh38
rs535636570 Chr.15: 59438316-59438316 on Build GRCh38
rs148415789 Chr.15: 59438307-59438307 on Build GRCh38
rs373717618 Chr.15: 59438304-59438304 on Build GRCh38
rs1005808063 Chr.15: 59438461-59438461 on Build GRCh38
rs1018361106 Chr.15: 59438468-59438468 on Build GRCh38
rs556658830 Chr.15: 59438276-59438276 on Build GRCh38

View Full Product Details