Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 62164361-62164860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD4B
Entrez Gene ID
388125
Gene Name
C2 calcium dependent domain containing 4B
Gene Aliases
FAM148B, NLF2
UniGene
Hs.144372
Gene Chromosome Location
Chr.15: 62163535-62165732 on Build GRCh38
Transcripts
NM_001007595.2 XM_017022170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
229802 c.478C>T p.P160S Chr.15: 62164507-62164507 on Build GRCh38
4799669 c.178G>T p.A60S Chr.15: 62164807-62164807 on Build GRCh38
6413382 c.175G>T p.A59S Chr.15: 62164810-62164810 on Build GRCh38
5729766 c.528C>T p.L176L Chr.15: 62164457-62164457 on Build GRCh38
4804165 c.125C>A p.P42H Chr.15: 62164860-62164860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771006417 Chr.15: 62164412-62164412 on Build GRCh38
rs761125408 Chr.15: 62164818-62164827 on Build GRCh38
rs1040198450 Chr.15: 62164607-62164607 on Build GRCh38
rs766095864 Chr.15: 62164758-62164758 on Build GRCh38
rs958847256 Chr.15: 62164481-62164481 on Build GRCh38
rs753854527 Chr.15: 62164618-62164618 on Build GRCh38
rs749906175 Chr.15: 62164632-62164632 on Build GRCh38
rs998281067 Chr.15: 62164617-62164617 on Build GRCh38
rs1018296862 Chr.15: 62164456-62164456 on Build GRCh38
rs1021438100 Chr.15: 62164785-62164785 on Build GRCh38
rs561369974 Chr.15: 62164417-62164417 on Build GRCh38
rs528963694 Chr.15: 62164441-62164441 on Build GRCh38
rs576329402 Chr.15: 62164390-62164390 on Build GRCh38
rs916979191 Chr.15: 62164500-62164500 on Build GRCh38
rs759750643 Chr.15: 62164439-62164439 on Build GRCh38
rs146941362 Chr.15: 62164783-62164783 on Build GRCh38
rs765154588 Chr.15: 62164740-62164740 on Build GRCh38
rs1017229725 Chr.15: 62164480-62164480 on Build GRCh38
rs8025811 Chr.15: 62164746-62164746 on Build GRCh38
rs1034231185 Chr.15: 62164829-62164829 on Build GRCh38
rs768402307 Chr.15: 62164719-62164719 on Build GRCh38
rs746006182 Chr.15: 62164705-62164705 on Build GRCh38
rs1001446279 Chr.15: 62164819-62164819 on Build GRCh38
rs1005584448 Chr.15: 62164634-62164634 on Build GRCh38
rs756583829 Chr.15: 62164665-62164665 on Build GRCh38
rs568495090 Chr.15: 62164749-62164749 on Build GRCh38
rs968510345 Chr.15: 62164791-62164791 on Build GRCh38
rs889057159 Chr.15: 62164573-62164573 on Build GRCh38
rs552064552 Chr.15: 62164649-62164650 on Build GRCh38
rs935664379 Chr.15: 62164708-62164708 on Build GRCh38
rs372753423 Chr.15: 62164396-62164396 on Build GRCh38
rs991678636 Chr.15: 62164499-62164499 on Build GRCh38
rs1047128328 Chr.15: 62164762-62164762 on Build GRCh38
rs924298061 Chr.15: 62164540-62164541 on Build GRCh38
rs779346787 Chr.15: 62164797-62164797 on Build GRCh38
rs887105259 Chr.15: 62164633-62164633 on Build GRCh38
rs778988906 Chr.15: 62164703-62164703 on Build GRCh38
rs1054288887 Chr.15: 62164556-62164556 on Build GRCh38
rs951674313 Chr.15: 62164477-62164477 on Build GRCh38
rs984385678 Chr.15: 62164479-62164479 on Build GRCh38
rs775939393 Chr.15: 62164707-62164708 on Build GRCh38
rs776302952 Chr.15: 62164734-62164734 on Build GRCh38
rs749756812 Chr.15: 62164697-62164697 on Build GRCh38
rs530840238 Chr.15: 62164741-62164741 on Build GRCh38
rs772289303 Chr.15: 62164440-62164440 on Build GRCh38
rs887214186 Chr.15: 62164763-62164763 on Build GRCh38
rs772180933 Chr.15: 62164809-62164809 on Build GRCh38
rs753085681 Chr.15: 62164661-62164661 on Build GRCh38
rs949842115 Chr.15: 62164517-62164517 on Build GRCh38
rs917030149 Chr.15: 62164386-62164386 on Build GRCh38
rs977452856 Chr.15: 62164526-62164526 on Build GRCh38
rs764744304 Chr.15: 62164628-62164628 on Build GRCh38
rs929163771 Chr.15: 62164757-62164757 on Build GRCh38
rs559489998 Chr.15: 62164471-62164471 on Build GRCh38
rs943300338 Chr.15: 62164730-62164730 on Build GRCh38
rs1000120400 Chr.15: 62164404-62164404 on Build GRCh38
rs972012527 Chr.15: 62164672-62164672 on Build GRCh38
rs551557912 Chr.15: 62164653-62164653 on Build GRCh38
rs778027575 Chr.15: 62164688-62164688 on Build GRCh38
rs775739269 Chr.15: 62164716-62164716 on Build GRCh38
rs540803070 Chr.15: 62164462-62164462 on Build GRCh38
rs373990152 Chr.15: 62164774-62164774 on Build GRCh38
rs377184066 Chr.15: 62164671-62164671 on Build GRCh38
rs569923492 Chr.15: 62164717-62164717 on Build GRCh38
rs943185818 Chr.15: 62164583-62164584 on Build GRCh38
rs1025650029 Chr.15: 62164476-62164476 on Build GRCh38
rs370815414 Chr.15: 62164723-62164723 on Build GRCh38
rs761741672 Chr.15: 62164739-62164739 on Build GRCh38
rs745926724 Chr.15: 62164800-62164800 on Build GRCh38
rs1024482308 Chr.15: 62164670-62164670 on Build GRCh38
rs533026330 Chr.15: 62164621-62164621 on Build GRCh38
rs771357729 Chr.15: 62164698-62164698 on Build GRCh38
rs535925453 Chr.15: 62164769-62164769 on Build GRCh38
rs772268924 Chr.15: 62164713-62164713 on Build GRCh38
rs758118204 Chr.15: 62164649-62164649 on Build GRCh38
rs763999485 Chr.15: 62164460-62164460 on Build GRCh38
rs760521041 Chr.15: 62164450-62164450 on Build GRCh38
rs901217849 Chr.15: 62164612-62164612 on Build GRCh38
rs960194122 Chr.15: 62164836-62164836 on Build GRCh38
rs1034715540 Chr.15: 62164508-62164508 on Build GRCh38
rs935701129 Chr.15: 62164554-62164554 on Build GRCh38
rs757781866 Chr.15: 62164790-62164790 on Build GRCh38
rs901672341 Chr.15: 62164751-62164751 on Build GRCh38
rs903065850 Chr.15: 62164398-62164398 on Build GRCh38
rs991626390 Chr.15: 62164667-62164667 on Build GRCh38
rs894696085 Chr.15: 62164793-62164793 on Build GRCh38

View Full Product Details