Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63912025-63912524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAPK2
Entrez Gene ID
23604
Gene Name
death associated protein kinase 2
Gene Aliases
DRP-1, DRP1
UniGene
Hs.237886
Gene Chromosome Location
Chr.15: 63907036-64046470 on Build GRCh38
Transcripts
NM_014326.3 XM_011521413.2 XM_011521414.1 XM_011521417.1 XM_011521421.2 XM_017022044.1 XM_017022045.1 XM_017022046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5849528 c.859-1G>A p.? Chr.15: 63912198-63912198 on Build GRCh38
4831109 c.933C>A p.V311V Chr.15: 63912123-63912123 on Build GRCh38
6142537 c.872A>T p.Q291L Chr.15: 63912184-63912184 on Build GRCh38
1374012 c.930T>C p.Y310Y Chr.15: 63912126-63912126 on Build GRCh38
434133 c.899T>A p.V300E Chr.15: 63912157-63912157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879576538 Chr.15: 63912491-63912491 on Build GRCh38
rs1026455822 Chr.15: 63912292-63912292 on Build GRCh38
rs772372955 Chr.15: 63912112-63912112 on Build GRCh38
rs756826098 Chr.15: 63912336-63912336 on Build GRCh38
rs778662437 Chr.15: 63912488-63912488 on Build GRCh38
rs372139206 Chr.15: 63912239-63912239 on Build GRCh38
rs78177217 Chr.15: 63912059-63912059 on Build GRCh38
rs759815702 Chr.15: 63912123-63912123 on Build GRCh38
rs776636585 Chr.15: 63912122-63912122 on Build GRCh38
rs557490333 Chr.15: 63912288-63912288 on Build GRCh38
rs540660830 Chr.15: 63912370-63912370 on Build GRCh38
rs561811662 Chr.15: 63912343-63912343 on Build GRCh38
rs929500345 Chr.15: 63912463-63912463 on Build GRCh38
rs554284723 Chr.15: 63912258-63912258 on Build GRCh38
rs149186606 Chr.15: 63912195-63912195 on Build GRCh38
rs975736851 Chr.15: 63912129-63912129 on Build GRCh38
rs146826506 Chr.15: 63912262-63912262 on Build GRCh38
rs952268575 Chr.15: 63912305-63912305 on Build GRCh38
rs763307335 Chr.15: 63912245-63912245 on Build GRCh38
rs367871582 Chr.15: 63912475-63912475 on Build GRCh38
rs761060412 Chr.15: 63912320-63912320 on Build GRCh38
rs188760559 Chr.15: 63912219-63912219 on Build GRCh38
rs370099103 Chr.15: 63912420-63912421 on Build GRCh38
rs750636736 Chr.15: 63912149-63912149 on Build GRCh38
rs754938406 Chr.15: 63912172-63912172 on Build GRCh38
rs770535575 Chr.15: 63912202-63912202 on Build GRCh38
rs758204210 Chr.15: 63912078-63912078 on Build GRCh38
rs894444792 Chr.15: 63912103-63912103 on Build GRCh38
rs552799151 Chr.15: 63912083-63912083 on Build GRCh38
rs750258659 Chr.15: 63912077-63912077 on Build GRCh38
rs747691041 Chr.15: 63912089-63912089 on Build GRCh38
rs769949313 Chr.15: 63912221-63912221 on Build GRCh38
rs747614906 Chr.15: 63912194-63912194 on Build GRCh38
rs898019066 Chr.15: 63912130-63912130 on Build GRCh38
rs543284700 Chr.15: 63912314-63912314 on Build GRCh38
rs761897052 Chr.15: 63912125-63912125 on Build GRCh38
rs907469723 Chr.15: 63912283-63912283 on Build GRCh38
rs764461608 Chr.15: 63912485-63912485 on Build GRCh38
rs367753124 Chr.15: 63912076-63912076 on Build GRCh38
rs761122652 Chr.15: 63912116-63912116 on Build GRCh38
rs765387382 Chr.15: 63912127-63912127 on Build GRCh38
rs374641253 Chr.15: 63912061-63912061 on Build GRCh38
rs144968721 Chr.15: 63912170-63912170 on Build GRCh38
rs748435388 Chr.15: 63912101-63912102 on Build GRCh38
rs532839869 Chr.15: 63912461-63912461 on Build GRCh38
rs922908593 Chr.15: 63912400-63912400 on Build GRCh38
rs1038384449 Chr.15: 63912371-63912371 on Build GRCh38
rs528916354 Chr.15: 63912368-63912368 on Build GRCh38
rs925174778 Chr.15: 63912435-63912435 on Build GRCh38
rs534353075 Chr.15: 63912217-63912217 on Build GRCh38
rs766325954 Chr.15: 63912169-63912169 on Build GRCh38
rs894567854 Chr.15: 63912480-63912480 on Build GRCh38
rs73450762 Chr.15: 63912062-63912062 on Build GRCh38
rs538497620 Chr.15: 63912186-63912186 on Build GRCh38
rs370370764 Chr.15: 63912085-63912085 on Build GRCh38
rs1021529335 Chr.15: 63912265-63912265 on Build GRCh38
rs140986181 Chr.15: 63912403-63912403 on Build GRCh38
rs936558233 Chr.15: 63912438-63912438 on Build GRCh38
rs1007947012 Chr.15: 63912204-63912204 on Build GRCh38
rs780608846 Chr.15: 63912088-63912088 on Build GRCh38
rs150322169 Chr.15: 63912115-63912115 on Build GRCh38
rs199659770 Chr.15: 63912211-63912211 on Build GRCh38
rs113878140 Chr.15: 63912075-63912075 on Build GRCh38
rs978031794 Chr.15: 63912410-63912410 on Build GRCh38
rs775896992 Chr.15: 63912210-63912210 on Build GRCh38
rs368792810 Chr.15: 63912196-63912196 on Build GRCh38
rs867137886 Chr.15: 63912281-63912281 on Build GRCh38
rs775872514 Chr.15: 63912319-63912319 on Build GRCh38
rs370209874 Chr.15: 63912164-63912164 on Build GRCh38
rs753921574 Chr.15: 63912064-63912064 on Build GRCh38
rs967638434 Chr.15: 63912280-63912280 on Build GRCh38
rs575948137 Chr.15: 63912291-63912291 on Build GRCh38
rs759834243 Chr.15: 63912058-63912058 on Build GRCh38
rs764655277 Chr.15: 63912121-63912121 on Build GRCh38
rs769342474 Chr.15: 63912093-63912093 on Build GRCh38
rs971614261 Chr.15: 63912069-63912069 on Build GRCh38
rs185243000 Chr.15: 63912086-63912086 on Build GRCh38
rs4569204 Chr.15: 63912073-63912073 on Build GRCh38
rs765091422 Chr.15: 63912270-63912270 on Build GRCh38
rs945236342 Chr.15: 63912165-63912165 on Build GRCh38
rs781124569 Chr.15: 63912189-63912189 on Build GRCh38
rs867622129 Chr.15: 63912494-63912494 on Build GRCh38
rs1025438780 Chr.15: 63912192-63912192 on Build GRCh38
rs945287540 Chr.15: 63912405-63912405 on Build GRCh38
rs772407366 Chr.15: 63912185-63912185 on Build GRCh38
rs377753351 Chr.15: 63912097-63912097 on Build GRCh38
rs572444831 Chr.15: 63912261-63912261 on Build GRCh38
rs773404306 Chr.15: 63912233-63912233 on Build GRCh38

View Full Product Details