Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65579041-65579547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA9
Entrez Gene ID
81556
Gene Name
von Willebrand factor A domain containing 9
Gene Aliases
C15orf44
UniGene
Hs.6686
Gene Chromosome Location
Chr.15: 65578757-65611289 on Build GRCh38
Transcripts
NM_001136043.2 NM_001207058.1 NM_001207059.1 NR_045104.1 NR_045105.2 NR_045106.2 NR_045901.1 XM_006720708.3 XM_006720709.3 XM_006720711.2 XM_011522101.2 XM_017022644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4813195 c.1605T>C p.Y535Y Chr.15: 65579468-65579468 on Build GRCh38
2256635 c.1581C>T p.T527T Chr.15: 65579492-65579492 on Build GRCh38
1582292 c.1496A>G p.Y499C Chr.15: 65579469-65579469 on Build GRCh38
3377629 c.1632G>A p.T544T Chr.15: 65579441-65579441 on Build GRCh38
416861 c.1483G>C p.E495Q Chr.15: 65579482-65579482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905974608 Chr.15: 65579273-65579273 on Build GRCh38
rs770640770 Chr.15: 65579491-65579491 on Build GRCh38
rs762413272 Chr.15: 65579507-65579507 on Build GRCh38
rs977631524 Chr.15: 65579456-65579456 on Build GRCh38
rs913192514 Chr.15: 65579220-65579220 on Build GRCh38
rs543017461 Chr.15: 65579270-65579270 on Build GRCh38
rs748957170 Chr.15: 65579400-65579400 on Build GRCh38
rs914550148 Chr.15: 65579442-65579442 on Build GRCh38
rs781037827 Chr.15: 65579370-65579370 on Build GRCh38
rs879173903 Chr.15: 65579314-65579314 on Build GRCh38
rs3743169 Chr.15: 65579101-65579101 on Build GRCh38
rs762756375 Chr.15: 65579137-65579137 on Build GRCh38
rs752538670 Chr.15: 65579381-65579382 on Build GRCh38
rs777138857 Chr.15: 65579505-65579505 on Build GRCh38
rs971167757 Chr.15: 65579199-65579199 on Build GRCh38
rs747940838 Chr.15: 65579378-65579378 on Build GRCh38
rs748902130 Chr.15: 65579483-65579483 on Build GRCh38
rs771549509 Chr.15: 65579503-65579503 on Build GRCh38
rs950063104 Chr.15: 65579254-65579254 on Build GRCh38
rs1048872262 Chr.15: 65579353-65579353 on Build GRCh38
rs758722812 Chr.15: 65579453-65579453 on Build GRCh38
rs567080619 Chr.15: 65579163-65579163 on Build GRCh38
rs1034843705 Chr.15: 65579321-65579321 on Build GRCh38
rs761040810 Chr.15: 65579433-65579433 on Build GRCh38
rs759347980 Chr.15: 65579378-65579380 on Build GRCh38
rs770242616 Chr.15: 65579512-65579512 on Build GRCh38
rs753123146 Chr.15: 65579427-65579427 on Build GRCh38
rs997205698 Chr.15: 65579421-65579421 on Build GRCh38
rs745353624 Chr.15: 65579496-65579496 on Build GRCh38
rs764641210 Chr.15: 65579438-65579438 on Build GRCh38
rs773945014 Chr.15: 65579417-65579417 on Build GRCh38
rs762255473 Chr.15: 65579441-65579441 on Build GRCh38
rs751387170 Chr.15: 65579377-65579378 on Build GRCh38
rs777045881 Chr.15: 65579440-65579440 on Build GRCh38
rs781181850 Chr.15: 65579474-65579474 on Build GRCh38
rs575493112 Chr.15: 65579196-65579196 on Build GRCh38
rs914789394 Chr.15: 65579092-65579092 on Build GRCh38
rs887679806 Chr.15: 65579158-65579158 on Build GRCh38
rs138028542 Chr.15: 65579182-65579182 on Build GRCh38
rs542736272 Chr.15: 65579380-65579380 on Build GRCh38
rs372521742 Chr.15: 65579303-65579303 on Build GRCh38
rs1027598226 Chr.15: 65579144-65579144 on Build GRCh38
rs767350473 Chr.15: 65579382-65579384 on Build GRCh38
rs372145138 Chr.15: 65579522-65579522 on Build GRCh38
rs773925803 Chr.15: 65579495-65579495 on Build GRCh38
rs10914 Chr.15: 65579082-65579082 on Build GRCh38
rs750545819 Chr.15: 65579501-65579527 on Build GRCh38
rs777551938 Chr.15: 65579480-65579480 on Build GRCh38
rs770363902 Chr.15: 65579401-65579401 on Build GRCh38
rs926145174 Chr.15: 65579162-65579162 on Build GRCh38
rs766201119 Chr.15: 65579371-65579375 on Build GRCh38
rs765709152 Chr.15: 65579451-65579451 on Build GRCh38
rs1040493698 Chr.15: 65579127-65579127 on Build GRCh38
rs751633940 Chr.15: 65579463-65579463 on Build GRCh38
rs559239976 Chr.15: 65579187-65579187 on Build GRCh38
rs766318959 Chr.15: 65579460-65579460 on Build GRCh38
rs1028761188 Chr.15: 65579128-65579131 on Build GRCh38
rs750605772 Chr.15: 65579452-65579452 on Build GRCh38
rs1041920713 Chr.15: 65579107-65579107 on Build GRCh38
rs772842120 Chr.15: 65579394-65579394 on Build GRCh38
rs761657300 Chr.15: 65579251-65579251 on Build GRCh38
rs141034504 Chr.15: 65579473-65579473 on Build GRCh38
rs773733142 Chr.15: 65579520-65579520 on Build GRCh38
rs1040164373 Chr.15: 65579475-65579475 on Build GRCh38
rs187402322 Chr.15: 65579490-65579490 on Build GRCh38
rs368498639 Chr.15: 65579478-65579478 on Build GRCh38
rs773016288 Chr.15: 65579113-65579113 on Build GRCh38
rs11484 Chr.15: 65579360-65579360 on Build GRCh38
rs778839370 Chr.15: 65579403-65579404 on Build GRCh38
rs573600764 Chr.15: 65579087-65579087 on Build GRCh38
rs553174820 Chr.15: 65579139-65579139 on Build GRCh38
rs891276029 Chr.15: 65579110-65579110 on Build GRCh38
rs756101783 Chr.15: 65579387-65579388 on Build GRCh38
rs796115282 Chr.15: 65579383-65579383 on Build GRCh38
rs936260183 Chr.15: 65579173-65579173 on Build GRCh38

View Full Product Details