Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 88621159-88621673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AEN
Entrez Gene ID
64782
Gene Name
apoptosis enhancing nuclease
Gene Aliases
ISG20L1, pp12744
UniGene
Hs.436102
Gene Chromosome Location
Chr.15: 88604601-88632281 on Build GRCh38
Transcripts
NM_022767.3 XM_005254966.1 XM_005254967.1 XM_006720645.3 XM_011521905.2 XM_017022489.1 XM_017022490.1

SNP Detail

SNP ID Chromosome Location
rs1000122387 Chr.15: 88621469-88621469 on Build GRCh38
rs902666189 Chr.15: 88621460-88621460 on Build GRCh38
rs978782039 Chr.15: 88621198-88621198 on Build GRCh38
rs1032518868 Chr.15: 88621475-88621475 on Build GRCh38
rs570090640 Chr.15: 88621299-88621299 on Build GRCh38
rs913441276 Chr.15: 88621197-88621197 on Build GRCh38
rs117218125 Chr.15: 88621350-88621350 on Build GRCh38
rs930565520 Chr.15: 88621400-88621400 on Build GRCh38
rs1019770631 Chr.15: 88621326-88621326 on Build GRCh38
rs900544157 Chr.15: 88621477-88621477 on Build GRCh38
rs981623238 Chr.15: 88621361-88621361 on Build GRCh38
rs1022583985 Chr.15: 88621514-88621514 on Build GRCh38
rs386786159 Chr.15: 88621401-88621403 on Build GRCh38
rs1036922886 Chr.15: 88621405-88621405 on Build GRCh38
rs952795364 Chr.15: 88621490-88621490 on Build GRCh38
rs545680983 Chr.15: 88621342-88621342 on Build GRCh38
rs1055209493 Chr.15: 88621257-88621257 on Build GRCh38
rs1049765292 Chr.15: 88621422-88621422 on Build GRCh38
rs944064364 Chr.15: 88621591-88621591 on Build GRCh38
rs906178834 Chr.15: 88621232-88621232 on Build GRCh38
rs1033006046 Chr.15: 88621485-88621485 on Build GRCh38
rs578229257 Chr.15: 88621336-88621336 on Build GRCh38
rs553579596 Chr.15: 88621321-88621321 on Build GRCh38
rs907764187 Chr.15: 88621651-88621651 on Build GRCh38
rs958292076 Chr.15: 88621483-88621483 on Build GRCh38
rs898487176 Chr.15: 88621314-88621314 on Build GRCh38
rs899044985 Chr.15: 88621266-88621266 on Build GRCh38
rs1049711435 Chr.15: 88621589-88621589 on Build GRCh38
rs1041104768 Chr.15: 88621606-88621606 on Build GRCh38
rs1053252135 Chr.15: 88621195-88621195 on Build GRCh38
rs73451723 Chr.15: 88621182-88621182 on Build GRCh38
rs1008656298 Chr.15: 88621466-88621466 on Build GRCh38
rs967317807 Chr.15: 88621329-88621329 on Build GRCh38
rs995519480 Chr.15: 88621315-88621315 on Build GRCh38
rs924275890 Chr.15: 88621203-88621203 on Build GRCh38
rs569672982 Chr.15: 88621181-88621181 on Build GRCh38
rs937763377 Chr.15: 88621378-88621378 on Build GRCh38
rs961211186 Chr.15: 88621549-88621549 on Build GRCh38
rs145808040 Chr.15: 88621267-88621267 on Build GRCh38
rs966991587 Chr.15: 88621184-88621184 on Build GRCh38
rs375618035 Chr.15: 88621316-88621316 on Build GRCh38
rs911205351 Chr.15: 88621590-88621590 on Build GRCh38
rs999616571 Chr.15: 88621642-88621642 on Build GRCh38
rs1037380035 Chr.15: 88621306-88621306 on Build GRCh38
rs989831267 Chr.15: 88621222-88621222 on Build GRCh38
rs991068654 Chr.15: 88621509-88621509 on Build GRCh38
rs936258021 Chr.15: 88621193-88621193 on Build GRCh38
rs1054798806 Chr.15: 88621385-88621385 on Build GRCh38
rs948493024 Chr.15: 88621368-88621368 on Build GRCh38
rs555777931 Chr.15: 88621183-88621183 on Build GRCh38
rs981572503 Chr.15: 88621536-88621536 on Build GRCh38
rs937666454 Chr.15: 88621255-88621255 on Build GRCh38
rs927667948 Chr.15: 88621367-88621367 on Build GRCh38
rs559962082 Chr.15: 88621423-88621423 on Build GRCh38
rs560975808 Chr.15: 88621516-88621516 on Build GRCh38
rs920256844 Chr.15: 88621595-88621595 on Build GRCh38
rs971650063 Chr.15: 88621525-88621525 on Build GRCh38
rs935691722 Chr.15: 88621215-88621215 on Build GRCh38
rs931262717 Chr.15: 88621414-88621414 on Build GRCh38
rs928470463 Chr.15: 88621399-88621399 on Build GRCh38
rs376662874 Chr.15: 88621335-88621335 on Build GRCh38
rs948522285 Chr.15: 88621244-88621244 on Build GRCh38
rs1026098097 Chr.15: 88621304-88621304 on Build GRCh38
rs188843509 Chr.15: 88621577-88621577 on Build GRCh38
rs878875068 Chr.15: 88621409-88621409 on Build GRCh38
rs898432693 Chr.15: 88621410-88621410 on Build GRCh38
rs528153262 Chr.15: 88621519-88621519 on Build GRCh38
rs758627929 Chr.15: 88621436-88621436 on Build GRCh38
rs889875859 Chr.15: 88621320-88621320 on Build GRCh38
rs11858257 Chr.15: 88621643-88621643 on Build GRCh38
rs556854069 Chr.15: 88621180-88621180 on Build GRCh38
rs886303132 Chr.15: 88621312-88621312 on Build GRCh38
rs939879070 Chr.15: 88621403-88621403 on Build GRCh38
rs915681957 Chr.15: 88621236-88621236 on Build GRCh38
rs970152497 Chr.15: 88621515-88621515 on Build GRCh38
rs1020139548 Chr.15: 88621433-88621433 on Build GRCh38
rs972563443 Chr.15: 88621571-88621571 on Build GRCh38
rs981637766 Chr.15: 88621390-88621390 on Build GRCh38
rs531286588 Chr.15: 88621372-88621372 on Build GRCh38
rs1008307725 Chr.15: 88621430-88621430 on Build GRCh38
rs546312450 Chr.15: 88621573-88621573 on Build GRCh38
rs902600866 Chr.15: 88621638-88621638 on Build GRCh38
rs939950519 Chr.15: 88621303-88621303 on Build GRCh38
rs1008777744 Chr.15: 88621322-88621322 on Build GRCh38
rs759983530 Chr.15: 88621332-88621332 on Build GRCh38
rs957012780 Chr.15: 88621341-88621341 on Build GRCh38

View Full Product Details