Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 88631385-88631923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AEN
Entrez Gene ID
64782
Gene Name
apoptosis enhancing nuclease
Gene Aliases
ISG20L1, pp12744
UniGene
Hs.436102
Gene Chromosome Location
Chr.15: 88604601-88632281 on Build GRCh38
Transcripts
NM_022767.3 XM_005254966.1 XM_005254967.1 XM_006720645.3 XM_011521905.2 XM_017022489.1 XM_017022490.1

SNP Detail

SNP ID Chromosome Location
rs1009061404 Chr.15: 88631653-88631653 on Build GRCh38
rs761931693 Chr.15: 88631876-88631876 on Build GRCh38
rs1048696182 Chr.15: 88631492-88631492 on Build GRCh38
rs1047793717 Chr.15: 88631792-88631792 on Build GRCh38
rs770132836 Chr.15: 88631801-88631801 on Build GRCh38
rs577574131 Chr.15: 88631479-88631479 on Build GRCh38
rs1028501078 Chr.15: 88631668-88631668 on Build GRCh38
rs913819790 Chr.15: 88631687-88631687 on Build GRCh38
rs1031497449 Chr.15: 88631853-88631853 on Build GRCh38
rs899141699 Chr.15: 88631618-88631618 on Build GRCh38
rs778133763 Chr.15: 88631658-88631658 on Build GRCh38
rs115789711 Chr.15: 88631458-88631458 on Build GRCh38
rs754064749 Chr.15: 88631848-88631849 on Build GRCh38
rs532149008 Chr.15: 88631437-88631437 on Build GRCh38
rs1031548511 Chr.15: 88631556-88631556 on Build GRCh38
rs374237404 Chr.15: 88631827-88631827 on Build GRCh38
rs747654696 Chr.15: 88631555-88631555 on Build GRCh38
rs116836487 Chr.15: 88631877-88631877 on Build GRCh38
rs920677991 Chr.15: 88631768-88631768 on Build GRCh38
rs999034691 Chr.15: 88631834-88631834 on Build GRCh38
rs568875863 Chr.15: 88631579-88631579 on Build GRCh38
rs966634662 Chr.15: 88631701-88631701 on Build GRCh38
rs965481625 Chr.15: 88631658-88631662 on Build GRCh38
rs371680691 Chr.15: 88631541-88631541 on Build GRCh38
rs1045832817 Chr.15: 88631427-88631427 on Build GRCh38
rs989381446 Chr.15: 88631532-88631532 on Build GRCh38
rs945176364 Chr.15: 88631552-88631552 on Build GRCh38
rs576578123 Chr.15: 88631657-88631658 on Build GRCh38
rs546883482 Chr.15: 88631772-88631772 on Build GRCh38
rs771359728 Chr.15: 88631683-88631683 on Build GRCh38
rs142747428 Chr.15: 88631448-88631448 on Build GRCh38
rs775778085 Chr.15: 88631803-88631803 on Build GRCh38
rs936350955 Chr.15: 88631520-88631521 on Build GRCh38
rs915938347 Chr.15: 88631423-88631423 on Build GRCh38
rs966240018 Chr.15: 88631549-88631549 on Build GRCh38
rs888796097 Chr.15: 88631498-88631498 on Build GRCh38
rs937860083 Chr.15: 88631764-88631764 on Build GRCh38
rs1037141023 Chr.15: 88631744-88631744 on Build GRCh38
rs529769772 Chr.15: 88631818-88631818 on Build GRCh38
rs928648687 Chr.15: 88631889-88631889 on Build GRCh38
rs915889364 Chr.15: 88631660-88631660 on Build GRCh38
rs981461616 Chr.15: 88631888-88631888 on Build GRCh38
rs1047969707 Chr.15: 88631637-88631640 on Build GRCh38
rs1055187260 Chr.15: 88631765-88631765 on Build GRCh38
rs780464770 Chr.15: 88631722-88631722 on Build GRCh38
rs1055517997 Chr.15: 88631595-88631595 on Build GRCh38
rs764340767 Chr.15: 88631812-88631812 on Build GRCh38
rs957691310 Chr.15: 88631675-88631675 on Build GRCh38
rs778916238 Chr.15: 88631885-88631885 on Build GRCh38
rs1019371096 Chr.15: 88631457-88631457 on Build GRCh38
rs1049986450 Chr.15: 88631793-88631793 on Build GRCh38
rs886730660 Chr.15: 88631638-88631638 on Build GRCh38
rs981897903 Chr.15: 88631718-88631718 on Build GRCh38
rs560770899 Chr.15: 88631658-88631658 on Build GRCh38
rs957256953 Chr.15: 88631592-88631592 on Build GRCh38
rs957206790 Chr.15: 88631857-88631857 on Build GRCh38
rs928755719 Chr.15: 88631442-88631442 on Build GRCh38
rs777169791 Chr.15: 88631767-88631767 on Build GRCh38
rs948784507 Chr.15: 88631425-88631425 on Build GRCh38
rs940211330 Chr.15: 88631443-88631443 on Build GRCh38
rs144859537 Chr.15: 88631838-88631838 on Build GRCh38
rs146546255 Chr.15: 88631529-88631529 on Build GRCh38
rs2278598 Chr.15: 88631517-88631517 on Build GRCh38
rs1020608690 Chr.15: 88631892-88631892 on Build GRCh38
rs543101779 Chr.15: 88631417-88631417 on Build GRCh38
rs961470090 Chr.15: 88631899-88631899 on Build GRCh38
rs928727568 Chr.15: 88631715-88631715 on Build GRCh38
rs570147012 Chr.15: 88631865-88631865 on Build GRCh38
rs115459440 Chr.15: 88631623-88631623 on Build GRCh38
rs940137002 Chr.15: 88631730-88631730 on Build GRCh38
rs528336609 Chr.15: 88631703-88631703 on Build GRCh38
rs141259959 Chr.15: 88631808-88631808 on Build GRCh38

View Full Product Details