Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1339229-1339729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP3
Entrez Gene ID
8938
Gene Name
BAI1 associated protein 3
Gene Aliases
BAP3
UniGene
Hs.458427
Gene Chromosome Location
Chr.16: 1333605-1349441 on Build GRCh38
Transcripts
NM_001199096.1 NM_001199097.1 NM_001199098.1 NM_001199099.1 NM_001286464.1 NM_003933.4 XM_011522728.1 XM_011522729.1 XM_011522730.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4630163 c.411C>T p.G137G Chr.16: 1339555-1339555 on Build GRCh38
471315 c.423G>A p.E141E Chr.16: 1339513-1339513 on Build GRCh38
5743720 c.391T>C p.Y131H Chr.16: 1339535-1339535 on Build GRCh38
2143513 c.486G>T p.E162D Chr.16: 1339576-1339576 on Build GRCh38
5743719 c.445T>C p.Y149H Chr.16: 1339535-1339535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014766671 Chr.16: 1339327-1339327 on Build GRCh38
rs148244582 Chr.16: 1339686-1339686 on Build GRCh38
rs756899333 Chr.16: 1339460-1339460 on Build GRCh38
rs369732219 Chr.16: 1339637-1339637 on Build GRCh38
rs188806500 Chr.16: 1339256-1339256 on Build GRCh38
rs536530061 Chr.16: 1339388-1339388 on Build GRCh38
rs372954811 Chr.16: 1339649-1339649 on Build GRCh38
rs565255025 Chr.16: 1339453-1339453 on Build GRCh38
rs751046433 Chr.16: 1339500-1339500 on Build GRCh38
rs201425803 Chr.16: 1339492-1339492 on Build GRCh38
rs1009884326 Chr.16: 1339395-1339395 on Build GRCh38
rs771315907 Chr.16: 1339292-1339292 on Build GRCh38
rs367845210 Chr.16: 1339284-1339284 on Build GRCh38
rs961845414 Chr.16: 1339436-1339436 on Build GRCh38
rs117780030 Chr.16: 1339465-1339465 on Build GRCh38
rs1033890563 Chr.16: 1339378-1339378 on Build GRCh38
rs971941910 Chr.16: 1339257-1339257 on Build GRCh38
rs757783175 Chr.16: 1339445-1339445 on Build GRCh38
rs923080069 Chr.16: 1339259-1339259 on Build GRCh38
rs950097625 Chr.16: 1339501-1339501 on Build GRCh38
rs750097987 Chr.16: 1339554-1339554 on Build GRCh38
rs374580348 Chr.16: 1339570-1339570 on Build GRCh38
rs1050326475 Chr.16: 1339268-1339268 on Build GRCh38
rs372874239 Chr.16: 1339528-1339528 on Build GRCh38
rs78268478 Chr.16: 1339628-1339628 on Build GRCh38
rs753733338 Chr.16: 1339640-1339640 on Build GRCh38
rs945365408 Chr.16: 1339674-1339674 on Build GRCh38
rs866185784 Chr.16: 1339698-1339698 on Build GRCh38
rs753039186 Chr.16: 1339654-1339654 on Build GRCh38
rs192028603 Chr.16: 1339377-1339377 on Build GRCh38
rs533648313 Chr.16: 1339627-1339627 on Build GRCh38
rs371283238 Chr.16: 1339512-1339512 on Build GRCh38
rs562830339 Chr.16: 1339709-1339709 on Build GRCh38
rs1019041265 Chr.16: 1339400-1339400 on Build GRCh38
rs747040960 Chr.16: 1339527-1339527 on Build GRCh38
rs551464937 Chr.16: 1339398-1339398 on Build GRCh38
rs763970958 Chr.16: 1339541-1339541 on Build GRCh38
rs773881878 Chr.16: 1339542-1339542 on Build GRCh38
rs780446143 Chr.16: 1339462-1339462 on Build GRCh38
rs144888270 Chr.16: 1339567-1339567 on Build GRCh38
rs765039943 Chr.16: 1339290-1339290 on Build GRCh38
rs113227869 Chr.16: 1339265-1339265 on Build GRCh38
rs772341229 Chr.16: 1339264-1339264 on Build GRCh38
rs757355425 Chr.16: 1339574-1339574 on Build GRCh38
rs747270262 Chr.16: 1339476-1339476 on Build GRCh38
rs1030499988 Chr.16: 1339576-1339576 on Build GRCh38
rs557488567 Chr.16: 1339581-1339581 on Build GRCh38
rs1054573298 Chr.16: 1339516-1339516 on Build GRCh38
rs773418345 Chr.16: 1339471-1339471 on Build GRCh38
rs200855970 Chr.16: 1339556-1339556 on Build GRCh38
rs767169250 Chr.16: 1339544-1339544 on Build GRCh38
rs984599861 Chr.16: 1339593-1339593 on Build GRCh38
rs772097774 Chr.16: 1339469-1339469 on Build GRCh38
rs772743434 Chr.16: 1339630-1339630 on Build GRCh38
rs940622180 Chr.16: 1339490-1339490 on Build GRCh38
rs779735780 Chr.16: 1339619-1339619 on Build GRCh38
rs1033255246 Chr.16: 1339390-1339390 on Build GRCh38
rs776594670 Chr.16: 1339483-1339483 on Build GRCh38
rs1010041374 Chr.16: 1339310-1339310 on Build GRCh38
rs369126457 Chr.16: 1339511-1339511 on Build GRCh38
rs113817326 Chr.16: 1339393-1339393 on Build GRCh38
rs751484936 Chr.16: 1339582-1339582 on Build GRCh38
rs954413321 Chr.16: 1339580-1339580 on Build GRCh38
rs1016314276 Chr.16: 1339396-1339396 on Build GRCh38
rs556026206 Chr.16: 1339383-1339383 on Build GRCh38
rs1046581894 Chr.16: 1339356-1339356 on Build GRCh38
rs927360078 Chr.16: 1339666-1339666 on Build GRCh38
rs972212338 Chr.16: 1339454-1339454 on Build GRCh38
rs368421949 Chr.16: 1339622-1339622 on Build GRCh38
rs763660506 Chr.16: 1339498-1339498 on Build GRCh38
rs775234980 Chr.16: 1339494-1339494 on Build GRCh38
rs910596998 Chr.16: 1339269-1339269 on Build GRCh38
rs751696341 Chr.16: 1339573-1339573 on Build GRCh38
rs573543196 Chr.16: 1339665-1339665 on Build GRCh38
rs758903417 Chr.16: 1339517-1339517 on Build GRCh38
rs778053041 Chr.16: 1339520-1339520 on Build GRCh38
rs142556073 Chr.16: 1339539-1339539 on Build GRCh38
rs749605753 Chr.16: 1339463-1339463 on Build GRCh38
rs773508188 Chr.16: 1339273-1339273 on Build GRCh38
rs768260176 Chr.16: 1339374-1339374 on Build GRCh38
rs751271391 Chr.16: 1339459-1339459 on Build GRCh38
rs573457843 Chr.16: 1339287-1339287 on Build GRCh38
rs761756651 Chr.16: 1339260-1339260 on Build GRCh38
rs373188358 Chr.16: 1339623-1339623 on Build GRCh38
rs752285683 Chr.16: 1339295-1339295 on Build GRCh38
rs376349828 Chr.16: 1339285-1339285 on Build GRCh38
rs762673539 Chr.16: 1339510-1339510 on Build GRCh38
rs748270308 Chr.16: 1339467-1339467 on Build GRCh38
rs760963108 Chr.16: 1339275-1339275 on Build GRCh38
rs769536428 Chr.16: 1339538-1339538 on Build GRCh38
rs370207479 Chr.16: 1339493-1339493 on Build GRCh38
rs770027564 Chr.16: 1339597-1339597 on Build GRCh38
rs913935506 Chr.16: 1339656-1339656 on Build GRCh38
rs151334572 Chr.16: 1339571-1339571 on Build GRCh38
rs974493458 Chr.16: 1339433-1339433 on Build GRCh38
rs895876188 Chr.16: 1339325-1339325 on Build GRCh38
rs147709691 Chr.16: 1339505-1339505 on Build GRCh38
rs547870649 Chr.16: 1339477-1339477 on Build GRCh38
rs9708682 Chr.16: 1339359-1339359 on Build GRCh38
rs761244619 Chr.16: 1339543-1339543 on Build GRCh38
rs1044508177 Chr.16: 1339342-1339342 on Build GRCh38
rs374144851 Chr.16: 1339464-1339464 on Build GRCh38
rs892566492 Chr.16: 1339301-1339301 on Build GRCh38
rs965792007 Chr.16: 1339416-1339416 on Build GRCh38
rs762839563 Chr.16: 1339540-1339540 on Build GRCh38
rs765556289 Chr.16: 1339564-1339564 on Build GRCh38
rs367660388 Chr.16: 1339507-1339507 on Build GRCh38
rs762935325 Chr.16: 1339495-1339495 on Build GRCh38
rs35223486 Chr.16: 1339545-1339546 on Build GRCh38

View Full Product Details