Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1349787-1350277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSR3
Entrez Gene ID
115939
Gene Name
TSR3, acp transferase ribosome maturation factor
Gene Aliases
C16orf42
UniGene
Hs.134846
Gene Chromosome Location
Chr.16: 1349240-1351872 on Build GRCh38
Transcripts
NM_001001410.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666964 c.651G>A p.A217A Chr.16: 1350110-1350110 on Build GRCh38
3506073 c.582C>T p.F194F Chr.16: 1350179-1350179 on Build GRCh38
4666965 c.633G>A p.P211P Chr.16: 1350128-1350128 on Build GRCh38
6325496 c.596G>A p.R199H Chr.16: 1350165-1350165 on Build GRCh38
7085229 c.595C>A p.R199S Chr.16: 1350166-1350166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757025347 Chr.16: 1349884-1349884 on Build GRCh38
rs779028143 Chr.16: 1349868-1349873 on Build GRCh38
rs779507996 Chr.16: 1350125-1350125 on Build GRCh38
rs558014633 Chr.16: 1350128-1350128 on Build GRCh38
rs757791437 Chr.16: 1350045-1350045 on Build GRCh38
rs1027394258 Chr.16: 1349830-1349830 on Build GRCh38
rs774553826 Chr.16: 1350012-1350012 on Build GRCh38
rs765918112 Chr.16: 1350174-1350174 on Build GRCh38
rs778415056 Chr.16: 1350187-1350187 on Build GRCh38
rs143030507 Chr.16: 1350165-1350165 on Build GRCh38
rs142802890 Chr.16: 1350232-1350232 on Build GRCh38
rs764224735 Chr.16: 1349875-1349875 on Build GRCh38
rs895006618 Chr.16: 1350177-1350177 on Build GRCh38
rs372809768 Chr.16: 1349859-1349859 on Build GRCh38
rs761782920 Chr.16: 1350013-1350013 on Build GRCh38
rs762008010 Chr.16: 1349921-1349921 on Build GRCh38
rs1003337137 Chr.16: 1350044-1350044 on Build GRCh38
rs759044782 Chr.16: 1350179-1350179 on Build GRCh38
rs768669545 Chr.16: 1349873-1349874 on Build GRCh38
rs200343582 Chr.16: 1349967-1349967 on Build GRCh38
rs773364768 Chr.16: 1349960-1349961 on Build GRCh38
rs775329353 Chr.16: 1350139-1350139 on Build GRCh38
rs757945435 Chr.16: 1350093-1350093 on Build GRCh38
rs141978358 Chr.16: 1350092-1350092 on Build GRCh38
rs906338834 Chr.16: 1349820-1349820 on Build GRCh38
rs766979180 Chr.16: 1350152-1350152 on Build GRCh38
rs377359576 Chr.16: 1350254-1350254 on Build GRCh38
rs951537189 Chr.16: 1349841-1349841 on Build GRCh38
rs751764753 Chr.16: 1349878-1349878 on Build GRCh38
rs775210664 Chr.16: 1350212-1350212 on Build GRCh38
rs752098196 Chr.16: 1350010-1350011 on Build GRCh38
rs769009010 Chr.16: 1350011-1350011 on Build GRCh38
rs183711057 Chr.16: 1349992-1349992 on Build GRCh38
rs761027314 Chr.16: 1350079-1350079 on Build GRCh38
rs761342480 Chr.16: 1350228-1350228 on Build GRCh38
rs770804513 Chr.16: 1349839-1349840 on Build GRCh38
rs780123786 Chr.16: 1349897-1349897 on Build GRCh38
rs768217236 Chr.16: 1349905-1349905 on Build GRCh38
rs770547806 Chr.16: 1350058-1350058 on Build GRCh38
rs773465601 Chr.16: 1350129-1350129 on Build GRCh38
rs1052197090 Chr.16: 1349983-1349983 on Build GRCh38
rs148141495 Chr.16: 1350065-1350065 on Build GRCh38
rs906287978 Chr.16: 1350027-1350027 on Build GRCh38
rs199629353 Chr.16: 1349960-1349960 on Build GRCh38
rs139006693 Chr.16: 1350157-1350157 on Build GRCh38
rs917702750 Chr.16: 1350133-1350133 on Build GRCh38
rs773909730 Chr.16: 1349910-1349910 on Build GRCh38
rs755228580 Chr.16: 1350123-1350123 on Build GRCh38
rs753065771 Chr.16: 1350245-1350245 on Build GRCh38
rs774035457 Chr.16: 1349823-1349823 on Build GRCh38
rs369337216 Chr.16: 1349958-1349958 on Build GRCh38
rs776760370 Chr.16: 1349889-1349890 on Build GRCh38
rs201725773 Chr.16: 1350110-1350110 on Build GRCh38
rs747580455 Chr.16: 1349977-1349977 on Build GRCh38
rs111929985 Chr.16: 1349994-1349994 on Build GRCh38
rs748555427 Chr.16: 1350127-1350127 on Build GRCh38
rs375746230 Chr.16: 1349890-1349890 on Build GRCh38
rs760063483 Chr.16: 1350062-1350064 on Build GRCh38
rs763499630 Chr.16: 1349997-1349997 on Build GRCh38
rs767626640 Chr.16: 1349952-1349952 on Build GRCh38
rs755841038 Chr.16: 1350164-1350164 on Build GRCh38
rs200565292 Chr.16: 1350204-1350204 on Build GRCh38
rs995829433 Chr.16: 1350019-1350019 on Build GRCh38
rs56707013 Chr.16: 1349978-1349978 on Build GRCh38
rs777432530 Chr.16: 1350046-1350046 on Build GRCh38
rs758781838 Chr.16: 1350247-1350247 on Build GRCh38
rs1054953125 Chr.16: 1350176-1350176 on Build GRCh38
rs201696319 Chr.16: 1350170-1350170 on Build GRCh38
rs752260875 Chr.16: 1350082-1350100 on Build GRCh38
rs780808158 Chr.16: 1350121-1350121 on Build GRCh38
rs367714564 Chr.16: 1349846-1349846 on Build GRCh38
rs763051249 Chr.16: 1349942-1349942 on Build GRCh38
rs372845875 Chr.16: 1349959-1349959 on Build GRCh38
rs1043621709 Chr.16: 1350201-1350202 on Build GRCh38
rs200822692 Chr.16: 1349860-1349860 on Build GRCh38
rs773573593 Chr.16: 1349844-1349844 on Build GRCh38
rs969197452 Chr.16: 1349877-1349877 on Build GRCh38
rs767159335 Chr.16: 1350146-1350146 on Build GRCh38
rs771340303 Chr.16: 1350136-1350136 on Build GRCh38
rs375400644 Chr.16: 1350144-1350144 on Build GRCh38
rs746614922 Chr.16: 1350052-1350052 on Build GRCh38
rs753790049 Chr.16: 1349970-1349970 on Build GRCh38
rs1017661178 Chr.16: 1350251-1350251 on Build GRCh38
rs747184246 Chr.16: 1349922-1349922 on Build GRCh38
rs760581168 Chr.16: 1350016-1350016 on Build GRCh38
rs766051684 Chr.16: 1350241-1350241 on Build GRCh38
rs756697549 Chr.16: 1350145-1350145 on Build GRCh38
rs756184815 Chr.16: 1349896-1349896 on Build GRCh38
rs111267196 Chr.16: 1350227-1350227 on Build GRCh38
rs996888230 Chr.16: 1350229-1350229 on Build GRCh38
rs761356974 Chr.16: 1349950-1349951 on Build GRCh38
rs367737359 Chr.16: 1350059-1350059 on Build GRCh38
rs777027510 Chr.16: 1349851-1349851 on Build GRCh38
rs759381551 Chr.16: 1350137-1350137 on Build GRCh38
rs759891341 Chr.16: 1349938-1349938 on Build GRCh38
rs117781264 Chr.16: 1350239-1350239 on Build GRCh38
rs777594223 Chr.16: 1350241-1350242 on Build GRCh38
rs368854136 Chr.16: 1349966-1349966 on Build GRCh38
rs142109530 Chr.16: 1350111-1350111 on Build GRCh38
rs372514570 Chr.16: 1350240-1350240 on Build GRCh38
rs371843849 Chr.16: 1349945-1349945 on Build GRCh38
rs377633003 Chr.16: 1349999-1349999 on Build GRCh38
rs768593176 Chr.16: 1350226-1350226 on Build GRCh38
rs760085640 Chr.16: 1349855-1349855 on Build GRCh38
rs745623676 Chr.16: 1349891-1349891 on Build GRCh38
rs773592112 Chr.16: 1350077-1350077 on Build GRCh38
rs766687285 Chr.16: 1350000-1350001 on Build GRCh38
rs764185089 Chr.16: 1349944-1349944 on Build GRCh38
rs755525157 Chr.16: 1350106-1350107 on Build GRCh38
rs775178105 Chr.16: 1349839-1349839 on Build GRCh38
rs954796172 Chr.16: 1350100-1350100 on Build GRCh38
rs754984027 Chr.16: 1350020-1350020 on Build GRCh38
rs370888956 Chr.16: 1350014-1350014 on Build GRCh38
rs766730874 Chr.16: 1349847-1349847 on Build GRCh38
rs776639568 Chr.16: 1350090-1350090 on Build GRCh38
rs777094302 Chr.16: 1349936-1349936 on Build GRCh38
rs372183764 Chr.16: 1350131-1350131 on Build GRCh38
rs781497708 Chr.16: 1350203-1350203 on Build GRCh38
rs762833838 Chr.16: 1350141-1350141 on Build GRCh38
rs778431583 Chr.16: 1350253-1350253 on Build GRCh38
rs113774682 Chr.16: 1350107-1350107 on Build GRCh38
rs965418486 Chr.16: 1350103-1350103 on Build GRCh38
rs555149094 Chr.16: 1350087-1350087 on Build GRCh38
rs72779215 Chr.16: 1350018-1350018 on Build GRCh38
rs778534973 Chr.16: 1349974-1349974 on Build GRCh38
rs764014118 Chr.16: 1350143-1350143 on Build GRCh38
rs765236752 Chr.16: 1350091-1350091 on Build GRCh38
rs965534530 Chr.16: 1349889-1349889 on Build GRCh38
rs757813762 Chr.16: 1350256-1350256 on Build GRCh38
rs748266154 Chr.16: 1350064-1350064 on Build GRCh38
rs140020940 Chr.16: 1350223-1350223 on Build GRCh38
rs777392996 Chr.16: 1349986-1349986 on Build GRCh38
rs754293003 Chr.16: 1350215-1350215 on Build GRCh38
rs369589750 Chr.16: 1349941-1349941 on Build GRCh38
rs375137026 Chr.16: 1350194-1350194 on Build GRCh38
rs774660643 Chr.16: 1350062-1350062 on Build GRCh38
rs747791504 Chr.16: 1349915-1349915 on Build GRCh38
rs778712535 Chr.16: 1350028-1350028 on Build GRCh38
rs771826060 Chr.16: 1349980-1349980 on Build GRCh38
rs558963521 Chr.16: 1349962-1349966 on Build GRCh38
rs752311981 Chr.16: 1350033-1350033 on Build GRCh38
rs749675012 Chr.16: 1350122-1350122 on Build GRCh38
rs745788806 Chr.16: 1349870-1349872 on Build GRCh38
rs766990986 Chr.16: 1350061-1350062 on Build GRCh38
rs1026548543 Chr.16: 1349817-1349817 on Build GRCh38
rs747102484 Chr.16: 1350189-1350189 on Build GRCh38
rs750609781 Chr.16: 1350015-1350015 on Build GRCh38
rs770023401 Chr.16: 1350205-1350205 on Build GRCh38
rs754473087 Chr.16: 1349972-1349972 on Build GRCh38

View Full Product Details