Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1349977-1350476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSR3
Entrez Gene ID
115939
Gene Name
TSR3, acp transferase ribosome maturation factor
Gene Aliases
C16orf42
UniGene
Hs.134846
Gene Chromosome Location
Chr.16: 1349240-1351872 on Build GRCh38
Transcripts
NM_001001410.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666964 c.651G>A p.A217A Chr.16: 1350110-1350110 on Build GRCh38
3506073 c.582C>T p.F194F Chr.16: 1350179-1350179 on Build GRCh38
4666965 c.633G>A p.P211P Chr.16: 1350128-1350128 on Build GRCh38
6325496 c.596G>A p.R199H Chr.16: 1350165-1350165 on Build GRCh38
7085229 c.595C>A p.R199S Chr.16: 1350166-1350166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779507996 Chr.16: 1350125-1350125 on Build GRCh38
rs558014633 Chr.16: 1350128-1350128 on Build GRCh38
rs749085077 Chr.16: 1350267-1350267 on Build GRCh38
rs757791437 Chr.16: 1350045-1350045 on Build GRCh38
rs774553826 Chr.16: 1350012-1350012 on Build GRCh38
rs927645023 Chr.16: 1350314-1350314 on Build GRCh38
rs765918112 Chr.16: 1350174-1350174 on Build GRCh38
rs778415056 Chr.16: 1350187-1350187 on Build GRCh38
rs143030507 Chr.16: 1350165-1350165 on Build GRCh38
rs142802890 Chr.16: 1350232-1350232 on Build GRCh38
rs895006618 Chr.16: 1350177-1350177 on Build GRCh38
rs1000826301 Chr.16: 1350444-1350444 on Build GRCh38
rs761782920 Chr.16: 1350013-1350013 on Build GRCh38
rs1003337137 Chr.16: 1350044-1350044 on Build GRCh38
rs759044782 Chr.16: 1350179-1350179 on Build GRCh38
rs931201998 Chr.16: 1350323-1350323 on Build GRCh38
rs757945435 Chr.16: 1350093-1350093 on Build GRCh38
rs775329353 Chr.16: 1350139-1350139 on Build GRCh38
rs141978358 Chr.16: 1350092-1350092 on Build GRCh38
rs1028270292 Chr.16: 1350439-1350439 on Build GRCh38
rs377359576 Chr.16: 1350254-1350254 on Build GRCh38
rs766979180 Chr.16: 1350152-1350152 on Build GRCh38
rs775210664 Chr.16: 1350212-1350212 on Build GRCh38
rs752098196 Chr.16: 1350010-1350011 on Build GRCh38
rs769009010 Chr.16: 1350011-1350011 on Build GRCh38
rs761027314 Chr.16: 1350079-1350079 on Build GRCh38
rs761342480 Chr.16: 1350228-1350228 on Build GRCh38
rs951071956 Chr.16: 1350292-1350292 on Build GRCh38
rs901259262 Chr.16: 1350427-1350427 on Build GRCh38
rs770547806 Chr.16: 1350058-1350058 on Build GRCh38
rs773465601 Chr.16: 1350129-1350129 on Build GRCh38
rs906287978 Chr.16: 1350027-1350027 on Build GRCh38
rs148141495 Chr.16: 1350065-1350065 on Build GRCh38
rs139006693 Chr.16: 1350157-1350157 on Build GRCh38
rs917702750 Chr.16: 1350133-1350133 on Build GRCh38
rs755228580 Chr.16: 1350123-1350123 on Build GRCh38
rs753065771 Chr.16: 1350245-1350245 on Build GRCh38
rs926335610 Chr.16: 1350317-1350317 on Build GRCh38
rs201725773 Chr.16: 1350110-1350110 on Build GRCh38
rs748555427 Chr.16: 1350127-1350127 on Build GRCh38
rs781167188 Chr.16: 1350258-1350258 on Build GRCh38
rs760063483 Chr.16: 1350062-1350064 on Build GRCh38
rs747202730 Chr.16: 1350345-1350345 on Build GRCh38
rs548655266 Chr.16: 1350355-1350355 on Build GRCh38
rs755841038 Chr.16: 1350164-1350164 on Build GRCh38
rs984070457 Chr.16: 1350337-1350337 on Build GRCh38
rs200565292 Chr.16: 1350204-1350204 on Build GRCh38
rs1043954306 Chr.16: 1350368-1350368 on Build GRCh38
rs527945921 Chr.16: 1350272-1350272 on Build GRCh38
rs983109658 Chr.16: 1350308-1350308 on Build GRCh38
rs995829433 Chr.16: 1350019-1350019 on Build GRCh38
rs777432530 Chr.16: 1350046-1350046 on Build GRCh38
rs758781838 Chr.16: 1350247-1350247 on Build GRCh38
rs1054953125 Chr.16: 1350176-1350176 on Build GRCh38
rs201696319 Chr.16: 1350170-1350170 on Build GRCh38
rs752260875 Chr.16: 1350082-1350100 on Build GRCh38
rs780808158 Chr.16: 1350121-1350121 on Build GRCh38
rs939861515 Chr.16: 1350387-1350387 on Build GRCh38
rs1043621709 Chr.16: 1350201-1350202 on Build GRCh38
rs199682343 Chr.16: 1350260-1350260 on Build GRCh38
rs767159335 Chr.16: 1350146-1350146 on Build GRCh38
rs771340303 Chr.16: 1350136-1350136 on Build GRCh38
rs375400644 Chr.16: 1350144-1350144 on Build GRCh38
rs746614922 Chr.16: 1350052-1350052 on Build GRCh38
rs1017661178 Chr.16: 1350251-1350251 on Build GRCh38
rs373399857 Chr.16: 1350273-1350273 on Build GRCh38
rs760581168 Chr.16: 1350016-1350016 on Build GRCh38
rs766051684 Chr.16: 1350241-1350241 on Build GRCh38
rs756697549 Chr.16: 1350145-1350145 on Build GRCh38
rs1008865739 Chr.16: 1350430-1350430 on Build GRCh38
rs111267196 Chr.16: 1350227-1350227 on Build GRCh38
rs996888230 Chr.16: 1350229-1350229 on Build GRCh38
rs367737359 Chr.16: 1350059-1350059 on Build GRCh38
rs72779216 Chr.16: 1350282-1350282 on Build GRCh38
rs759381551 Chr.16: 1350137-1350137 on Build GRCh38
rs117781264 Chr.16: 1350239-1350239 on Build GRCh38
rs777594223 Chr.16: 1350241-1350242 on Build GRCh38
rs772664816 Chr.16: 1350286-1350286 on Build GRCh38
rs142109530 Chr.16: 1350111-1350111 on Build GRCh38
rs372514570 Chr.16: 1350240-1350240 on Build GRCh38
rs935272772 Chr.16: 1350385-1350385 on Build GRCh38
rs377633003 Chr.16: 1349999-1349999 on Build GRCh38
rs753504103 Chr.16: 1350264-1350264 on Build GRCh38
rs535217640 Chr.16: 1350307-1350307 on Build GRCh38
rs768593176 Chr.16: 1350226-1350226 on Build GRCh38
rs1054859055 Chr.16: 1350322-1350322 on Build GRCh38
rs908351086 Chr.16: 1350375-1350375 on Build GRCh38
rs980834982 Chr.16: 1350315-1350315 on Build GRCh38
rs773592112 Chr.16: 1350077-1350077 on Build GRCh38
rs766687285 Chr.16: 1350000-1350001 on Build GRCh38
rs755525157 Chr.16: 1350106-1350107 on Build GRCh38
rs954796172 Chr.16: 1350100-1350100 on Build GRCh38
rs754984027 Chr.16: 1350020-1350020 on Build GRCh38
rs370888956 Chr.16: 1350014-1350014 on Build GRCh38
rs372183764 Chr.16: 1350131-1350131 on Build GRCh38
rs776639568 Chr.16: 1350090-1350090 on Build GRCh38
rs781497708 Chr.16: 1350203-1350203 on Build GRCh38
rs778431583 Chr.16: 1350253-1350253 on Build GRCh38
rs762833838 Chr.16: 1350141-1350141 on Build GRCh38
rs113774682 Chr.16: 1350107-1350107 on Build GRCh38
rs965418486 Chr.16: 1350103-1350103 on Build GRCh38
rs72779215 Chr.16: 1350018-1350018 on Build GRCh38
rs555149094 Chr.16: 1350087-1350087 on Build GRCh38
rs764014118 Chr.16: 1350143-1350143 on Build GRCh38
rs765236752 Chr.16: 1350091-1350091 on Build GRCh38
rs1040896874 Chr.16: 1350414-1350414 on Build GRCh38
rs748266154 Chr.16: 1350064-1350064 on Build GRCh38
rs757813762 Chr.16: 1350256-1350256 on Build GRCh38
rs140020940 Chr.16: 1350223-1350223 on Build GRCh38
rs754293003 Chr.16: 1350215-1350215 on Build GRCh38
rs377651251 Chr.16: 1350266-1350266 on Build GRCh38
rs568745282 Chr.16: 1350408-1350408 on Build GRCh38
rs375137026 Chr.16: 1350194-1350194 on Build GRCh38
rs774660643 Chr.16: 1350062-1350062 on Build GRCh38
rs778712535 Chr.16: 1350028-1350028 on Build GRCh38
rs146399843 Chr.16: 1350431-1350431 on Build GRCh38
rs752311981 Chr.16: 1350033-1350033 on Build GRCh38
rs1050173574 Chr.16: 1350433-1350433 on Build GRCh38
rs904541789 Chr.16: 1350441-1350441 on Build GRCh38
rs551816175 Chr.16: 1350259-1350259 on Build GRCh38
rs749675012 Chr.16: 1350122-1350122 on Build GRCh38
rs983729980 Chr.16: 1350448-1350448 on Build GRCh38
rs766990986 Chr.16: 1350061-1350062 on Build GRCh38
rs747102484 Chr.16: 1350189-1350189 on Build GRCh38
rs750609781 Chr.16: 1350015-1350015 on Build GRCh38
rs770023401 Chr.16: 1350205-1350205 on Build GRCh38
rs946537543 Chr.16: 1350357-1350357 on Build GRCh38

View Full Product Details