Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1827408-1827944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAHD1
Entrez Gene ID
81889
Gene Name
fumarylacetoacetate hydrolase domain containing 1
Gene Aliases
C16orf36, YISKL
UniGene
Hs.513265
Gene Chromosome Location
Chr.16: 1827224-1840207 on Build GRCh38
Transcripts
NM_001018104.2 NM_001142398.1 NM_031208.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4169015 c.481G>T p.E161* Chr.16: 1827710-1827710 on Build GRCh38
4682304 c.426C>T p.D142D Chr.16: 1827655-1827655 on Build GRCh38
4128752 c.388C>A p.P130T Chr.16: 1827617-1827617 on Build GRCh38
1189136 c.616G>C p.E206Q Chr.16: 1827845-1827845 on Build GRCh38
4419557 c.468C>A p.L156L Chr.16: 1827697-1827697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766424745 Chr.16: 1827677-1827677 on Build GRCh38
rs370268598 Chr.16: 1827545-1827545 on Build GRCh38
rs746937126 Chr.16: 1827784-1827784 on Build GRCh38
rs377324379 Chr.16: 1827695-1827695 on Build GRCh38
rs201750977 Chr.16: 1827549-1827549 on Build GRCh38
rs140989113 Chr.16: 1827739-1827739 on Build GRCh38
rs759771469 Chr.16: 1827863-1827863 on Build GRCh38
rs752659525 Chr.16: 1827591-1827591 on Build GRCh38
rs577168355 Chr.16: 1827705-1827705 on Build GRCh38
rs148559699 Chr.16: 1827681-1827681 on Build GRCh38
rs757341156 Chr.16: 1827623-1827624 on Build GRCh38
rs201234911 Chr.16: 1827439-1827439 on Build GRCh38
rs774363462 Chr.16: 1827631-1827631 on Build GRCh38
rs774951188 Chr.16: 1827802-1827802 on Build GRCh38
rs150008635 Chr.16: 1827903-1827903 on Build GRCh38
rs1010113397 Chr.16: 1827628-1827628 on Build GRCh38
rs979125330 Chr.16: 1827502-1827502 on Build GRCh38
rs369979388 Chr.16: 1827792-1827792 on Build GRCh38
rs771241120 Chr.16: 1827507-1827507 on Build GRCh38
rs751388046 Chr.16: 1827448-1827448 on Build GRCh38
rs750445720 Chr.16: 1827699-1827700 on Build GRCh38
rs3743853 Chr.16: 1827557-1827557 on Build GRCh38
rs746275903 Chr.16: 1827612-1827612 on Build GRCh38
rs886448634 Chr.16: 1827548-1827548 on Build GRCh38
rs148244001 Chr.16: 1827436-1827436 on Build GRCh38
rs769862861 Chr.16: 1827525-1827525 on Build GRCh38
rs781520734 Chr.16: 1827606-1827606 on Build GRCh38
rs752619417 Chr.16: 1827440-1827440 on Build GRCh38
rs141622344 Chr.16: 1827617-1827617 on Build GRCh38
rs778963849 Chr.16: 1827771-1827771 on Build GRCh38
rs755198562 Chr.16: 1827767-1827767 on Build GRCh38
rs145930740 Chr.16: 1827575-1827575 on Build GRCh38
rs576596560 Chr.16: 1827476-1827476 on Build GRCh38
rs770782031 Chr.16: 1827741-1827741 on Build GRCh38
rs764254165 Chr.16: 1827882-1827882 on Build GRCh38
rs919812912 Chr.16: 1827777-1827777 on Build GRCh38
rs776355120 Chr.16: 1827742-1827742 on Build GRCh38
rs773125108 Chr.16: 1827609-1827609 on Build GRCh38
rs765297181 Chr.16: 1827637-1827638 on Build GRCh38
rs923600441 Chr.16: 1827511-1827511 on Build GRCh38
rs749352792 Chr.16: 1827572-1827572 on Build GRCh38
rs144517611 Chr.16: 1827619-1827619 on Build GRCh38
rs759211556 Chr.16: 1827744-1827744 on Build GRCh38
rs768153149 Chr.16: 1827823-1827823 on Build GRCh38
rs757184553 Chr.16: 1827892-1827892 on Build GRCh38
rs896488658 Chr.16: 1827852-1827852 on Build GRCh38
rs752922681 Chr.16: 1827772-1827772 on Build GRCh38
rs3743854 Chr.16: 1827493-1827493 on Build GRCh38
rs763611579 Chr.16: 1827751-1827751 on Build GRCh38
rs777226982 Chr.16: 1827861-1827861 on Build GRCh38
rs772186508 Chr.16: 1827652-1827652 on Build GRCh38
rs374040401 Chr.16: 1827708-1827708 on Build GRCh38
rs377760925 Chr.16: 1827600-1827600 on Build GRCh38
rs367867345 Chr.16: 1827738-1827738 on Build GRCh38
rs372337381 Chr.16: 1827707-1827707 on Build GRCh38
rs769658361 Chr.16: 1827795-1827795 on Build GRCh38
rs138540253 Chr.16: 1827585-1827585 on Build GRCh38
rs142676593 Chr.16: 1827559-1827559 on Build GRCh38
rs565822052 Chr.16: 1827877-1827877 on Build GRCh38
rs13166 Chr.16: 1827912-1827912 on Build GRCh38
rs779029732 Chr.16: 1827713-1827713 on Build GRCh38
rs369398052 Chr.16: 1827514-1827514 on Build GRCh38
rs765648079 Chr.16: 1827871-1827871 on Build GRCh38
rs150350443 Chr.16: 1827531-1827531 on Build GRCh38
rs762283022 Chr.16: 1827691-1827691 on Build GRCh38
rs759813779 Chr.16: 1827523-1827523 on Build GRCh38
rs770055326 Chr.16: 1827562-1827562 on Build GRCh38
rs753464371 Chr.16: 1827594-1827594 on Build GRCh38
rs778302216 Chr.16: 1827550-1827550 on Build GRCh38
rs752899746 Chr.16: 1827876-1827876 on Build GRCh38
rs763017079 Chr.16: 1827443-1827443 on Build GRCh38
rs761150370 Chr.16: 1827855-1827855 on Build GRCh38
rs191965297 Chr.16: 1827451-1827451 on Build GRCh38
rs773910707 Chr.16: 1827841-1827841 on Build GRCh38
rs767397644 Chr.16: 1827566-1827568 on Build GRCh38
rs755670267 Chr.16: 1827461-1827461 on Build GRCh38
rs780948061 Chr.16: 1827893-1827893 on Build GRCh38
rs772309233 Chr.16: 1827721-1827721 on Build GRCh38
rs372228511 Chr.16: 1827848-1827848 on Build GRCh38
rs182875865 Chr.16: 1827789-1827789 on Build GRCh38
rs940814012 Chr.16: 1827760-1827760 on Build GRCh38
rs912453163 Chr.16: 1827529-1827529 on Build GRCh38
rs768645041 Chr.16: 1827574-1827574 on Build GRCh38
rs142900137 Chr.16: 1827688-1827688 on Build GRCh38
rs747432454 Chr.16: 1827503-1827503 on Build GRCh38
rs777951334 Chr.16: 1827494-1827494 on Build GRCh38
rs776287940 Chr.16: 1827674-1827674 on Build GRCh38
rs781015874 Chr.16: 1827791-1827791 on Build GRCh38
rs369492494 Chr.16: 1827528-1827528 on Build GRCh38
rs984910988 Chr.16: 1827740-1827740 on Build GRCh38
rs147870961 Chr.16: 1827796-1827796 on Build GRCh38
rs747492216 Chr.16: 1827551-1827551 on Build GRCh38
rs746083064 Chr.16: 1827561-1827561 on Build GRCh38
rs374720354 Chr.16: 1827783-1827783 on Build GRCh38
rs375302754 Chr.16: 1827627-1827627 on Build GRCh38
rs765976075 Chr.16: 1827543-1827543 on Build GRCh38
rs754460625 Chr.16: 1827596-1827596 on Build GRCh38
rs752036004 Chr.16: 1827601-1827601 on Build GRCh38
rs776652666 Chr.16: 1827427-1827427 on Build GRCh38
rs200357596 Chr.16: 1827542-1827542 on Build GRCh38
rs761968538 Chr.16: 1827546-1827546 on Build GRCh38
rs748279685 Chr.16: 1827717-1827717 on Build GRCh38
rs979179314 Chr.16: 1827578-1827578 on Build GRCh38
rs748596830 Chr.16: 1827490-1827490 on Build GRCh38
rs144948758 Chr.16: 1827746-1827746 on Build GRCh38
rs770872053 Chr.16: 1827673-1827673 on Build GRCh38
rs756417278 Chr.16: 1827701-1827701 on Build GRCh38
rs766174715 Chr.16: 1827592-1827592 on Build GRCh38
rs760507332 Chr.16: 1827670-1827670 on Build GRCh38
rs749488492 Chr.16: 1827623-1827623 on Build GRCh38
rs150430426 Chr.16: 1827753-1827753 on Build GRCh38
rs759338353 Chr.16: 1827535-1827535 on Build GRCh38
rs754205563 Chr.16: 1827763-1827763 on Build GRCh38
rs200823071 Chr.16: 1827533-1827533 on Build GRCh38
rs768048752 Chr.16: 1827696-1827696 on Build GRCh38
rs750146072 Chr.16: 1827459-1827459 on Build GRCh38
rs757825291 Chr.16: 1827602-1827602 on Build GRCh38
rs751735330 Chr.16: 1827885-1827885 on Build GRCh38
rs1050488709 Chr.16: 1827785-1827785 on Build GRCh38
rs1056905733 Chr.16: 1827845-1827845 on Build GRCh38
rs369838638 Chr.16: 1827433-1827433 on Build GRCh38
rs760382793 Chr.16: 1827591-1827591 on Build GRCh38
rs187630331 Chr.16: 1827426-1827426 on Build GRCh38
rs866513855 Chr.16: 1827737-1827737 on Build GRCh38
rs3743852 Chr.16: 1827697-1827697 on Build GRCh38
rs958523576 Chr.16: 1827736-1827736 on Build GRCh38
rs376770299 Chr.16: 1827452-1827452 on Build GRCh38
rs961171649 Chr.16: 1827435-1827435 on Build GRCh38
rs745835853 Chr.16: 1827899-1827899 on Build GRCh38
rs141586613 Chr.16: 1827822-1827822 on Build GRCh38
rs886499438 Chr.16: 1827665-1827665 on Build GRCh38
rs981176936 Chr.16: 1827487-1827487 on Build GRCh38
rs766836010 Chr.16: 1827859-1827859 on Build GRCh38
rs940778463 Chr.16: 1827833-1827833 on Build GRCh38
rs779894104 Chr.16: 1827905-1827905 on Build GRCh38
rs748104752 Chr.16: 1827637-1827637 on Build GRCh38
rs752991519 Chr.16: 1827780-1827780 on Build GRCh38
rs759635482 Chr.16: 1827766-1827766 on Build GRCh38
rs766755458 Chr.16: 1827755-1827755 on Build GRCh38
rs759113509 Chr.16: 1827675-1827675 on Build GRCh38
rs773201120 Chr.16: 1827588-1827588 on Build GRCh38
rs933540553 Chr.16: 1827782-1827782 on Build GRCh38
rs533518287 Chr.16: 1827750-1827750 on Build GRCh38

View Full Product Details