Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1837811-1838349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAHD1
Entrez Gene ID
81889
Gene Name
fumarylacetoacetate hydrolase domain containing 1
Gene Aliases
C16orf36, YISKL
UniGene
Hs.513265
Gene Chromosome Location
Chr.16: 1827224-1840207 on Build GRCh38
Transcripts
NM_001018104.2 NM_001142398.1 NM_001163560.2 NM_152764.2

Gene Symbol
MEIOB
Entrez Gene ID
254528
Gene Name
meiosis specific with OB domains
Gene Aliases
C16orf73, gs129
UniGene
Hs.729604
Gene Chromosome Location
Chr.16: 1833983-1872178 on Build GRCh38
Transcripts
NM_001018104.2 NM_001142398.1 NM_001163560.2 NM_152764.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5516721 c.1219-7delT p.? Chr.16: 1837877-1837877 on Build GRCh38
3396223 c.662A>G p.K221R Chr.16: 1838041-1838041 on Build GRCh38
4506020 c.708C>T p.L236L Chr.16: 1838087-1838087 on Build GRCh38
6197435 c.657G>A p.S219S Chr.16: 1838036-1838036 on Build GRCh38
7255644 c.712G>C p.G238R Chr.16: 1838091-1838091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756466372 Chr.16: 1838088-1838088 on Build GRCh38
rs908265672 Chr.16: 1838008-1838008 on Build GRCh38
rs67309640 Chr.16: 1838176-1838176 on Build GRCh38
rs759300021 Chr.16: 1837877-1837877 on Build GRCh38
rs1012180974 Chr.16: 1838217-1838217 on Build GRCh38
rs926206475 Chr.16: 1838229-1838229 on Build GRCh38
rs532086106 Chr.16: 1838065-1838065 on Build GRCh38
rs1033371445 Chr.16: 1838119-1838119 on Build GRCh38
rs781670886 Chr.16: 1838077-1838077 on Build GRCh38
rs952510165 Chr.16: 1838125-1838125 on Build GRCh38
rs542264304 Chr.16: 1838214-1838214 on Build GRCh38
rs976889271 Chr.16: 1838184-1838184 on Build GRCh38
rs780340817 Chr.16: 1838128-1838128 on Build GRCh38
rs147632019 Chr.16: 1838210-1838210 on Build GRCh38
rs922739070 Chr.16: 1838191-1838191 on Build GRCh38
rs930732785 Chr.16: 1837916-1837916 on Build GRCh38
rs185788889 Chr.16: 1838306-1838306 on Build GRCh38
rs777123699 Chr.16: 1837998-1837998 on Build GRCh38
rs542509711 Chr.16: 1837885-1837885 on Build GRCh38
rs73490142 Chr.16: 1837993-1837993 on Build GRCh38
rs970642903 Chr.16: 1838096-1838096 on Build GRCh38
rs922894271 Chr.16: 1838029-1838029 on Build GRCh38
rs552182711 Chr.16: 1838081-1838081 on Build GRCh38
rs1004759569 Chr.16: 1837972-1837972 on Build GRCh38
rs763969224 Chr.16: 1838090-1838090 on Build GRCh38
rs765011693 Chr.16: 1837887-1837887 on Build GRCh38
rs1053123382 Chr.16: 1838287-1838287 on Build GRCh38
rs1009444008 Chr.16: 1838080-1838080 on Build GRCh38
rs62038406 Chr.16: 1838307-1838307 on Build GRCh38
rs749823022 Chr.16: 1838056-1838056 on Build GRCh38
rs891889313 Chr.16: 1838301-1838301 on Build GRCh38
rs769142272 Chr.16: 1838062-1838062 on Build GRCh38
rs75334169 Chr.16: 1837958-1837958 on Build GRCh38
rs926249420 Chr.16: 1837985-1837985 on Build GRCh38
rs201214840 Chr.16: 1837886-1837887 on Build GRCh38
rs763631463 Chr.16: 1838022-1838022 on Build GRCh38
rs912115051 Chr.16: 1838101-1838101 on Build GRCh38
rs956399534 Chr.16: 1838090-1838090 on Build GRCh38
rs758734618 Chr.16: 1837887-1837888 on Build GRCh38
rs921941878 Chr.16: 1838142-1838142 on Build GRCh38
rs904493446 Chr.16: 1838041-1838041 on Build GRCh38
rs1049175935 Chr.16: 1838160-1838160 on Build GRCh38
rs908331786 Chr.16: 1838159-1838159 on Build GRCh38
rs926138084 Chr.16: 1837874-1837874 on Build GRCh38
rs760674658 Chr.16: 1837893-1837894 on Build GRCh38
rs1053642441 Chr.16: 1838075-1838075 on Build GRCh38
rs1024931036 Chr.16: 1837890-1837890 on Build GRCh38
rs141132983 Chr.16: 1837887-1837887 on Build GRCh38
rs900568344 Chr.16: 1837886-1837886 on Build GRCh38
rs528391738 Chr.16: 1838089-1838089 on Build GRCh38
rs774117210 Chr.16: 1837876-1837877 on Build GRCh38
rs746567623 Chr.16: 1838004-1838004 on Build GRCh38
rs142700439 Chr.16: 1838118-1838118 on Build GRCh38
rs62038405 Chr.16: 1838263-1838263 on Build GRCh38
rs940218784 Chr.16: 1838019-1838019 on Build GRCh38
rs74891344 Chr.16: 1838036-1838036 on Build GRCh38
rs556504006 Chr.16: 1838082-1838082 on Build GRCh38
rs989026693 Chr.16: 1838093-1838093 on Build GRCh38
rs199775735 Chr.16: 1837846-1837846 on Build GRCh38
rs573518333 Chr.16: 1837840-1837840 on Build GRCh38
rs774141768 Chr.16: 1838212-1838212 on Build GRCh38
rs66509847 Chr.16: 1837936-1837936 on Build GRCh38
rs751059643 Chr.16: 1838032-1838032 on Build GRCh38
rs769235284 Chr.16: 1837963-1837964 on Build GRCh38
rs934882655 Chr.16: 1837876-1837876 on Build GRCh38

View Full Product Details