Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1839557-1840054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAHD1
Entrez Gene ID
81889
Gene Name
fumarylacetoacetate hydrolase domain containing 1
Gene Aliases
C16orf36, YISKL
UniGene
Hs.513265
Gene Chromosome Location
Chr.16: 1827224-1840207 on Build GRCh38
Transcripts
NM_001018104.2 NM_001142398.1 NM_001163560.2 NM_152764.2

Gene Symbol
MEIOB
Entrez Gene ID
254528
Gene Name
meiosis specific with OB domains
Gene Aliases
C16orf73, gs129
UniGene
Hs.729604
Gene Chromosome Location
Chr.16: 1833983-1872178 on Build GRCh38
Transcripts
NM_001018104.2 NM_001142398.1 NM_001163560.2 NM_152764.2

SNP Detail

SNP ID Chromosome Location
rs181171688 Chr.16: 1839741-1839741 on Build GRCh38
rs972100149 Chr.16: 1839694-1839694 on Build GRCh38
rs150321971 Chr.16: 1839739-1839739 on Build GRCh38
rs754164242 Chr.16: 1839764-1839764 on Build GRCh38
rs993447669 Chr.16: 1839946-1839946 on Build GRCh38
rs560353775 Chr.16: 1839920-1839920 on Build GRCh38
rs938897002 Chr.16: 1839870-1839870 on Build GRCh38
rs989838739 Chr.16: 1839816-1839816 on Build GRCh38
rs907344900 Chr.16: 1839585-1839585 on Build GRCh38
rs1006260780 Chr.16: 1839646-1839646 on Build GRCh38
rs758736751 Chr.16: 1839923-1839923 on Build GRCh38
rs942575349 Chr.16: 1839787-1839787 on Build GRCh38
rs3813759 Chr.16: 1839615-1839615 on Build GRCh38
rs569282812 Chr.16: 1840005-1840005 on Build GRCh38
rs942201742 Chr.16: 1839613-1839613 on Build GRCh38
rs868398497 Chr.16: 1839945-1839945 on Build GRCh38
rs3848347 Chr.16: 1839906-1839906 on Build GRCh38
rs1014265425 Chr.16: 1839649-1839649 on Build GRCh38
rs3813760 Chr.16: 1839644-1839644 on Build GRCh38
rs945795769 Chr.16: 1839840-1839840 on Build GRCh38
rs1016644922 Chr.16: 1839662-1839662 on Build GRCh38
rs529426854 Chr.16: 1839924-1839924 on Build GRCh38
rs3813761 Chr.16: 1839713-1839713 on Build GRCh38
rs140234821 Chr.16: 1839738-1839738 on Build GRCh38
rs1041510458 Chr.16: 1839849-1839849 on Build GRCh38
rs549451638 Chr.16: 1839961-1839961 on Build GRCh38
rs1048172024 Chr.16: 1839583-1839583 on Build GRCh38
rs748816242 Chr.16: 1839587-1839588 on Build GRCh38
rs962115079 Chr.16: 1839666-1839666 on Build GRCh38
rs1003023013 Chr.16: 1839602-1839602 on Build GRCh38
rs939781159 Chr.16: 1840027-1840027 on Build GRCh38
rs1030409376 Chr.16: 1839950-1839950 on Build GRCh38
rs3848346 Chr.16: 1839820-1839820 on Build GRCh38
rs906937612 Chr.16: 1839868-1839868 on Build GRCh38
rs755295778 Chr.16: 1839774-1839774 on Build GRCh38
rs894734687 Chr.16: 1839919-1839919 on Build GRCh38
rs958391377 Chr.16: 1839768-1839768 on Build GRCh38
rs534171954 Chr.16: 1839693-1839693 on Build GRCh38
rs138279147 Chr.16: 1840022-1840022 on Build GRCh38
rs572638224 Chr.16: 1839716-1839716 on Build GRCh38
rs546507588 Chr.16: 1839917-1839917 on Build GRCh38
rs186419402 Chr.16: 1839786-1839786 on Build GRCh38

View Full Product Details