Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3493405-3493898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf90
Entrez Gene ID
646174
Gene Name
chromosome 16 open reading frame 90
Gene Aliases
-
UniGene
Hs.632224
Gene Chromosome Location
Chr.16: 3493484-3495521 on Build GRCh38
Transcripts
NM_001080524.1 XM_005255502.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1609310 c.567G>C p.E189D Chr.16: 3493846-3493846 on Build GRCh38
5971790 c.524G>A p.R175H Chr.16: 3493864-3493864 on Build GRCh38
5527571 c.538G>A p.E180K Chr.16: 3493850-3493850 on Build GRCh38
970457 c.543G>T p.K181N Chr.16: 3493870-3493870 on Build GRCh38
1588457 c.518G>T p.R173I Chr.16: 3493870-3493870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044333265 Chr.16: 3493483-3493483 on Build GRCh38
rs988755450 Chr.16: 3493738-3493738 on Build GRCh38
rs576452497 Chr.16: 3493767-3493767 on Build GRCh38
rs935363628 Chr.16: 3493458-3493458 on Build GRCh38
rs921243080 Chr.16: 3493771-3493771 on Build GRCh38
rs751226976 Chr.16: 3493867-3493867 on Build GRCh38
rs545548538 Chr.16: 3493443-3493443 on Build GRCh38
rs1008512434 Chr.16: 3493546-3493546 on Build GRCh38
rs756364790 Chr.16: 3493632-3493632 on Build GRCh38
rs771979630 Chr.16: 3493759-3493759 on Build GRCh38
rs781040809 Chr.16: 3493795-3493795 on Build GRCh38
rs550178847 Chr.16: 3493735-3493735 on Build GRCh38
rs199544224 Chr.16: 3493799-3493799 on Build GRCh38
rs1050424565 Chr.16: 3493764-3493764 on Build GRCh38
rs956071089 Chr.16: 3493605-3493605 on Build GRCh38
rs1000452259 Chr.16: 3493770-3493770 on Build GRCh38
rs573144588 Chr.16: 3493634-3493634 on Build GRCh38
rs761254338 Chr.16: 3493841-3493841 on Build GRCh38
rs764746989 Chr.16: 3493848-3493848 on Build GRCh38
rs371884531 Chr.16: 3493550-3493551 on Build GRCh38
rs904877733 Chr.16: 3493724-3493724 on Build GRCh38
rs116855661 Chr.16: 3493660-3493660 on Build GRCh38
rs994948664 Chr.16: 3493548-3493549 on Build GRCh38
rs920549998 Chr.16: 3493659-3493659 on Build GRCh38
rs532131459 Chr.16: 3493549-3493549 on Build GRCh38
rs536228833 Chr.16: 3493713-3493713 on Build GRCh38
rs759479731 Chr.16: 3493831-3493831 on Build GRCh38
rs9932807 Chr.16: 3493445-3493445 on Build GRCh38
rs74005822 Chr.16: 3493527-3493527 on Build GRCh38
rs898943274 Chr.16: 3493711-3493711 on Build GRCh38
rs1042962142 Chr.16: 3493493-3493493 on Build GRCh38
rs185591789 Chr.16: 3493576-3493576 on Build GRCh38
rs190822077 Chr.16: 3493822-3493822 on Build GRCh38
rs940550651 Chr.16: 3493677-3493677 on Build GRCh38
rs1047592422 Chr.16: 3493699-3493699 on Build GRCh38
rs1004558058 Chr.16: 3493542-3493542 on Build GRCh38
rs969280797 Chr.16: 3493559-3493559 on Build GRCh38
rs944423523 Chr.16: 3493716-3493716 on Build GRCh38
rs9923182 Chr.16: 3493820-3493820 on Build GRCh38
rs551507734 Chr.16: 3493526-3493526 on Build GRCh38
rs780666914 Chr.16: 3493876-3493876 on Build GRCh38
rs886222739 Chr.16: 3493538-3493538 on Build GRCh38
rs1031759305 Chr.16: 3493577-3493577 on Build GRCh38
rs754224523 Chr.16: 3493850-3493850 on Build GRCh38
rs749260760 Chr.16: 3493811-3493812 on Build GRCh38
rs371199813 Chr.16: 3493864-3493864 on Build GRCh38
rs527942155 Chr.16: 3493461-3493461 on Build GRCh38
rs1015114558 Chr.16: 3493543-3493543 on Build GRCh38
rs754652937 Chr.16: 3493869-3493869 on Build GRCh38
rs371165617 Chr.16: 3493804-3493804 on Build GRCh38
rs774646659 Chr.16: 3493740-3493740 on Build GRCh38
rs745967379 Chr.16: 3493628-3493628 on Build GRCh38
rs140776833 Chr.16: 3493554-3493554 on Build GRCh38
rs774653873 Chr.16: 3493824-3493824 on Build GRCh38
rs748921643 Chr.16: 3493629-3493629 on Build GRCh38
rs934020280 Chr.16: 3493772-3493772 on Build GRCh38
rs750995800 Chr.16: 3493791-3493791 on Build GRCh38
rs547565496 Chr.16: 3493555-3493555 on Build GRCh38
rs569937045 Chr.16: 3493794-3493794 on Build GRCh38
rs201461202 Chr.16: 3493832-3493832 on Build GRCh38
rs956841731 Chr.16: 3493737-3493737 on Build GRCh38
rs1023683517 Chr.16: 3493719-3493719 on Build GRCh38
rs992920769 Chr.16: 3493627-3493627 on Build GRCh38
rs775783720 Chr.16: 3493834-3493834 on Build GRCh38
rs758306474 Chr.16: 3493506-3493506 on Build GRCh38
rs778838146 Chr.16: 3493673-3493673 on Build GRCh38
rs907715039 Chr.16: 3493715-3493715 on Build GRCh38
rs890292407 Chr.16: 3493535-3493535 on Build GRCh38
rs200770058 Chr.16: 3493865-3493865 on Build GRCh38
rs552303823 Chr.16: 3493551-3493551 on Build GRCh38
rs928123423 Chr.16: 3493458-3493461 on Build GRCh38
rs565178135 Chr.16: 3493664-3493666 on Build GRCh38
rs548119016 Chr.16: 3493491-3493491 on Build GRCh38
rs948610452 Chr.16: 3493482-3493482 on Build GRCh38
rs545174053 Chr.16: 3493781-3493781 on Build GRCh38
rs561615840 Chr.16: 3493519-3493519 on Build GRCh38
rs373573539 Chr.16: 3493502-3493502 on Build GRCh38
rs75072041 Chr.16: 3493734-3493734 on Build GRCh38
rs527477171 Chr.16: 3493548-3493548 on Build GRCh38

View Full Product Details