Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3726218-3726723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBBP
Entrez Gene ID
1387
Gene Name
CREB binding protein
Gene Aliases
CBP, KAT3A, RSTS
UniGene
Hs.459759
Gene Chromosome Location
Chr.16: 3725054-3880727 on Build GRCh38
Transcripts
NM_001079846.1 NM_004380.2 XM_005255124.4 XM_005255125.4 XM_006720848.3 XM_011522381.2 XM_017022944.1

SNP Detail

SNP ID Chromosome Location
rs144692279 Chr.16: 3726333-3726333 on Build GRCh38
rs756547297 Chr.16: 3726596-3726596 on Build GRCh38
rs572303083 Chr.16: 3726547-3726547 on Build GRCh38
rs113302498 Chr.16: 3726427-3726427 on Build GRCh38
rs894598464 Chr.16: 3726426-3726426 on Build GRCh38
rs1057133031 Chr.16: 3726405-3726405 on Build GRCh38
rs531708637 Chr.16: 3726300-3726300 on Build GRCh38
rs912342904 Chr.16: 3726332-3726332 on Build GRCh38
rs888779065 Chr.16: 3726628-3726628 on Build GRCh38
rs571576565 Chr.16: 3726240-3726240 on Build GRCh38
rs975208548 Chr.16: 3726498-3726498 on Build GRCh38
rs773950779 Chr.16: 3726546-3726546 on Build GRCh38
rs890739628 Chr.16: 3726598-3726598 on Build GRCh38
rs567146441 Chr.16: 3726447-3726447 on Build GRCh38
rs897583090 Chr.16: 3726683-3726683 on Build GRCh38
rs879026614 Chr.16: 3726239-3726240 on Build GRCh38
rs772893885 Chr.16: 3726355-3726358 on Build GRCh38
rs1037811228 Chr.16: 3726537-3726537 on Build GRCh38
rs1041719694 Chr.16: 3726270-3726270 on Build GRCh38
rs1039628447 Chr.16: 3726248-3726248 on Build GRCh38
rs567290539 Chr.16: 3726243-3726243 on Build GRCh38
rs1635399 Chr.16: 3726412-3726412 on Build GRCh38
rs1020990661 Chr.16: 3726249-3726249 on Build GRCh38
rs960332016 Chr.16: 3726242-3726243 on Build GRCh38
rs975281024 Chr.16: 3726350-3726350 on Build GRCh38
rs1008980597 Chr.16: 3726636-3726636 on Build GRCh38
rs1007856937 Chr.16: 3726634-3726634 on Build GRCh38
rs3179352 Chr.16: 3726306-3726306 on Build GRCh38
rs535953791 Chr.16: 3726244-3726244 on Build GRCh38
rs1007930662 Chr.16: 3726463-3726463 on Build GRCh38
rs560270968 Chr.16: 3726274-3726274 on Build GRCh38
rs993320256 Chr.16: 3726684-3726684 on Build GRCh38
rs993781761 Chr.16: 3726453-3726453 on Build GRCh38
rs962341717 Chr.16: 3726241-3726241 on Build GRCh38
rs972315788 Chr.16: 3726462-3726462 on Build GRCh38
rs574027491 Chr.16: 3726276-3726276 on Build GRCh38
rs1019297532 Chr.16: 3726652-3726652 on Build GRCh38
rs766652133 Chr.16: 3726428-3726440 on Build GRCh38
rs538759549 Chr.16: 3726519-3726519 on Build GRCh38
rs924554715 Chr.16: 3726516-3726516 on Build GRCh38
rs951910424 Chr.16: 3726686-3726686 on Build GRCh38
rs1019369576 Chr.16: 3726466-3726466 on Build GRCh38
rs1040088386 Chr.16: 3726676-3726676 on Build GRCh38
rs948815023 Chr.16: 3726531-3726531 on Build GRCh38
rs914451578 Chr.16: 3726351-3726351 on Build GRCh38
rs34148810 Chr.16: 3726628-3726629 on Build GRCh38
rs547057736 Chr.16: 3726242-3726242 on Build GRCh38
rs375123238 Chr.16: 3726422-3726422 on Build GRCh38
rs1018765427 Chr.16: 3726285-3726285 on Build GRCh38
rs946246645 Chr.16: 3726359-3726359 on Build GRCh38
rs879039927 Chr.16: 3726497-3726497 on Build GRCh38
rs986530338 Chr.16: 3726326-3726326 on Build GRCh38
rs549516833 Chr.16: 3726504-3726504 on Build GRCh38
rs113182995 Chr.16: 3726413-3726425 on Build GRCh38
rs369136324 Chr.16: 3726245-3726245 on Build GRCh38
rs867727493 Chr.16: 3726297-3726297 on Build GRCh38
rs572342292 Chr.16: 3726254-3726254 on Build GRCh38
rs34630159 Chr.16: 3726427-3726439 on Build GRCh38
rs867579617 Chr.16: 3726439-3726439 on Build GRCh38
rs951067233 Chr.16: 3726459-3726459 on Build GRCh38
rs542660820 Chr.16: 3726290-3726290 on Build GRCh38
rs938708024 Chr.16: 3726389-3726389 on Build GRCh38
rs80164886 Chr.16: 3726425-3726425 on Build GRCh38
rs936223792 Chr.16: 3726580-3726580 on Build GRCh38
rs899312186 Chr.16: 3726538-3726538 on Build GRCh38
rs558763100 Chr.16: 3726522-3726522 on Build GRCh38
rs1028683030 Chr.16: 3726388-3726388 on Build GRCh38
rs150542689 Chr.16: 3726245-3726246 on Build GRCh38
rs897169094 Chr.16: 3726240-3726240 on Build GRCh38
rs2286005 Chr.16: 3726246-3726246 on Build GRCh38
rs904782975 Chr.16: 3726377-3726377 on Build GRCh38
rs1028715420 Chr.16: 3726264-3726264 on Build GRCh38
rs924655701 Chr.16: 3726381-3726381 on Build GRCh38
rs927285049 Chr.16: 3726500-3726500 on Build GRCh38
rs904755288 Chr.16: 3726557-3726557 on Build GRCh38
rs1050697899 Chr.16: 3726589-3726589 on Build GRCh38
rs917326198 Chr.16: 3726536-3726536 on Build GRCh38
rs957363361 Chr.16: 3726491-3726502 on Build GRCh38
rs752807752 Chr.16: 3726428-3726439 on Build GRCh38
rs938690373 Chr.16: 3726520-3726520 on Build GRCh38
rs1027005459 Chr.16: 3726458-3726458 on Build GRCh38
rs541345097 Chr.16: 3726263-3726263 on Build GRCh38
rs565169280 Chr.16: 3726317-3726317 on Build GRCh38
rs899379726 Chr.16: 3726442-3726442 on Build GRCh38
rs963703226 Chr.16: 3726301-3726301 on Build GRCh38
rs386787936 Chr.16: 3726427-3726442 on Build GRCh38
rs548111383 Chr.16: 3726360-3726360 on Build GRCh38
rs536196290 Chr.16: 3726467-3726467 on Build GRCh38
rs903829824 Chr.16: 3726494-3726494 on Build GRCh38
rs370838201 Chr.16: 3726244-3726245 on Build GRCh38
rs903230076 Chr.16: 3726271-3726271 on Build GRCh38
rs376154310 Chr.16: 3726505-3726505 on Build GRCh38
rs558800596 Chr.16: 3726247-3726247 on Build GRCh38
rs977348577 Chr.16: 3726506-3726506 on Build GRCh38
rs1025321267 Chr.16: 3726307-3726307 on Build GRCh38
rs937408994 Chr.16: 3726322-3726322 on Build GRCh38
rs963760107 Chr.16: 3726685-3726685 on Build GRCh38
rs1042083799 Chr.16: 3726368-3726368 on Build GRCh38
rs534603318 Chr.16: 3726625-3726625 on Build GRCh38
rs149315725 Chr.16: 3726243-3726244 on Build GRCh38
rs78989292 Chr.16: 3726528-3726528 on Build GRCh38
rs562603203 Chr.16: 3726298-3726298 on Build GRCh38

View Full Product Details