Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4574894-4575388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf96
Entrez Gene ID
342346
Gene Name
chromosome 16 open reading frame 96
Gene Aliases
-
UniGene
Hs.684212
Gene Chromosome Location
Chr.16: 4538709-4600455 on Build GRCh38
Transcripts
NM_001145011.1 XM_005255298.3 XM_005255301.3 XM_011522475.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4961047 c.852G>A p.E284E Chr.16: 4575332-4575332 on Build GRCh38
6522548 c.658C>A p.L220I Chr.16: 4575023-4575023 on Build GRCh38
4806294 c.790G>T p.A264S Chr.16: 4575270-4575270 on Build GRCh38
4846395 c.690C>T p.T230T Chr.16: 4575055-4575055 on Build GRCh38
4511407 c.867C>T p.L289L Chr.16: 4575347-4575347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767954720 Chr.16: 4575128-4575128 on Build GRCh38
rs558135189 Chr.16: 4574930-4574930 on Build GRCh38
rs140680907 Chr.16: 4575350-4575350 on Build GRCh38
rs375733441 Chr.16: 4575291-4575291 on Build GRCh38
rs769192653 Chr.16: 4575190-4575190 on Build GRCh38
rs766128669 Chr.16: 4575019-4575019 on Build GRCh38
rs955733042 Chr.16: 4575183-4575183 on Build GRCh38
rs368217239 Chr.16: 4575105-4575105 on Build GRCh38
rs964289006 Chr.16: 4575212-4575212 on Build GRCh38
rs964235535 Chr.16: 4575003-4575003 on Build GRCh38
rs567810633 Chr.16: 4574955-4574955 on Build GRCh38
rs554799912 Chr.16: 4575162-4575162 on Build GRCh38
rs755620336 Chr.16: 4575173-4575173 on Build GRCh38
rs948520506 Chr.16: 4575299-4575299 on Build GRCh38
rs1019843177 Chr.16: 4575001-4575001 on Build GRCh38
rs907078563 Chr.16: 4575349-4575349 on Build GRCh38
rs138390932 Chr.16: 4574939-4574939 on Build GRCh38
rs951558791 Chr.16: 4574965-4574965 on Build GRCh38
rs186761749 Chr.16: 4575131-4575131 on Build GRCh38
rs552683971 Chr.16: 4575101-4575101 on Build GRCh38
rs376298041 Chr.16: 4574949-4574949 on Build GRCh38
rs764425471 Chr.16: 4575085-4575085 on Build GRCh38
rs1022561873 Chr.16: 4575217-4575217 on Build GRCh38
rs576640190 Chr.16: 4575247-4575247 on Build GRCh38
rs766876725 Chr.16: 4575284-4575284 on Build GRCh38
rs183130326 Chr.16: 4575363-4575363 on Build GRCh38
rs988386388 Chr.16: 4574935-4574935 on Build GRCh38
rs1052824125 Chr.16: 4575358-4575358 on Build GRCh38
rs771124202 Chr.16: 4574929-4574929 on Build GRCh38
rs376258492 Chr.16: 4575224-4575224 on Build GRCh38
rs9673241 Chr.16: 4575125-4575125 on Build GRCh38
rs921473823 Chr.16: 4575267-4575267 on Build GRCh38
rs1256768 Chr.16: 4575273-4575273 on Build GRCh38
rs9922488 Chr.16: 4575148-4575148 on Build GRCh38
rs777206248 Chr.16: 4575211-4575211 on Build GRCh38
rs113353601 Chr.16: 4574936-4574936 on Build GRCh38
rs76048912 Chr.16: 4575163-4575163 on Build GRCh38
rs572401766 Chr.16: 4575281-4575281 on Build GRCh38
rs753518030 Chr.16: 4575251-4575251 on Build GRCh38
rs975612390 Chr.16: 4575234-4575234 on Build GRCh38
rs1023103012 Chr.16: 4575106-4575106 on Build GRCh38
rs977527216 Chr.16: 4575017-4575017 on Build GRCh38
rs779135465 Chr.16: 4575367-4575367 on Build GRCh38
rs182165251 Chr.16: 4575041-4575041 on Build GRCh38
rs956949197 Chr.16: 4574918-4574918 on Build GRCh38
rs975850437 Chr.16: 4575010-4575010 on Build GRCh38
rs772007572 Chr.16: 4575142-4575142 on Build GRCh38
rs1026265919 Chr.16: 4574954-4574954 on Build GRCh38
rs898369187 Chr.16: 4575334-4575334 on Build GRCh38
rs935168016 Chr.16: 4575046-4575046 on Build GRCh38
rs558164554 Chr.16: 4575094-4575094 on Build GRCh38
rs549479446 Chr.16: 4575011-4575011 on Build GRCh38
rs775746017 Chr.16: 4574945-4574945 on Build GRCh38
rs1019727564 Chr.16: 4575195-4575195 on Build GRCh38
rs545505381 Chr.16: 4575268-4575268 on Build GRCh38
rs917753507 Chr.16: 4575290-4575290 on Build GRCh38
rs1056279790 Chr.16: 4575092-4575092 on Build GRCh38
rs779622953 Chr.16: 4575246-4575246 on Build GRCh38
rs995545436 Chr.16: 4575129-4575129 on Build GRCh38
rs62037460 Chr.16: 4574976-4574976 on Build GRCh38
rs940022778 Chr.16: 4575277-4575277 on Build GRCh38
rs191374978 Chr.16: 4575165-4575165 on Build GRCh38
rs921369497 Chr.16: 4574962-4574962 on Build GRCh38
rs931013430 Chr.16: 4575336-4575336 on Build GRCh38
rs771181497 Chr.16: 4575130-4575130 on Build GRCh38
rs752234400 Chr.16: 4575285-4575285 on Build GRCh38
rs975878059 Chr.16: 4574958-4574958 on Build GRCh38
rs147242097 Chr.16: 4575317-4575317 on Build GRCh38
rs556718262 Chr.16: 4575240-4575240 on Build GRCh38
rs751320483 Chr.16: 4575363-4575364 on Build GRCh38
rs751158306 Chr.16: 4575028-4575028 on Build GRCh38
rs112229339 Chr.16: 4575016-4575016 on Build GRCh38
rs780032758 Chr.16: 4575106-4575106 on Build GRCh38
rs886836446 Chr.16: 4575159-4575159 on Build GRCh38

View Full Product Details