Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4576445-4576913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf96
Entrez Gene ID
342346
Gene Name
chromosome 16 open reading frame 96
Gene Aliases
-
UniGene
Hs.684212
Gene Chromosome Location
Chr.16: 4538709-4600455 on Build GRCh38
Transcripts
NM_001145011.1 XM_005255298.3 XM_005255301.3 XM_011522475.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5982160 c.1965C>A p.A655A Chr.16: 4576445-4576445 on Build GRCh38
6021406 c.1987C>A p.L663M Chr.16: 4576467-4576467 on Build GRCh38
5473747 c.2092G>A p.D698N Chr.16: 4576572-4576572 on Build GRCh38
3691014 c.2073C>T p.A691A Chr.16: 4576553-4576553 on Build GRCh38
4621508 c.2067C>G p.H689Q Chr.16: 4576547-4576547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889725957 Chr.16: 4576479-4576479 on Build GRCh38
rs7206416 Chr.16: 4576723-4576723 on Build GRCh38
rs1008742274 Chr.16: 4576586-4576586 on Build GRCh38
rs535167070 Chr.16: 4576611-4576611 on Build GRCh38
rs546166791 Chr.16: 4576763-4576763 on Build GRCh38
rs780185982 Chr.16: 4576874-4576874 on Build GRCh38
rs746949687 Chr.16: 4576759-4576759 on Build GRCh38
rs542044443 Chr.16: 4576791-4576791 on Build GRCh38
rs574714408 Chr.16: 4576527-4576527 on Build GRCh38
rs114312438 Chr.16: 4576571-4576571 on Build GRCh38
rs765797259 Chr.16: 4576550-4576550 on Build GRCh38
rs541912775 Chr.16: 4576869-4576869 on Build GRCh38
rs952247577 Chr.16: 4576887-4576887 on Build GRCh38
rs530537472 Chr.16: 4576876-4576876 on Build GRCh38
rs1009921230 Chr.16: 4576826-4576826 on Build GRCh38
rs561727217 Chr.16: 4576870-4576870 on Build GRCh38
rs971871408 Chr.16: 4576621-4576621 on Build GRCh38
rs778874139 Chr.16: 4576477-4576477 on Build GRCh38
rs537821585 Chr.16: 4576825-4576829 on Build GRCh38
rs891696419 Chr.16: 4576814-4576814 on Build GRCh38
rs568120685 Chr.16: 4576670-4576670 on Build GRCh38
rs927559199 Chr.16: 4576713-4576713 on Build GRCh38
rs1000502351 Chr.16: 4576487-4576487 on Build GRCh38
rs935786984 Chr.16: 4576805-4576805 on Build GRCh38
rs374671207 Chr.16: 4576529-4576529 on Build GRCh38
rs554716380 Chr.16: 4576620-4576620 on Build GRCh38
rs1027718566 Chr.16: 4576626-4576626 on Build GRCh38
rs1024081690 Chr.16: 4576554-4576554 on Build GRCh38
rs373816170 Chr.16: 4576575-4576575 on Build GRCh38
rs1054378084 Chr.16: 4576808-4576808 on Build GRCh38
rs993632636 Chr.16: 4576866-4576866 on Build GRCh38
rs369792596 Chr.16: 4576578-4576578 on Build GRCh38
rs146459816 Chr.16: 4576790-4576790 on Build GRCh38
rs1000115649 Chr.16: 4576524-4576524 on Build GRCh38
rs1008856572 Chr.16: 4576818-4576818 on Build GRCh38
rs976026255 Chr.16: 4576786-4576786 on Build GRCh38
rs992955738 Chr.16: 4576730-4576730 on Build GRCh38
rs180929327 Chr.16: 4576803-4576803 on Build GRCh38
rs908092962 Chr.16: 4576733-4576733 on Build GRCh38
rs780725297 Chr.16: 4576588-4576588 on Build GRCh38
rs755828741 Chr.16: 4576593-4576593 on Build GRCh38
rs1023965919 Chr.16: 4576863-4576863 on Build GRCh38
rs750892765 Chr.16: 4576551-4576551 on Build GRCh38
rs758232639 Chr.16: 4576678-4576678 on Build GRCh38
rs897688700 Chr.16: 4576865-4576865 on Build GRCh38
rs7201176 Chr.16: 4576880-4576880 on Build GRCh38
rs866746093 Chr.16: 4576523-4576523 on Build GRCh38
rs577147105 Chr.16: 4576731-4576731 on Build GRCh38
rs971143641 Chr.16: 4576617-4576617 on Build GRCh38
rs761014024 Chr.16: 4576506-4576506 on Build GRCh38
rs201341219 Chr.16: 4576547-4576547 on Build GRCh38
rs144023335 Chr.16: 4576702-4576702 on Build GRCh38
rs904498351 Chr.16: 4576769-4576769 on Build GRCh38
rs1050064638 Chr.16: 4576793-4576793 on Build GRCh38

View Full Product Details