Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4804662-4805158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYR1
Entrez Gene ID
84656
Gene Name
glyoxylate reductase 1 homolog
Gene Aliases
BM045, HIBDL, N-PAC, NP60
UniGene
Hs.387255
Gene Chromosome Location
Chr.16: 4803203-4847388 on Build GRCh38
Transcripts
NM_001308096.1 NM_001324096.1 NM_001324097.1 NM_001324098.1 NM_032569.3 NR_136695.1 NR_136696.1 NR_136697.1 NR_136698.1 NR_136699.1 NR_136700.1 XM_005255639.4 XM_011522716.2 XM_011522717.1 XM_017023786.1 XM_017023787.1 XM_017023788.1 XM_017023789.1

SNP Detail

SNP ID Chromosome Location
rs750970373 Chr.16: 4804922-4804922 on Build GRCh38
rs1023579071 Chr.16: 4804799-4804799 on Build GRCh38
rs1035269772 Chr.16: 4804772-4804772 on Build GRCh38
rs1000190972 Chr.16: 4804740-4804740 on Build GRCh38
rs55911609 Chr.16: 4804926-4804926 on Build GRCh38
rs372373125 Chr.16: 4805104-4805104 on Build GRCh38
rs184634826 Chr.16: 4805135-4805135 on Build GRCh38
rs1012475765 Chr.16: 4804996-4804996 on Build GRCh38
rs886209751 Chr.16: 4804925-4804925 on Build GRCh38
rs1023209641 Chr.16: 4804940-4804941 on Build GRCh38
rs879047689 Chr.16: 4804934-4804935 on Build GRCh38
rs1047463644 Chr.16: 4804762-4804762 on Build GRCh38
rs1045265550 Chr.16: 4805028-4805028 on Build GRCh38
rs933178311 Chr.16: 4804915-4804915 on Build GRCh38
rs7498674 Chr.16: 4804875-4804875 on Build GRCh38
rs923129102 Chr.16: 4804898-4804898 on Build GRCh38
rs1037359863 Chr.16: 4804929-4804929 on Build GRCh38
rs765765561 Chr.16: 4804895-4804904 on Build GRCh38
rs1031813585 Chr.16: 4804970-4804971 on Build GRCh38
rs143169843 Chr.16: 4804720-4804720 on Build GRCh38
rs1043799127 Chr.16: 4804691-4804691 on Build GRCh38
rs59515878 Chr.16: 4804971-4804972 on Build GRCh38
rs974568292 Chr.16: 4804877-4804877 on Build GRCh38
rs556910257 Chr.16: 4804876-4804876 on Build GRCh38
rs373831597 Chr.16: 4804933-4804934 on Build GRCh38
rs902143808 Chr.16: 4804713-4804713 on Build GRCh38
rs941809870 Chr.16: 4805075-4805075 on Build GRCh38
rs896756785 Chr.16: 4804988-4804988 on Build GRCh38
rs1037411756 Chr.16: 4805088-4805088 on Build GRCh38
rs1005847824 Chr.16: 4804783-4804783 on Build GRCh38
rs990213834 Chr.16: 4805003-4805003 on Build GRCh38
rs879019389 Chr.16: 4804937-4804938 on Build GRCh38
rs867895165 Chr.16: 4804969-4804971 on Build GRCh38
rs947753855 Chr.16: 4805102-4805109 on Build GRCh38
rs142552275 Chr.16: 4804941-4804942 on Build GRCh38
rs1003522127 Chr.16: 4805119-4805124 on Build GRCh38
rs914562346 Chr.16: 4804863-4804863 on Build GRCh38
rs948946962 Chr.16: 4805072-4805101 on Build GRCh38
rs934997890 Chr.16: 4804978-4804978 on Build GRCh38
rs542471110 Chr.16: 4805083-4805083 on Build GRCh38
rs1021672636 Chr.16: 4805004-4805004 on Build GRCh38
rs894796506 Chr.16: 4804930-4804930 on Build GRCh38
rs971586968 Chr.16: 4804820-4804820 on Build GRCh38
rs965950330 Chr.16: 4805118-4805118 on Build GRCh38
rs921743281 Chr.16: 4804924-4804924 on Build GRCh38
rs569824796 Chr.16: 4804987-4804987 on Build GRCh38
rs1012400651 Chr.16: 4804789-4804789 on Build GRCh38
rs761905006 Chr.16: 4804710-4804711 on Build GRCh38
rs911797604 Chr.16: 4804873-4804873 on Build GRCh38
rs534568859 Chr.16: 4805101-4805101 on Build GRCh38
rs1003060458 Chr.16: 4804966-4804970 on Build GRCh38
rs538466642 Chr.16: 4805045-4805045 on Build GRCh38
rs983319131 Chr.16: 4805060-4805060 on Build GRCh38
rs547409474 Chr.16: 4804801-4804801 on Build GRCh38
rs150457072 Chr.16: 4804784-4804784 on Build GRCh38
rs762027640 Chr.16: 4804790-4804790 on Build GRCh38
rs150211381 Chr.16: 4804961-4804962 on Build GRCh38
rs560980591 Chr.16: 4804804-4804804 on Build GRCh38
rs995570941 Chr.16: 4804825-4804825 on Build GRCh38
rs184126747 Chr.16: 4804859-4804859 on Build GRCh38
rs117581021 Chr.16: 4805047-4805047 on Build GRCh38
rs191382860 Chr.16: 4805076-4805076 on Build GRCh38
rs879198164 Chr.16: 4804935-4804936 on Build GRCh38
rs956083857 Chr.16: 4804840-4804840 on Build GRCh38
rs766715537 Chr.16: 4804963-4804964 on Build GRCh38
rs867223872 Chr.16: 4804933-4804933 on Build GRCh38
rs190687610 Chr.16: 4804972-4804972 on Build GRCh38
rs1025720513 Chr.16: 4805061-4805061 on Build GRCh38
rs964731656 Chr.16: 4805011-4805011 on Build GRCh38
rs149944297 Chr.16: 4804934-4804935 on Build GRCh38
rs974600279 Chr.16: 4805130-4805130 on Build GRCh38
rs886157512 Chr.16: 4804775-4804775 on Build GRCh38
rs951490969 Chr.16: 4805100-4805100 on Build GRCh38
rs1013625673 Chr.16: 4804934-4804934 on Build GRCh38
rs1026067667 Chr.16: 4804827-4804827 on Build GRCh38
rs772453064 Chr.16: 4804828-4804828 on Build GRCh38
rs916914932 Chr.16: 4804892-4804905 on Build GRCh38
rs962800142 Chr.16: 4804800-4804800 on Build GRCh38
rs898765755 Chr.16: 4804788-4804788 on Build GRCh38
rs143624351 Chr.16: 4804934-4804937 on Build GRCh38
rs901656216 Chr.16: 4804717-4804717 on Build GRCh38

View Full Product Details