Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29798885-29799381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF22
Entrez Gene ID
3835
Gene Name
kinesin family member 22
Gene Aliases
A-328A3.2, KID, KNSL4, OBP, OBP-1, OBP-2, SEMDJL2
UniGene
Hs.612151
Gene Chromosome Location
Chr.16: 29790062-29805385 on Build GRCh38
Transcripts
NM_001256269.1 NM_001256270.1 NM_007317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4059932 c.574T>A p.S192T Chr.16: 29798999-29798999 on Build GRCh38
6320492 c.607G>T p.G203W Chr.16: 29799032-29799032 on Build GRCh38
4059933 c.725G>A p.R242H Chr.16: 29799150-29799150 on Build GRCh38
7134149 c.576G>C p.S192S Chr.16: 29799001-29799001 on Build GRCh38
5384684 c.711C>T p.T237T Chr.16: 29799136-29799136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542248557 Chr.16: 29799199-29799200 on Build GRCh38
rs755606500 Chr.16: 29798971-29798971 on Build GRCh38
rs760208125 Chr.16: 29799018-29799018 on Build GRCh38
rs558298646 Chr.16: 29799306-29799306 on Build GRCh38
rs746614468 Chr.16: 29798979-29798980 on Build GRCh38
rs201572301 Chr.16: 29799030-29799030 on Build GRCh38
rs772915372 Chr.16: 29799301-29799301 on Build GRCh38
rs754587489 Chr.16: 29799029-29799030 on Build GRCh38
rs771128685 Chr.16: 29799093-29799093 on Build GRCh38
rs756641324 Chr.16: 29799155-29799155 on Build GRCh38
rs774929756 Chr.16: 29799014-29799014 on Build GRCh38
rs761429507 Chr.16: 29799036-29799036 on Build GRCh38
rs544303336 Chr.16: 29798911-29798911 on Build GRCh38
rs776146951 Chr.16: 29799221-29799221 on Build GRCh38
rs753911757 Chr.16: 29799261-29799261 on Build GRCh38
rs139563547 Chr.16: 29799305-29799305 on Build GRCh38
rs776978629 Chr.16: 29799352-29799352 on Build GRCh38
rs369123318 Chr.16: 29798938-29798938 on Build GRCh38
rs770910651 Chr.16: 29799016-29799016 on Build GRCh38
rs371507469 Chr.16: 29798947-29798947 on Build GRCh38
rs781023894 Chr.16: 29799295-29799295 on Build GRCh38
rs763552072 Chr.16: 29799029-29799029 on Build GRCh38
rs748255068 Chr.16: 29798986-29798986 on Build GRCh38
rs757445899 Chr.16: 29799180-29799180 on Build GRCh38
rs760011053 Chr.16: 29799139-29799139 on Build GRCh38
rs768035064 Chr.16: 29799149-29799149 on Build GRCh38
rs550717370 Chr.16: 29799150-29799150 on Build GRCh38
rs760651443 Chr.16: 29799127-29799127 on Build GRCh38
rs372923745 Chr.16: 29799192-29799192 on Build GRCh38
rs1054144039 Chr.16: 29799296-29799296 on Build GRCh38
rs749516467 Chr.16: 29799092-29799092 on Build GRCh38
rs762743747 Chr.16: 29799234-29799234 on Build GRCh38
rs768199147 Chr.16: 29799130-29799130 on Build GRCh38
rs1021842236 Chr.16: 29798939-29798939 on Build GRCh38
rs202226507 Chr.16: 29798987-29798987 on Build GRCh38
rs375298225 Chr.16: 29799205-29799205 on Build GRCh38
rs747676044 Chr.16: 29799082-29799082 on Build GRCh38
rs926687990 Chr.16: 29799154-29799154 on Build GRCh38
rs776068017 Chr.16: 29799241-29799241 on Build GRCh38
rs779756888 Chr.16: 29799282-29799282 on Build GRCh38
rs1024180320 Chr.16: 29799066-29799066 on Build GRCh38
rs377204303 Chr.16: 29799000-29799000 on Build GRCh38
rs760972864 Chr.16: 29799346-29799346 on Build GRCh38
rs777907674 Chr.16: 29799160-29799160 on Build GRCh38
rs753871059 Chr.16: 29799179-29799179 on Build GRCh38
rs767449435 Chr.16: 29799013-29799013 on Build GRCh38
rs769166321 Chr.16: 29799300-29799300 on Build GRCh38
rs373688433 Chr.16: 29799257-29799257 on Build GRCh38
rs768481686 Chr.16: 29799211-29799211 on Build GRCh38
rs780326874 Chr.16: 29798951-29798951 on Build GRCh38
rs761181341 Chr.16: 29799254-29799254 on Build GRCh38
rs143337481 Chr.16: 29799280-29799280 on Build GRCh38
rs764557291 Chr.16: 29799351-29799351 on Build GRCh38
rs752273599 Chr.16: 29798930-29798930 on Build GRCh38
rs147345328 Chr.16: 29799294-29799294 on Build GRCh38
rs756629323 Chr.16: 29799198-29799198 on Build GRCh38
rs200255598 Chr.16: 29798931-29798931 on Build GRCh38
rs113436342 Chr.16: 29798936-29798936 on Build GRCh38
rs771954040 Chr.16: 29799110-29799110 on Build GRCh38
rs377092746 Chr.16: 29799226-29799226 on Build GRCh38
rs891464618 Chr.16: 29799349-29799349 on Build GRCh38
rs776071468 Chr.16: 29799135-29799135 on Build GRCh38
rs765346261 Chr.16: 29799274-29799274 on Build GRCh38
rs780709408 Chr.16: 29799052-29799053 on Build GRCh38
rs746126363 Chr.16: 29799193-29799193 on Build GRCh38
rs757250411 Chr.16: 29799273-29799273 on Build GRCh38
rs766761438 Chr.16: 29799040-29799040 on Build GRCh38
rs554780628 Chr.16: 29799298-29799298 on Build GRCh38
rs140332579 Chr.16: 29799057-29799057 on Build GRCh38
rs761249591 Chr.16: 29799224-29799224 on Build GRCh38
rs538207110 Chr.16: 29799068-29799068 on Build GRCh38
rs745683317 Chr.16: 29799103-29799103 on Build GRCh38
rs201659270 Chr.16: 29799120-29799120 on Build GRCh38
rs778704987 Chr.16: 29799094-29799094 on Build GRCh38
rs761461411 Chr.16: 29799136-29799136 on Build GRCh38
rs751925003 Chr.16: 29799138-29799138 on Build GRCh38
rs1054392 Chr.16: 29799089-29799089 on Build GRCh38
rs781770633 Chr.16: 29799049-29799049 on Build GRCh38
rs190757548 Chr.16: 29799048-29799048 on Build GRCh38
rs773517779 Chr.16: 29798990-29798990 on Build GRCh38
rs373330722 Chr.16: 29799279-29799279 on Build GRCh38
rs370334885 Chr.16: 29799322-29799322 on Build GRCh38
rs750713541 Chr.16: 29798944-29798944 on Build GRCh38
rs767001936 Chr.16: 29798928-29798928 on Build GRCh38
rs767984235 Chr.16: 29799240-29799240 on Build GRCh38
rs369142316 Chr.16: 29799230-29799230 on Build GRCh38
rs11545431 Chr.16: 29799001-29799001 on Build GRCh38
rs369995078 Chr.16: 29799082-29799082 on Build GRCh38
rs781352160 Chr.16: 29798975-29798975 on Build GRCh38
rs929602607 Chr.16: 29799204-29799204 on Build GRCh38
rs773176288 Chr.16: 29799227-29799227 on Build GRCh38
rs746216092 Chr.16: 29798955-29798955 on Build GRCh38
rs11545428 Chr.16: 29799004-29799004 on Build GRCh38
rs775371597 Chr.16: 29799114-29799114 on Build GRCh38
rs374024953 Chr.16: 29799137-29799137 on Build GRCh38
rs768729822 Chr.16: 29799286-29799286 on Build GRCh38
rs760893454 Chr.16: 29799199-29799199 on Build GRCh38

View Full Product Details