Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29802612-29803120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF22
Entrez Gene ID
3835
Gene Name
kinesin family member 22
Gene Aliases
A-328A3.2, KID, KNSL4, OBP, OBP-1, OBP-2, SEMDJL2
UniGene
Hs.612151
Gene Chromosome Location
Chr.16: 29790062-29805385 on Build GRCh38
Transcripts
NM_001256269.1 NM_001256270.1 NM_007317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1301836 c.1441G>A p.E481K Chr.16: 29802929-29802929 on Build GRCh38
3110919 c.1343G>A p.R448H Chr.16: 29802831-29802831 on Build GRCh38
5508639 c.1315A>G p.M439V Chr.16: 29802803-29802803 on Build GRCh38
7313786 c.1325G>T p.R442L Chr.16: 29802813-29802813 on Build GRCh38
3508572 c.1330C>T p.L444F Chr.16: 29802818-29802818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995558505 Chr.16: 29803094-29803094 on Build GRCh38
rs774808227 Chr.16: 29802926-29802926 on Build GRCh38
rs754947238 Chr.16: 29802979-29802979 on Build GRCh38
rs141728416 Chr.16: 29802827-29802827 on Build GRCh38
rs2450399 Chr.16: 29802913-29802913 on Build GRCh38
rs141172780 Chr.16: 29803037-29803037 on Build GRCh38
rs767966150 Chr.16: 29802937-29802937 on Build GRCh38
rs764188334 Chr.16: 29802956-29802956 on Build GRCh38
rs985653640 Chr.16: 29802982-29802982 on Build GRCh38
rs570149400 Chr.16: 29803027-29803027 on Build GRCh38
rs750680224 Chr.16: 29802955-29802955 on Build GRCh38
rs768036085 Chr.16: 29802895-29802895 on Build GRCh38
rs779821702 Chr.16: 29802890-29802890 on Build GRCh38
rs935279128 Chr.16: 29802826-29802826 on Build GRCh38
rs752098611 Chr.16: 29802735-29802735 on Build GRCh38
rs555996211 Chr.16: 29803050-29803050 on Build GRCh38
rs757028180 Chr.16: 29802764-29802764 on Build GRCh38
rs767639850 Chr.16: 29802740-29802740 on Build GRCh38
rs764267174 Chr.16: 29802934-29802934 on Build GRCh38
rs777888954 Chr.16: 29802751-29802751 on Build GRCh38
rs756120432 Chr.16: 29802862-29802862 on Build GRCh38
rs778742395 Chr.16: 29802776-29802776 on Build GRCh38
rs764285568 Chr.16: 29802863-29802863 on Build GRCh38
rs189904457 Chr.16: 29803091-29803091 on Build GRCh38
rs200809627 Chr.16: 29802891-29802891 on Build GRCh38
rs899921274 Chr.16: 29803030-29803030 on Build GRCh38
rs1043807128 Chr.16: 29802799-29802799 on Build GRCh38
rs768252159 Chr.16: 29802806-29802806 on Build GRCh38
rs371789170 Chr.16: 29802748-29802748 on Build GRCh38
rs759881029 Chr.16: 29802830-29802830 on Build GRCh38
rs147785290 Chr.16: 29802941-29802941 on Build GRCh38
rs771618349 Chr.16: 29803051-29803052 on Build GRCh38
rs536220400 Chr.16: 29803043-29803043 on Build GRCh38
rs775013851 Chr.16: 29802792-29802792 on Build GRCh38
rs757548979 Chr.16: 29802980-29802980 on Build GRCh38
rs752732730 Chr.16: 29802741-29802741 on Build GRCh38
rs145879411 Chr.16: 29802885-29802885 on Build GRCh38
rs368889638 Chr.16: 29802970-29802970 on Build GRCh38
rs753925181 Chr.16: 29802944-29802944 on Build GRCh38
rs1002381556 Chr.16: 29803055-29803055 on Build GRCh38
rs540131382 Chr.16: 29802995-29802995 on Build GRCh38
rs561685861 Chr.16: 29803054-29803054 on Build GRCh38
rs936639815 Chr.16: 29803031-29803032 on Build GRCh38
rs1025130662 Chr.16: 29802833-29802833 on Build GRCh38
rs764080777 Chr.16: 29802942-29802942 on Build GRCh38
rs753205824 Chr.16: 29802896-29802896 on Build GRCh38
rs1053808625 Chr.16: 29803037-29803038 on Build GRCh38
rs958495823 Chr.16: 29803083-29803083 on Build GRCh38
rs199715417 Chr.16: 29802791-29802791 on Build GRCh38
rs776122940 Chr.16: 29802812-29802812 on Build GRCh38
rs929915133 Chr.16: 29802978-29802978 on Build GRCh38
rs944112139 Chr.16: 29802697-29802697 on Build GRCh38
rs1034482563 Chr.16: 29803062-29803062 on Build GRCh38
rs896342388 Chr.16: 29802831-29802831 on Build GRCh38
rs946473701 Chr.16: 29802784-29802784 on Build GRCh38
rs781257402 Chr.16: 29802986-29802986 on Build GRCh38
rs890980383 Chr.16: 29802668-29802668 on Build GRCh38
rs551921234 Chr.16: 29803024-29803024 on Build GRCh38
rs749567581 Chr.16: 29802759-29802759 on Build GRCh38
rs1009809775 Chr.16: 29803038-29803038 on Build GRCh38
rs747735559 Chr.16: 29802903-29802903 on Build GRCh38
rs755608574 Chr.16: 29802736-29802736 on Build GRCh38
rs767933971 Chr.16: 29802857-29802857 on Build GRCh38
rs571968439 Chr.16: 29802715-29802715 on Build GRCh38
rs768444693 Chr.16: 29802692-29802694 on Build GRCh38
rs765833732 Chr.16: 29802975-29802975 on Build GRCh38
rs150131602 Chr.16: 29802859-29802859 on Build GRCh38
rs745681316 Chr.16: 29802884-29802884 on Build GRCh38
rs754050855 Chr.16: 29802869-29802869 on Build GRCh38
rs778617888 Chr.16: 29802883-29802883 on Build GRCh38
rs565948374 Chr.16: 29802951-29802951 on Build GRCh38
rs376040090 Chr.16: 29802788-29802788 on Build GRCh38
rs757545358 Chr.16: 29802875-29802875 on Build GRCh38
rs372337472 Chr.16: 29802798-29802798 on Build GRCh38
rs762022584 Chr.16: 29802911-29802913 on Build GRCh38
rs2516819 Chr.16: 29802962-29802962 on Build GRCh38
rs760062208 Chr.16: 29802836-29802836 on Build GRCh38
rs201631186 Chr.16: 29802813-29802813 on Build GRCh38
rs968240230 Chr.16: 29803010-29803010 on Build GRCh38
rs779954401 Chr.16: 29802969-29802969 on Build GRCh38
rs766897915 Chr.16: 29802721-29802721 on Build GRCh38
rs559884972 Chr.16: 29802662-29802662 on Build GRCh38
rs766845044 Chr.16: 29802822-29802822 on Build GRCh38
rs765471633 Chr.16: 29802954-29802954 on Build GRCh38
rs917862411 Chr.16: 29803064-29803064 on Build GRCh38

View Full Product Details