Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29816488-29816982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAGR1
Entrez Gene ID
79447
Gene Name
PAXIP1 associated glutamate rich protein 1
Gene Aliases
C16orf53, GAS, PA1
UniGene
Hs.702841
Gene Chromosome Location
Chr.16: 29816207-29822495 on Build GRCh38
Transcripts
NM_024516.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7251711 c.244G>C p.E82Q Chr.16: 29816769-29816769 on Build GRCh38
5971749 c.29C>T p.T10I Chr.16: 29816554-29816554 on Build GRCh38
7165679 c.119C>G p.S40C Chr.16: 29816644-29816644 on Build GRCh38
7002706 c.221C>A p.A74D Chr.16: 29816746-29816746 on Build GRCh38
5955483 c.303T>A p.D101E Chr.16: 29816828-29816828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758338974 Chr.16: 29816956-29816956 on Build GRCh38
rs943606661 Chr.16: 29816648-29816648 on Build GRCh38
rs746515233 Chr.16: 29816719-29816719 on Build GRCh38
rs960939549 Chr.16: 29816695-29816695 on Build GRCh38
rs534165002 Chr.16: 29816848-29816848 on Build GRCh38
rs753983836 Chr.16: 29816911-29816911 on Build GRCh38
rs745681136 Chr.16: 29816825-29816825 on Build GRCh38
rs778970803 Chr.16: 29816701-29816701 on Build GRCh38
rs779860747 Chr.16: 29816843-29816843 on Build GRCh38
rs780443230 Chr.16: 29816546-29816546 on Build GRCh38
rs375076291 Chr.16: 29816526-29816526 on Build GRCh38
rs369349605 Chr.16: 29816865-29816865 on Build GRCh38
rs759895360 Chr.16: 29816887-29816887 on Build GRCh38
rs757085801 Chr.16: 29816811-29816811 on Build GRCh38
rs550827638 Chr.16: 29816600-29816600 on Build GRCh38
rs867944841 Chr.16: 29816583-29816583 on Build GRCh38
rs768022179 Chr.16: 29816783-29816783 on Build GRCh38
rs201354663 Chr.16: 29816731-29816731 on Build GRCh38
rs754054325 Chr.16: 29816802-29816802 on Build GRCh38
rs777577339 Chr.16: 29816737-29816737 on Build GRCh38
rs918289522 Chr.16: 29816862-29816862 on Build GRCh38
rs370072946 Chr.16: 29816912-29816912 on Build GRCh38
rs1029443377 Chr.16: 29816585-29816585 on Build GRCh38
rs558886856 Chr.16: 29816903-29816903 on Build GRCh38
rs781381892 Chr.16: 29816569-29816569 on Build GRCh38
rs752120723 Chr.16: 29816660-29816660 on Build GRCh38
rs751073322 Chr.16: 29816599-29816605 on Build GRCh38
rs757058094 Chr.16: 29816700-29816700 on Build GRCh38
rs780693517 Chr.16: 29816663-29816668 on Build GRCh38
rs748407512 Chr.16: 29816572-29816572 on Build GRCh38
rs754539974 Chr.16: 29816611-29816612 on Build GRCh38
rs778813478 Chr.16: 29816817-29816817 on Build GRCh38
rs753000270 Chr.16: 29816557-29816557 on Build GRCh38
rs763365665 Chr.16: 29816744-29816744 on Build GRCh38
rs768003848 Chr.16: 29816896-29816896 on Build GRCh38
rs909600238 Chr.16: 29816815-29816815 on Build GRCh38
rs764981460 Chr.16: 29816556-29816556 on Build GRCh38
rs769752602 Chr.16: 29816712-29816712 on Build GRCh38
rs769936733 Chr.16: 29816579-29816579 on Build GRCh38
rs572999151 Chr.16: 29816844-29816844 on Build GRCh38
rs373200391 Chr.16: 29816884-29816884 on Build GRCh38
rs540688966 Chr.16: 29816583-29816585 on Build GRCh38
rs750176614 Chr.16: 29816954-29816954 on Build GRCh38
rs200743734 Chr.16: 29816697-29816697 on Build GRCh38
rs1043266857 Chr.16: 29816650-29816650 on Build GRCh38
rs375215655 Chr.16: 29816799-29816799 on Build GRCh38
rs764118426 Chr.16: 29816910-29816910 on Build GRCh38
rs759983516 Chr.16: 29816774-29816774 on Build GRCh38
rs772168243 Chr.16: 29816712-29816712 on Build GRCh38
rs748010806 Chr.16: 29816671-29816672 on Build GRCh38
rs745842510 Chr.16: 29816709-29816709 on Build GRCh38
rs1031459821 Chr.16: 29816710-29816710 on Build GRCh38
rs760337168 Chr.16: 29816623-29816623 on Build GRCh38
rs776217162 Chr.16: 29816742-29816742 on Build GRCh38
rs11546677 Chr.16: 29816511-29816511 on Build GRCh38
rs368639575 Chr.16: 29816554-29816554 on Build GRCh38
rs941009693 Chr.16: 29816549-29816549 on Build GRCh38
rs780031691 Chr.16: 29816962-29816962 on Build GRCh38
rs377615776 Chr.16: 29816901-29816901 on Build GRCh38
rs750741846 Chr.16: 29816641-29816641 on Build GRCh38
rs774727009 Chr.16: 29816752-29816752 on Build GRCh38
rs374459711 Chr.16: 29816941-29816941 on Build GRCh38
rs752751554 Chr.16: 29816790-29816790 on Build GRCh38
rs146149602 Chr.16: 29816537-29816537 on Build GRCh38
rs763236125 Chr.16: 29816654-29816654 on Build GRCh38
rs770834886 Chr.16: 29816604-29816604 on Build GRCh38
rs758045357 Chr.16: 29816576-29816579 on Build GRCh38
rs1022355821 Chr.16: 29816652-29816652 on Build GRCh38
rs774495877 Chr.16: 29816607-29816607 on Build GRCh38
rs758889974 Chr.16: 29816544-29816544 on Build GRCh38
rs775253992 Chr.16: 29816616-29816616 on Build GRCh38
rs1031459501 Chr.16: 29816676-29816676 on Build GRCh38
rs112242011 Chr.16: 29816743-29816743 on Build GRCh38
rs756298039 Chr.16: 29816692-29816692 on Build GRCh38
rs916729163 Chr.16: 29816725-29816725 on Build GRCh38
rs867495905 Chr.16: 29816955-29816955 on Build GRCh38
rs771755166 Chr.16: 29816613-29816613 on Build GRCh38
rs1036699712 Chr.16: 29816851-29816851 on Build GRCh38
rs763677620 Chr.16: 29816636-29816636 on Build GRCh38
rs867151087 Chr.16: 29816926-29816926 on Build GRCh38
rs777571525 Chr.16: 29816592-29816592 on Build GRCh38
rs755031511 Chr.16: 29816664-29816664 on Build GRCh38
rs34508592 Chr.16: 29816587-29816587 on Build GRCh38
rs571382632 Chr.16: 29816518-29816518 on Build GRCh38
rs767842802 Chr.16: 29816610-29816610 on Build GRCh38
rs752738892 Chr.16: 29816682-29816682 on Build GRCh38
rs758354996 Chr.16: 29816827-29816827 on Build GRCh38
rs747798431 Chr.16: 29816872-29816872 on Build GRCh38
rs769455895 Chr.16: 29816879-29816879 on Build GRCh38
rs750785330 Chr.16: 29816539-29816539 on Build GRCh38
rs773935759 Chr.16: 29816509-29816509 on Build GRCh38
rs879756491 Chr.16: 29816555-29816555 on Build GRCh38
rs184630497 Chr.16: 29816562-29816562 on Build GRCh38
rs889644757 Chr.16: 29816591-29816591 on Build GRCh38
rs775092459 Chr.16: 29816716-29816716 on Build GRCh38
rs554672402 Chr.16: 29816751-29816751 on Build GRCh38
rs767543291 Chr.16: 29816679-29816679 on Build GRCh38
rs762064824 Chr.16: 29816521-29816521 on Build GRCh38
rs868558474 Chr.16: 29816899-29816899 on Build GRCh38
rs1022592936 Chr.16: 29816762-29816762 on Build GRCh38
rs11546679 Chr.16: 29816655-29816655 on Build GRCh38
rs767273972 Chr.16: 29816514-29816514 on Build GRCh38
rs1050989106 Chr.16: 29816880-29816880 on Build GRCh38
rs777858853 Chr.16: 29816694-29816694 on Build GRCh38

View Full Product Details