Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30184129-30184633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1A
Entrez Gene ID
11151
Gene Name
coronin 1A
Gene Aliases
CLABP, CLIPINA, HCORO1, IMD8, TACO, p57
UniGene
Hs.415067
Gene Chromosome Location
Chr.16: 30183410-30189076 on Build GRCh38
Transcripts
NM_001193333.2 NM_007074.3 XM_011545714.2 XM_017022885.1 XM_017022886.1

SNP Detail

SNP ID Chromosome Location
rs1006073321 Chr.16: 30184392-30184392 on Build GRCh38
rs372973829 Chr.16: 30184289-30184289 on Build GRCh38
rs1052760722 Chr.16: 30184151-30184151 on Build GRCh38
rs116307333 Chr.16: 30184213-30184213 on Build GRCh38
rs773741586 Chr.16: 30184193-30184193 on Build GRCh38
rs576598709 Chr.16: 30184472-30184472 on Build GRCh38
rs947554535 Chr.16: 30184607-30184607 on Build GRCh38
rs1025591696 Chr.16: 30184171-30184171 on Build GRCh38
rs987489911 Chr.16: 30184357-30184357 on Build GRCh38
rs1002872326 Chr.16: 30184206-30184206 on Build GRCh38
rs72791248 Chr.16: 30184470-30184470 on Build GRCh38
rs925924791 Chr.16: 30184447-30184447 on Build GRCh38
rs554312370 Chr.16: 30184304-30184304 on Build GRCh38
rs1012992122 Chr.16: 30184353-30184353 on Build GRCh38
rs187250127 Chr.16: 30184419-30184419 on Build GRCh38
rs759773653 Chr.16: 30184573-30184574 on Build GRCh38
rs182549352 Chr.16: 30184169-30184169 on Build GRCh38
rs895470666 Chr.16: 30184540-30184540 on Build GRCh38
rs748616446 Chr.16: 30184158-30184158 on Build GRCh38
rs999240803 Chr.16: 30184321-30184321 on Build GRCh38
rs762831777 Chr.16: 30184606-30184606 on Build GRCh38
rs923002569 Chr.16: 30184487-30184487 on Build GRCh38
rs943176477 Chr.16: 30184383-30184383 on Build GRCh38
rs771250125 Chr.16: 30184209-30184209 on Build GRCh38
rs747429343 Chr.16: 30184195-30184195 on Build GRCh38
rs954823912 Chr.16: 30184356-30184356 on Build GRCh38
rs562603068 Chr.16: 30184594-30184594 on Build GRCh38
rs1044113650 Chr.16: 30184554-30184554 on Build GRCh38
rs910725929 Chr.16: 30184216-30184216 on Build GRCh38
rs536205411 Chr.16: 30184220-30184220 on Build GRCh38
rs765362619 Chr.16: 30184338-30184338 on Build GRCh38
rs1043340772 Chr.16: 30184254-30184254 on Build GRCh38
rs1016579447 Chr.16: 30184400-30184400 on Build GRCh38
rs963589908 Chr.16: 30184264-30184264 on Build GRCh38
rs954573077 Chr.16: 30184534-30184534 on Build GRCh38
rs78466713 Chr.16: 30184593-30184593 on Build GRCh38
rs1057159236 Chr.16: 30184329-30184329 on Build GRCh38
rs118107356 Chr.16: 30184368-30184368 on Build GRCh38
rs947585018 Chr.16: 30184249-30184249 on Build GRCh38
rs985884385 Chr.16: 30184553-30184553 on Build GRCh38
rs969208650 Chr.16: 30184369-30184369 on Build GRCh38
rs969561153 Chr.16: 30184192-30184192 on Build GRCh38
rs866773375 Chr.16: 30184344-30184344 on Build GRCh38
rs560063164 Chr.16: 30184579-30184579 on Build GRCh38
rs1052793224 Chr.16: 30184522-30184522 on Build GRCh38
rs868448826 Chr.16: 30184291-30184291 on Build GRCh38
rs949793040 Chr.16: 30184541-30184541 on Build GRCh38

View Full Product Details