Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50026265-50026664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNEP1R1
Entrez Gene ID
255919
Gene Name
CTD nuclear envelope phosphatase 1 regulatory subunit 1
Gene Aliases
C16orf69, NEP1-R1, NEP1R1, TMEM188, TMP125
UniGene
Hs.59134
Gene Chromosome Location
Chr.16: 50025206-50037088 on Build GRCh38
Transcripts
NM_001281789.1 NM_153261.5 NR_104042.1 NR_104043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1609375 c.138A>T p.G46G Chr.16: 50026457-50026457 on Build GRCh38
5779947 c.105T>C p.T35T Chr.16: 50026424-50026424 on Build GRCh38
5794087 c.140G>A p.R47H Chr.16: 50026459-50026459 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs569209008 Chr.16: 50026377-50026377 on Build GRCh38
rs559597829 Chr.16: 50026504-50026507 on Build GRCh38
rs1046958817 Chr.16: 50026528-50026528 on Build GRCh38
rs935027881 Chr.16: 50026631-50026631 on Build GRCh38
rs750912118 Chr.16: 50026417-50026417 on Build GRCh38
rs17213264 Chr.16: 50026292-50026292 on Build GRCh38
rs552364603 Chr.16: 50026373-50026373 on Build GRCh38
rs759435097 Chr.16: 50026505-50026505 on Build GRCh38
rs376368462 Chr.16: 50026352-50026352 on Build GRCh38
rs927082440 Chr.16: 50026494-50026494 on Build GRCh38
rs565589295 Chr.16: 50026311-50026313 on Build GRCh38
rs764542639 Chr.16: 50026382-50026382 on Build GRCh38
rs923726536 Chr.16: 50026630-50026630 on Build GRCh38
rs781748007 Chr.16: 50026392-50026392 on Build GRCh38
rs371032588 Chr.16: 50026371-50026371 on Build GRCh38
rs994299512 Chr.16: 50026476-50026476 on Build GRCh38
rs913429127 Chr.16: 50026451-50026451 on Build GRCh38
rs965126552 Chr.16: 50026355-50026355 on Build GRCh38
rs373002381 Chr.16: 50026380-50026380 on Build GRCh38
rs760477957 Chr.16: 50026369-50026369 on Build GRCh38
rs777408049 Chr.16: 50026469-50026469 on Build GRCh38
rs748020845 Chr.16: 50026384-50026384 on Build GRCh38
rs533512866 Chr.16: 50026545-50026545 on Build GRCh38
rs748750846 Chr.16: 50026596-50026596 on Build GRCh38
rs374230724 Chr.16: 50026597-50026597 on Build GRCh38
rs778326180 Chr.16: 50026440-50026440 on Build GRCh38
rs551320062 Chr.16: 50026637-50026637 on Build GRCh38
rs765285578 Chr.16: 50026379-50026379 on Build GRCh38
rs553452202 Chr.16: 50026618-50026618 on Build GRCh38
rs773057469 Chr.16: 50026367-50026367 on Build GRCh38
rs947560758 Chr.16: 50026477-50026477 on Build GRCh38
rs1016339662 Chr.16: 50026296-50026297 on Build GRCh38
rs766239146 Chr.16: 50026372-50026372 on Build GRCh38
rs540858490 Chr.16: 50026498-50026498 on Build GRCh38
rs888308445 Chr.16: 50026576-50026576 on Build GRCh38
rs376719250 Chr.16: 50026478-50026478 on Build GRCh38
rs772887076 Chr.16: 50026482-50026482 on Build GRCh38
rs769917132 Chr.16: 50026511-50026511 on Build GRCh38
rs201107349 Chr.16: 50026457-50026457 on Build GRCh38
rs190443682 Chr.16: 50026512-50026512 on Build GRCh38
rs965104512 Chr.16: 50026562-50026562 on Build GRCh38
rs747804577 Chr.16: 50026346-50026346 on Build GRCh38
rs747830183 Chr.16: 50026454-50026454 on Build GRCh38
rs775205748 Chr.16: 50026497-50026500 on Build GRCh38
rs576591148 Chr.16: 50026388-50026388 on Build GRCh38
rs769402287 Chr.16: 50026431-50026431 on Build GRCh38
rs897237516 Chr.16: 50026403-50026403 on Build GRCh38
rs757697295 Chr.16: 50026387-50026387 on Build GRCh38
rs1044801676 Chr.16: 50026338-50026338 on Build GRCh38
rs905838391 Chr.16: 50026361-50026361 on Build GRCh38

View Full Product Details