Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50035326-50035822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNEP1R1
Entrez Gene ID
255919
Gene Name
CTD nuclear envelope phosphatase 1 regulatory subunit 1
Gene Aliases
C16orf69, NEP1-R1, NEP1R1, TMEM188, TMP125
UniGene
Hs.59134
Gene Chromosome Location
Chr.16: 50025206-50037088 on Build GRCh38
Transcripts
NM_001281789.1 NM_153261.5 NR_104042.1 NR_104043.1

SNP Detail

SNP ID Chromosome Location
rs776453456 Chr.16: 50035398-50035399 on Build GRCh38
rs34922460 Chr.16: 50035649-50035650 on Build GRCh38
rs898149495 Chr.16: 50035597-50035597 on Build GRCh38
rs936274056 Chr.16: 50035412-50035412 on Build GRCh38
rs777977157 Chr.16: 50035411-50035411 on Build GRCh38
rs950314487 Chr.16: 50035753-50035753 on Build GRCh38
rs373420066 Chr.16: 50035501-50035501 on Build GRCh38
rs1008066879 Chr.16: 50035657-50035657 on Build GRCh38
rs766062916 Chr.16: 50035502-50035502 on Build GRCh38
rs967495096 Chr.16: 50035727-50035727 on Build GRCh38
rs916168183 Chr.16: 50035751-50035751 on Build GRCh38
rs955426587 Chr.16: 50035648-50035648 on Build GRCh38
rs746195751 Chr.16: 50035464-50035464 on Build GRCh38
rs55653857 Chr.16: 50035476-50035476 on Build GRCh38
rs185459967 Chr.16: 50035537-50035537 on Build GRCh38
rs747187457 Chr.16: 50035459-50035459 on Build GRCh38
rs757827144 Chr.16: 50035472-50035472 on Build GRCh38
rs950265235 Chr.16: 50035467-50035467 on Build GRCh38
rs1020792417 Chr.16: 50035680-50035680 on Build GRCh38
rs967421578 Chr.16: 50035381-50035381 on Build GRCh38
rs771129348 Chr.16: 50035461-50035461 on Build GRCh38
rs546362775 Chr.16: 50035730-50035730 on Build GRCh38
rs779233705 Chr.16: 50035495-50035495 on Build GRCh38
rs758379345 Chr.16: 50035403-50035403 on Build GRCh38
rs369053494 Chr.16: 50035463-50035463 on Build GRCh38
rs570087741 Chr.16: 50035401-50035402 on Build GRCh38
rs911980427 Chr.16: 50035366-50035367 on Build GRCh38
rs775949974 Chr.16: 50035477-50035477 on Build GRCh38
rs2041918 Chr.16: 50035401-50035401 on Build GRCh38
rs991875896 Chr.16: 50035748-50035748 on Build GRCh38
rs527312214 Chr.16: 50035625-50035625 on Build GRCh38
rs561603319 Chr.16: 50035544-50035544 on Build GRCh38
rs1003912631 Chr.16: 50035563-50035563 on Build GRCh38
rs531670648 Chr.16: 50035504-50035504 on Build GRCh38
rs562539278 Chr.16: 50035482-50035482 on Build GRCh38
rs754825142 Chr.16: 50035396-50035396 on Build GRCh38
rs372382445 Chr.16: 50035397-50035397 on Build GRCh38
rs770163275 Chr.16: 50035468-50035468 on Build GRCh38
rs914750060 Chr.16: 50035448-50035448 on Build GRCh38
rs370646179 Chr.16: 50035523-50035523 on Build GRCh38
rs1046280001 Chr.16: 50035528-50035528 on Build GRCh38
rs1038043074 Chr.16: 50035590-50035590 on Build GRCh38
rs1028256811 Chr.16: 50035646-50035646 on Build GRCh38
rs772835338 Chr.16: 50035499-50035499 on Build GRCh38
rs778946495 Chr.16: 50035398-50035398 on Build GRCh38
rs747715588 Chr.16: 50035469-50035469 on Build GRCh38
rs765018965 Chr.16: 50035402-50035403 on Build GRCh38
rs528092040 Chr.16: 50035546-50035546 on Build GRCh38
rs753629058 Chr.16: 50035384-50035384 on Build GRCh38
rs957408396 Chr.16: 50035747-50035747 on Build GRCh38
rs763553501 Chr.16: 50035478-50035478 on Build GRCh38
rs925970466 Chr.16: 50035746-50035746 on Build GRCh38

View Full Product Details