Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57460689-57461228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ9
Entrez Gene ID
57017
Gene Name
coenzyme Q9
Gene Aliases
C16orf49, COQ10D5
UniGene
Hs.513632
Gene Chromosome Location
Chr.16: 57447425-57461275 on Build GRCh38
Transcripts
NM_020312.3

SNP Detail

SNP ID Chromosome Location
rs223870 Chr.16: 57461114-57461114 on Build GRCh38
rs1018196913 Chr.16: 57460759-57460759 on Build GRCh38
rs779379396 Chr.16: 57460795-57460795 on Build GRCh38
rs543575586 Chr.16: 57460725-57460725 on Build GRCh38
rs914235589 Chr.16: 57460925-57460925 on Build GRCh38
rs780726490 Chr.16: 57461172-57461172 on Build GRCh38
rs773067298 Chr.16: 57461099-57461099 on Build GRCh38
rs112075830 Chr.16: 57461035-57461035 on Build GRCh38
rs902712552 Chr.16: 57461026-57461026 on Build GRCh38
rs1025528110 Chr.16: 57461111-57461111 on Build GRCh38
rs1007377640 Chr.16: 57461059-57461059 on Build GRCh38
rs111229926 Chr.16: 57460723-57460723 on Build GRCh38
rs886052182 Chr.16: 57460758-57460758 on Build GRCh38
rs1006894488 Chr.16: 57460728-57460728 on Build GRCh38
rs1043983387 Chr.16: 57461014-57461014 on Build GRCh38
rs748805729 Chr.16: 57460862-57460862 on Build GRCh38
rs202078553 Chr.16: 57461068-57461073 on Build GRCh38
rs1051660120 Chr.16: 57461044-57461044 on Build GRCh38
rs571758971 Chr.16: 57461162-57461162 on Build GRCh38
rs223869 Chr.16: 57461080-57461080 on Build GRCh38
rs139650688 Chr.16: 57461194-57461194 on Build GRCh38
rs765505205 Chr.16: 57461124-57461124 on Build GRCh38
rs928424637 Chr.16: 57460835-57460835 on Build GRCh38
rs1025453095 Chr.16: 57460805-57460805 on Build GRCh38
rs747859383 Chr.16: 57461010-57461010 on Build GRCh38
rs768190161 Chr.16: 57460872-57460872 on Build GRCh38
rs991164313 Chr.16: 57460900-57460900 on Build GRCh38
rs973574071 Chr.16: 57460908-57460908 on Build GRCh38
rs749128086 Chr.16: 57461202-57461202 on Build GRCh38
rs886052183 Chr.16: 57461189-57461189 on Build GRCh38
rs951231455 Chr.16: 57460811-57460811 on Build GRCh38
rs991234919 Chr.16: 57461170-57461170 on Build GRCh38
rs190372643 Chr.16: 57461002-57461002 on Build GRCh38
rs1035900826 Chr.16: 57461167-57461167 on Build GRCh38
rs560489131 Chr.16: 57460874-57460874 on Build GRCh38
rs995410830 Chr.16: 57461106-57461106 on Build GRCh38
rs891218580 Chr.16: 57461046-57461046 on Build GRCh38
rs553993142 Chr.16: 57460794-57460794 on Build GRCh38
rs115123016 Chr.16: 57461169-57461169 on Build GRCh38
rs550304608 Chr.16: 57461116-57461116 on Build GRCh38
rs373048666 Chr.16: 57460724-57460724 on Build GRCh38
rs751291933 Chr.16: 57461128-57461128 on Build GRCh38
rs532394134 Chr.16: 57460815-57460815 on Build GRCh38
rs547752339 Chr.16: 57460819-57460819 on Build GRCh38
rs376172393 Chr.16: 57460792-57460792 on Build GRCh38

View Full Product Details