Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57461117-57461614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ9
Entrez Gene ID
57017
Gene Name
coenzyme Q9
Gene Aliases
C16orf49, COQ10D5
UniGene
Hs.513632
Gene Chromosome Location
Chr.16: 57447425-57461275 on Build GRCh38
Transcripts
NM_020312.3

SNP Detail

SNP ID Chromosome Location
rs887040419 Chr.16: 57461421-57461421 on Build GRCh38
rs878862876 Chr.16: 57461315-57461315 on Build GRCh38
rs565315346 Chr.16: 57461356-57461356 on Build GRCh38
rs977057624 Chr.16: 57461274-57461274 on Build GRCh38
rs924219251 Chr.16: 57461276-57461276 on Build GRCh38
rs1047012802 Chr.16: 57461411-57461411 on Build GRCh38
rs772192873 Chr.16: 57461333-57461337 on Build GRCh38
rs759658598 Chr.16: 57461427-57461427 on Build GRCh38
rs1039747612 Chr.16: 57461368-57461368 on Build GRCh38
rs958766876 Chr.16: 57461449-57461449 on Build GRCh38
rs780726490 Chr.16: 57461172-57461172 on Build GRCh38
rs1035570140 Chr.16: 57461448-57461448 on Build GRCh38
rs1021398375 Chr.16: 57461518-57461518 on Build GRCh38
rs11547481 Chr.16: 57461206-57461206 on Build GRCh38
rs576041450 Chr.16: 57461505-57461505 on Build GRCh38
rs968416728 Chr.16: 57461521-57461521 on Build GRCh38
rs1051305384 Chr.16: 57461293-57461293 on Build GRCh38
rs571758971 Chr.16: 57461162-57461162 on Build GRCh38
rs554159552 Chr.16: 57461462-57461462 on Build GRCh38
rs924263925 Chr.16: 57461586-57461586 on Build GRCh38
rs139650688 Chr.16: 57461194-57461194 on Build GRCh38
rs534696904 Chr.16: 57461246-57461246 on Build GRCh38
rs142842163 Chr.16: 57461527-57461527 on Build GRCh38
rs535878960 Chr.16: 57461385-57461385 on Build GRCh38
rs554709298 Chr.16: 57461275-57461275 on Build GRCh38
rs891286239 Chr.16: 57461342-57461342 on Build GRCh38
rs935547343 Chr.16: 57461283-57461283 on Build GRCh38
rs3177052 Chr.16: 57461271-57461271 on Build GRCh38
rs749128086 Chr.16: 57461202-57461202 on Build GRCh38
rs886052183 Chr.16: 57461189-57461189 on Build GRCh38
rs977092140 Chr.16: 57461564-57461564 on Build GRCh38
rs991234919 Chr.16: 57461170-57461170 on Build GRCh38
rs140036561 Chr.16: 57461456-57461457 on Build GRCh38
rs1035900826 Chr.16: 57461167-57461167 on Build GRCh38
rs1012700499 Chr.16: 57461503-57461503 on Build GRCh38
rs115123016 Chr.16: 57461169-57461169 on Build GRCh38
rs995483837 Chr.16: 57461382-57461382 on Build GRCh38
rs886052184 Chr.16: 57461254-57461254 on Build GRCh38
rs898555130 Chr.16: 57461369-57461369 on Build GRCh38
rs558706588 Chr.16: 57461566-57461566 on Build GRCh38
rs779536722 Chr.16: 57461318-57461319 on Build GRCh38
rs942820078 Chr.16: 57461365-57461365 on Build GRCh38
rs1003197466 Chr.16: 57461442-57461442 on Build GRCh38

View Full Product Details